Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 154297863-154298370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT10
Entrez Gene ID
55568
Gene Name
polypeptide N-acetylgalactosaminyltransferase 10
Gene Aliases
GALNACT10, PPGALNACT10, PPGANTASE10
UniGene
Hs.631797
Gene Chromosome Location
Chr.5: 154190735-154420984 on Build GRCh38
Transcripts
NM_198321.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6293859 c.326G>A p.R109Q Chr.5: 154298004-154298004 on Build GRCh38
6349906 c.310G>C p.V104L Chr.5: 154297988-154297988 on Build GRCh38
1064713 c.296C>T p.T99I Chr.5: 154297974-154297974 on Build GRCh38
3612881 c.281G>A p.R94K Chr.5: 154297959-154297959 on Build GRCh38
6293858 c.326G>A p.R109Q Chr.5: 154298004-154298004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371894452 Chr.5: 154298099-154298099 on Build GRCh38
rs1054072887 Chr.5: 154298246-154298246 on Build GRCh38
rs1008835865 Chr.5: 154298224-154298224 on Build GRCh38
rs371591541 Chr.5: 154298026-154298026 on Build GRCh38
rs765066945 Chr.5: 154298108-154298108 on Build GRCh38
rs890227372 Chr.5: 154298057-154298057 on Build GRCh38
rs769212386 Chr.5: 154297893-154297893 on Build GRCh38
rs892309659 Chr.5: 154298258-154298258 on Build GRCh38
rs754420824 Chr.5: 154298030-154298030 on Build GRCh38
rs973621370 Chr.5: 154298072-154298072 on Build GRCh38
rs948764063 Chr.5: 154298196-154298196 on Build GRCh38
rs144763979 Chr.5: 154297920-154297920 on Build GRCh38
rs750157494 Chr.5: 154298021-154298021 on Build GRCh38
rs774367607 Chr.5: 154298082-154298082 on Build GRCh38
rs939642021 Chr.5: 154297984-154297984 on Build GRCh38
rs980212167 Chr.5: 154298214-154298214 on Build GRCh38
rs765038396 Chr.5: 154298004-154298004 on Build GRCh38
rs1011193491 Chr.5: 154298266-154298266 on Build GRCh38
rs1022377958 Chr.5: 154298294-154298294 on Build GRCh38
rs377043988 Chr.5: 154298121-154298121 on Build GRCh38
rs372500405 Chr.5: 154297904-154297904 on Build GRCh38
rs759683121 Chr.5: 154297979-154297979 on Build GRCh38
rs538296822 Chr.5: 154298069-154298069 on Build GRCh38
rs773329106 Chr.5: 154297914-154297914 on Build GRCh38
rs748567978 Chr.5: 154297976-154297976 on Build GRCh38
rs111468454 Chr.5: 154298191-154298191 on Build GRCh38
rs555089718 Chr.5: 154298090-154298090 on Build GRCh38
rs145894947 Chr.5: 154297990-154297990 on Build GRCh38
rs770477030 Chr.5: 154298277-154298277 on Build GRCh38
rs550238250 Chr.5: 154297901-154297901 on Build GRCh38
rs199678705 Chr.5: 154298089-154298089 on Build GRCh38
rs200119977 Chr.5: 154297898-154297898 on Build GRCh38
rs377201746 Chr.5: 154297905-154297905 on Build GRCh38
rs148586271 Chr.5: 154298176-154298176 on Build GRCh38
rs945176244 Chr.5: 154298285-154298285 on Build GRCh38
rs575034183 Chr.5: 154298104-154298104 on Build GRCh38
rs368289180 Chr.5: 154297892-154297892 on Build GRCh38
rs969372324 Chr.5: 154297960-154297960 on Build GRCh38
rs144259960 Chr.5: 154298027-154298027 on Build GRCh38
rs781674331 Chr.5: 154298034-154298034 on Build GRCh38
rs989595171 Chr.5: 154297919-154297919 on Build GRCh38
rs796265244 Chr.5: 154298229-154298229 on Build GRCh38
rs185108639 Chr.5: 154298177-154298177 on Build GRCh38
rs983330801 Chr.5: 154298023-154298023 on Build GRCh38
rs762388008 Chr.5: 154297895-154297905 on Build GRCh38
rs868753861 Chr.5: 154298162-154298162 on Build GRCh38
rs146580196 Chr.5: 154298044-154298044 on Build GRCh38
rs763598798 Chr.5: 154298092-154298095 on Build GRCh38
rs766487381 Chr.5: 154297975-154297975 on Build GRCh38
rs754327423 Chr.5: 154297891-154297891 on Build GRCh38
rs369164195 Chr.5: 154298067-154298067 on Build GRCh38
rs761877682 Chr.5: 154298107-154298107 on Build GRCh38
rs890355159 Chr.5: 154298194-154298194 on Build GRCh38
rs756983564 Chr.5: 154298070-154298070 on Build GRCh38
rs80134523 Chr.5: 154298243-154298243 on Build GRCh38
rs753767151 Chr.5: 154297987-154297987 on Build GRCh38
rs764129503 Chr.5: 154297986-154297986 on Build GRCh38
rs757136989 Chr.5: 154298001-154298001 on Build GRCh38
rs950913184 Chr.5: 154298133-154298133 on Build GRCh38
rs936027924 Chr.5: 154298245-154298246 on Build GRCh38
rs779659631 Chr.5: 154297890-154297890 on Build GRCh38
rs745447672 Chr.5: 154298333-154298333 on Build GRCh38

View Full Product Details