Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 154818737-154819240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAXDC2
Entrez Gene ID
10826
Gene Name
fatty acid hydroxylase domain containing 2
Gene Aliases
C5orf4
UniGene
Hs.519694
Gene Chromosome Location
Chr.5: 154817018-154850685 on Build GRCh38
Transcripts
NM_032385.4 XM_005268358.2 XM_006714753.2 XM_017008965.1 XM_017008966.1

SNP Detail

SNP ID Chromosome Location
rs916675951 Chr.5: 154818790-154818790 on Build GRCh38
rs1052674479 Chr.5: 154818965-154818965 on Build GRCh38
rs1014774956 Chr.5: 154819162-154819162 on Build GRCh38
rs1034437308 Chr.5: 154818872-154818872 on Build GRCh38
rs775036738 Chr.5: 154818906-154818906 on Build GRCh38
rs910996004 Chr.5: 154819033-154819033 on Build GRCh38
rs141201234 Chr.5: 154819217-154819217 on Build GRCh38
rs971885300 Chr.5: 154819109-154819109 on Build GRCh38
rs771358989 Chr.5: 154818949-154818949 on Build GRCh38
rs185230440 Chr.5: 154819089-154819089 on Build GRCh38
rs919569814 Chr.5: 154818950-154818950 on Build GRCh38
rs976917824 Chr.5: 154819067-154819067 on Build GRCh38
rs763350403 Chr.5: 154819016-154819017 on Build GRCh38
rs903997068 Chr.5: 154818915-154818915 on Build GRCh38
rs959182820 Chr.5: 154818891-154818891 on Build GRCh38
rs1016109791 Chr.5: 154819069-154819069 on Build GRCh38
rs904531608 Chr.5: 154819116-154819116 on Build GRCh38
rs867197893 Chr.5: 154819111-154819111 on Build GRCh38
rs997618312 Chr.5: 154819031-154819031 on Build GRCh38
rs773824004 Chr.5: 154818808-154818808 on Build GRCh38
rs557343148 Chr.5: 154818933-154818933 on Build GRCh38
rs148220263 Chr.5: 154818902-154818902 on Build GRCh38
rs992587376 Chr.5: 154819153-154819153 on Build GRCh38
rs537449634 Chr.5: 154818928-154818928 on Build GRCh38
rs963419157 Chr.5: 154818785-154818785 on Build GRCh38
rs758996114 Chr.5: 154819141-154819141 on Build GRCh38
rs11955563 Chr.5: 154818993-154818993 on Build GRCh38
rs545719500 Chr.5: 154819159-154819159 on Build GRCh38
rs1028638014 Chr.5: 154819038-154819038 on Build GRCh38
rs1013404274 Chr.5: 154818903-154818903 on Build GRCh38
rs937366465 Chr.5: 154819132-154819132 on Build GRCh38
rs902084775 Chr.5: 154819013-154819013 on Build GRCh38
rs1035637731 Chr.5: 154819134-154819134 on Build GRCh38
rs77231999 Chr.5: 154819136-154819136 on Build GRCh38
rs76047326 Chr.5: 154819211-154819211 on Build GRCh38
rs946279670 Chr.5: 154819096-154819096 on Build GRCh38
rs948139308 Chr.5: 154818819-154818819 on Build GRCh38
rs952389536 Chr.5: 154818956-154818956 on Build GRCh38
rs944172350 Chr.5: 154819056-154819056 on Build GRCh38
rs888878588 Chr.5: 154819040-154819040 on Build GRCh38
rs868135036 Chr.5: 154819070-154819070 on Build GRCh38
rs1043738274 Chr.5: 154819114-154819114 on Build GRCh38
rs985202285 Chr.5: 154818972-154818972 on Build GRCh38
rs117699984 Chr.5: 154819121-154819121 on Build GRCh38
rs1001567466 Chr.5: 154818864-154818864 on Build GRCh38

View Full Product Details