Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 156759472-156760012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SGCD
Entrez Gene ID
6444
Gene Name
sarcoglycan delta
Gene Aliases
35DAG, CMD1L, DAGD, SG-delta, SGCDP, SGD
UniGene
Hs.387207
Gene Chromosome Location
Chr.5: 155728636-156767788 on Build GRCh38
Transcripts
NM_000337.5 NM_001128209.1

SNP Detail

SNP ID Chromosome Location
rs750190359 Chr.5: 156759539-156759539 on Build GRCh38
rs911758552 Chr.5: 156759587-156759587 on Build GRCh38
rs182567689 Chr.5: 156759684-156759684 on Build GRCh38
rs886293325 Chr.5: 156759675-156759675 on Build GRCh38
rs184350278 Chr.5: 156759968-156759968 on Build GRCh38
rs542066860 Chr.5: 156759500-156759500 on Build GRCh38
rs192545549 Chr.5: 156759745-156759745 on Build GRCh38
rs552236592 Chr.5: 156759924-156759924 on Build GRCh38
rs187465743 Chr.5: 156759548-156759548 on Build GRCh38
rs930649939 Chr.5: 156759687-156759687 on Build GRCh38
rs763599917 Chr.5: 156759775-156759775 on Build GRCh38
rs368427648 Chr.5: 156759623-156759623 on Build GRCh38
rs562008848 Chr.5: 156759511-156759511 on Build GRCh38
rs954189649 Chr.5: 156759522-156759522 on Build GRCh38
rs1005464535 Chr.5: 156759538-156759538 on Build GRCh38
rs747715095 Chr.5: 156759829-156759829 on Build GRCh38
rs753750993 Chr.5: 156759715-156759715 on Build GRCh38
rs915149265 Chr.5: 156759581-156759582 on Build GRCh38
rs1001710737 Chr.5: 156759979-156759979 on Build GRCh38
rs907424872 Chr.5: 156759969-156759969 on Build GRCh38
rs890143848 Chr.5: 156759964-156759964 on Build GRCh38
rs966768608 Chr.5: 156759816-156759816 on Build GRCh38
rs200297937 Chr.5: 156759564-156759577 on Build GRCh38
rs1044317114 Chr.5: 156759585-156759585 on Build GRCh38
rs927237750 Chr.5: 156759642-156759642 on Build GRCh38
rs956204992 Chr.5: 156759779-156759779 on Build GRCh38
rs886060289 Chr.5: 156759947-156759947 on Build GRCh38
rs944557781 Chr.5: 156759842-156759842 on Build GRCh38
rs527466708 Chr.5: 156759857-156759857 on Build GRCh38
rs6886827 Chr.5: 156759753-156759753 on Build GRCh38
rs1008380599 Chr.5: 156759754-156759754 on Build GRCh38
rs72803040 Chr.5: 156759750-156759750 on Build GRCh38
rs190744152 Chr.5: 156759651-156759651 on Build GRCh38
rs886060290 Chr.5: 156759950-156759950 on Build GRCh38
rs147791150 Chr.5: 156759540-156759540 on Build GRCh38
rs886471923 Chr.5: 156759961-156759961 on Build GRCh38
rs1004562795 Chr.5: 156759701-156759701 on Build GRCh38
rs886436904 Chr.5: 156759724-156759724 on Build GRCh38
rs1050537304 Chr.5: 156759939-156759939 on Build GRCh38
rs898941748 Chr.5: 156759919-156759919 on Build GRCh38
rs886060287 Chr.5: 156759538-156759538 on Build GRCh38
rs771352762 Chr.5: 156759561-156759561 on Build GRCh38
rs934754578 Chr.5: 156759541-156759541 on Build GRCh38
rs927196473 Chr.5: 156759825-156759825 on Build GRCh38
rs558093663 Chr.5: 156759912-156759912 on Build GRCh38
rs989131493 Chr.5: 156759574-156759577 on Build GRCh38
rs532900623 Chr.5: 156759665-156759665 on Build GRCh38
rs947378407 Chr.5: 156759584-156759584 on Build GRCh38
rs886060288 Chr.5: 156759707-156759707 on Build GRCh38
rs1049605218 Chr.5: 156759934-156759934 on Build GRCh38
rs796973332 Chr.5: 156759512-156759512 on Build GRCh38
rs766108625 Chr.5: 156759563-156759563 on Build GRCh38
rs1019769618 Chr.5: 156759755-156759755 on Build GRCh38
rs980540265 Chr.5: 156759817-156759817 on Build GRCh38
rs529998490 Chr.5: 156759780-156759780 on Build GRCh38
rs914681383 Chr.5: 156759781-156759781 on Build GRCh38
rs562383472 Chr.5: 156759552-156759561 on Build GRCh38
rs1008454349 Chr.5: 156759965-156759965 on Build GRCh38
rs578018465 Chr.5: 156759925-156759925 on Build GRCh38
rs188034931 Chr.5: 156759741-156759741 on Build GRCh38
rs758099486 Chr.5: 156759843-156759843 on Build GRCh38
rs566068016 Chr.5: 156759815-156759815 on Build GRCh38
rs535200914 Chr.5: 156759830-156759830 on Build GRCh38
rs898789804 Chr.5: 156759739-156759739 on Build GRCh38
rs564187200 Chr.5: 156759649-156759649 on Build GRCh38
rs1039115010 Chr.5: 156759661-156759661 on Build GRCh38
rs959316465 Chr.5: 156759571-156759571 on Build GRCh38
rs1037450730 Chr.5: 156759861-156759861 on Build GRCh38
rs922015401 Chr.5: 156759863-156759863 on Build GRCh38

View Full Product Details