Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157058141-157058647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAVCR1
Entrez Gene ID
26762
Gene Name
hepatitis A virus cellular receptor 1
Gene Aliases
CD365, HAVCR, HAVCR-1, KIM-1, KIM1, TIM, TIM-1, TIM1, TIMD-1, TIMD1
UniGene
Hs.129711
Gene Chromosome Location
Chr.5: 156979443-157059119 on Build GRCh38
Transcripts
NM_001173393.2 NM_001308156.1 NM_012206.3

SNP Detail

SNP ID Chromosome Location
rs754009656 Chr.5: 157058354-157058354 on Build GRCh38
rs34333511 Chr.5: 157058357-157058357 on Build GRCh38
rs533978352 Chr.5: 157058530-157058530 on Build GRCh38
rs976066017 Chr.5: 157058501-157058501 on Build GRCh38
rs916266922 Chr.5: 157058576-157058576 on Build GRCh38
rs1050729282 Chr.5: 157058432-157058432 on Build GRCh38
rs1035184226 Chr.5: 157058311-157058311 on Build GRCh38
rs939016328 Chr.5: 157058426-157058426 on Build GRCh38
rs113841144 Chr.5: 157058249-157058249 on Build GRCh38
rs769050727 Chr.5: 157058243-157058243 on Build GRCh38
rs188071094 Chr.5: 157058360-157058360 on Build GRCh38
rs1050682530 Chr.5: 157058577-157058577 on Build GRCh38
rs980912810 Chr.5: 157058548-157058548 on Build GRCh38
rs909398760 Chr.5: 157058417-157058417 on Build GRCh38
rs1036629888 Chr.5: 157058292-157058292 on Build GRCh38
rs894875947 Chr.5: 157058455-157058455 on Build GRCh38
rs564533001 Chr.5: 157058436-157058436 on Build GRCh38
rs549682453 Chr.5: 157058238-157058238 on Build GRCh38
rs745443004 Chr.5: 157058522-157058522 on Build GRCh38
rs530444854 Chr.5: 157058520-157058520 on Build GRCh38
rs891493263 Chr.5: 157058321-157058321 on Build GRCh38
rs560450410 Chr.5: 157058256-157058256 on Build GRCh38
rs572590807 Chr.5: 157058419-157058419 on Build GRCh38
rs896785476 Chr.5: 157058425-157058425 on Build GRCh38
rs762367151 Chr.5: 157058298-157058298 on Build GRCh38
rs67960962 Chr.5: 157058175-157058175 on Build GRCh38
rs187880583 Chr.5: 157058470-157058470 on Build GRCh38
rs940864513 Chr.5: 157058528-157058528 on Build GRCh38
rs542142630 Chr.5: 157058490-157058490 on Build GRCh38
rs149029649 Chr.5: 157058313-157058313 on Build GRCh38
rs772786497 Chr.5: 157058328-157058328 on Build GRCh38
rs984906425 Chr.5: 157058521-157058521 on Build GRCh38
rs34819868 Chr.5: 157058365-157058365 on Build GRCh38
rs759170452 Chr.5: 157058275-157058276 on Build GRCh38
rs930043924 Chr.5: 157058252-157058252 on Build GRCh38
rs923962045 Chr.5: 157058221-157058221 on Build GRCh38
rs192925104 Chr.5: 157058560-157058560 on Build GRCh38
rs951628115 Chr.5: 157058535-157058535 on Build GRCh38
rs932112204 Chr.5: 157058387-157058387 on Build GRCh38
rs972244005 Chr.5: 157058531-157058531 on Build GRCh38
rs927187624 Chr.5: 157058379-157058379 on Build GRCh38
rs200213701 Chr.5: 157058576-157058577 on Build GRCh38
rs35045030 Chr.5: 157058616-157058616 on Build GRCh38
rs990405841 Chr.5: 157058570-157058570 on Build GRCh38
rs547218030 Chr.5: 157058301-157058301 on Build GRCh38
rs1043494260 Chr.5: 157058592-157058592 on Build GRCh38
rs985343586 Chr.5: 157058391-157058391 on Build GRCh38
rs185319476 Chr.5: 157058339-157058339 on Build GRCh38
rs71576844 Chr.5: 157058357-157058365 on Build GRCh38
rs146230565 Chr.5: 157058537-157058537 on Build GRCh38
rs969457409 Chr.5: 157058546-157058546 on Build GRCh38
rs562014460 Chr.5: 157058581-157058581 on Build GRCh38
rs978076109 Chr.5: 157058215-157058215 on Build GRCh38
rs183793800 Chr.5: 157058424-157058424 on Build GRCh38
rs566942060 Chr.5: 157058302-157058302 on Build GRCh38
rs190867546 Chr.5: 157058348-157058348 on Build GRCh38
rs570724147 Chr.5: 157058571-157058571 on Build GRCh38
rs903703335 Chr.5: 157058468-157058468 on Build GRCh38
rs141852075 Chr.5: 157058534-157058534 on Build GRCh38
rs999357754 Chr.5: 157058609-157058609 on Build GRCh38
rs558621598 Chr.5: 157058552-157058552 on Build GRCh38
rs144625732 Chr.5: 157058330-157058330 on Build GRCh38
rs1022524675 Chr.5: 157058457-157058457 on Build GRCh38
rs746913223 Chr.5: 157058456-157058456 on Build GRCh38
rs957249413 Chr.5: 157058565-157058565 on Build GRCh38
rs890892702 Chr.5: 157058335-157058335 on Build GRCh38
rs922002115 Chr.5: 157058364-157058364 on Build GRCh38
rs917074696 Chr.5: 157058285-157058290 on Build GRCh38
rs114680098 Chr.5: 157058291-157058291 on Build GRCh38
rs531782280 Chr.5: 157058404-157058404 on Build GRCh38
rs1030927719 Chr.5: 157058483-157058483 on Build GRCh38
rs531654216 Chr.5: 157058342-157058342 on Build GRCh38
rs909337555 Chr.5: 157058525-157058525 on Build GRCh38
rs912879251 Chr.5: 157058429-157058429 on Build GRCh38
rs903650959 Chr.5: 157058607-157058607 on Build GRCh38
rs955565852 Chr.5: 157058164-157058164 on Build GRCh38
rs980303157 Chr.5: 157058377-157058377 on Build GRCh38
rs758412988 Chr.5: 157058506-157058506 on Build GRCh38
rs202228419 Chr.5: 157058252-157058253 on Build GRCh38
rs917984239 Chr.5: 157058366-157058366 on Build GRCh38
rs935908503 Chr.5: 157058420-157058420 on Build GRCh38
rs9313422 Chr.5: 157058359-157058359 on Build GRCh38
rs1049263212 Chr.5: 157058285-157058285 on Build GRCh38
rs772271535 Chr.5: 157058610-157058610 on Build GRCh38
rs901740793 Chr.5: 157058623-157058623 on Build GRCh38
rs561521553 Chr.5: 157058286-157058286 on Build GRCh38
rs548005305 Chr.5: 157058370-157058370 on Build GRCh38
rs575168872 Chr.5: 157058449-157058449 on Build GRCh38
rs113072142 Chr.5: 157058508-157058508 on Build GRCh38
rs915142707 Chr.5: 157058198-157058198 on Build GRCh38
rs867895017 Chr.5: 157058329-157058329 on Build GRCh38
rs968028609 Chr.5: 157058201-157058201 on Build GRCh38
rs547181269 Chr.5: 157058544-157058544 on Build GRCh38
rs913120647 Chr.5: 157058574-157058574 on Build GRCh38
rs1030980347 Chr.5: 157058352-157058352 on Build GRCh38
rs944749683 Chr.5: 157058222-157058222 on Build GRCh38
rs945690253 Chr.5: 157058430-157058430 on Build GRCh38

View Full Product Details