Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176955871-176956383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UIMC1
Entrez Gene ID
51720
Gene Name
ubiquitin interaction motif containing 1
Gene Aliases
RAP80, X2HRIP110
UniGene
Hs.232721
Gene Chromosome Location
Chr.5: 176905005-177022640 on Build GRCh38
Transcripts
NM_001199297.2 NM_001199298.1 NM_001317961.1 NM_016290.4 XM_005265930.2 XM_005265932.2 XM_005265933.2 XM_005265934.2 XM_005265935.2 XM_005265936.2 XM_006714871.2 XM_006714874.2 XM_011534568.2 XM_011534570.1 XM_017009574.1 XM_017009575.1 XM_017009576.1 XM_017009577.1 XM_017009578.1 XM_017009579.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4347116 c.1334G>T p.C445F Chr.5: 176955964-176955964 on Build GRCh38
6816555 c.1277C>T p.T426I Chr.5: 176956021-176956021 on Build GRCh38
3761129 c.1304C>T p.P435L Chr.5: 176955994-176955994 on Build GRCh38
1696790 c.1297C>T p.L433F Chr.5: 176956001-176956001 on Build GRCh38
3365998 c.1323A>G p.E441E Chr.5: 176955975-176955975 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs35502369 Chr.5: 176956273-176956274 on Build GRCh38
rs78803247 Chr.5: 176955918-176955918 on Build GRCh38
rs750251795 Chr.5: 176956047-176956047 on Build GRCh38
rs537182705 Chr.5: 176956225-176956225 on Build GRCh38
rs776013991 Chr.5: 176955936-176955937 on Build GRCh38
rs570207678 Chr.5: 176956214-176956214 on Build GRCh38
rs763578136 Chr.5: 176956000-176956000 on Build GRCh38
rs913381493 Chr.5: 176956240-176956240 on Build GRCh38
rs768501661 Chr.5: 176956201-176956201 on Build GRCh38
rs758143931 Chr.5: 176956057-176956057 on Build GRCh38
rs974268988 Chr.5: 176956258-176956258 on Build GRCh38
rs774857484 Chr.5: 176955961-176955961 on Build GRCh38
rs944676390 Chr.5: 176956089-176956089 on Build GRCh38
rs371814083 Chr.5: 176955964-176955964 on Build GRCh38
rs139925001 Chr.5: 176956226-176956226 on Build GRCh38
rs775517359 Chr.5: 176956092-176956092 on Build GRCh38
rs777882159 Chr.5: 176955948-176955948 on Build GRCh38
rs1040580506 Chr.5: 176956104-176956104 on Build GRCh38
rs769817391 Chr.5: 176955946-176955946 on Build GRCh38
rs368570037 Chr.5: 176956304-176956304 on Build GRCh38
rs145456332 Chr.5: 176956256-176956256 on Build GRCh38
rs765092987 Chr.5: 176956029-176956029 on Build GRCh38
rs756359226 Chr.5: 176956076-176956076 on Build GRCh38
rs892909240 Chr.5: 176956222-176956222 on Build GRCh38
rs950157405 Chr.5: 176956317-176956317 on Build GRCh38
rs553395523 Chr.5: 176956243-176956243 on Build GRCh38
rs998530846 Chr.5: 176956179-176956179 on Build GRCh38
rs761670554 Chr.5: 176956020-176956020 on Build GRCh38
rs575757047 Chr.5: 176956124-176956124 on Build GRCh38
rs910459087 Chr.5: 176956060-176956060 on Build GRCh38
rs763421633 Chr.5: 176955956-176955957 on Build GRCh38
rs184562493 Chr.5: 176956242-176956242 on Build GRCh38
rs752906450 Chr.5: 176956075-176956075 on Build GRCh38
rs771871630 Chr.5: 176955990-176955990 on Build GRCh38
rs1016817773 Chr.5: 176956168-176956168 on Build GRCh38
rs766144173 Chr.5: 176956073-176956073 on Build GRCh38
rs772817453 Chr.5: 176956278-176956278 on Build GRCh38
rs1054118978 Chr.5: 176956204-176956204 on Build GRCh38
rs190195470 Chr.5: 176956352-176956352 on Build GRCh38
rs113479574 Chr.5: 176956090-176956090 on Build GRCh38
rs543227132 Chr.5: 176956358-176956358 on Build GRCh38
rs557704745 Chr.5: 176956323-176956323 on Build GRCh38
rs569038553 Chr.5: 176955993-176955993 on Build GRCh38
rs954150865 Chr.5: 176956266-176956266 on Build GRCh38
rs770895540 Chr.5: 176956043-176956043 on Build GRCh38
rs200428142 Chr.5: 176955949-176955949 on Build GRCh38
rs1027055066 Chr.5: 176956259-176956259 on Build GRCh38
rs539725669 Chr.5: 176956311-176956311 on Build GRCh38
rs912574095 Chr.5: 176956298-176956298 on Build GRCh38
rs3733876 Chr.5: 176955994-176955994 on Build GRCh38
rs749319392 Chr.5: 176956087-176956087 on Build GRCh38
rs567606599 Chr.5: 176956228-176956228 on Build GRCh38
rs561596222 Chr.5: 176955955-176955955 on Build GRCh38
rs972022438 Chr.5: 176956233-176956233 on Build GRCh38
rs963711288 Chr.5: 176956187-176956187 on Build GRCh38
rs533434764 Chr.5: 176956128-176956128 on Build GRCh38
rs777724310 Chr.5: 176956086-176956086 on Build GRCh38
rs551734095 Chr.5: 176956162-176956162 on Build GRCh38
rs926196944 Chr.5: 176956255-176956255 on Build GRCh38
rs985859480 Chr.5: 176956297-176956297 on Build GRCh38
rs776191688 Chr.5: 176956008-176956008 on Build GRCh38
rs1003129019 Chr.5: 176956237-176956237 on Build GRCh38

View Full Product Details