Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177066041-177066545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF346
Entrez Gene ID
23567
Gene Name
zinc finger protein 346
Gene Aliases
JAZ, Zfp346
UniGene
Hs.484259
Gene Chromosome Location
Chr.5: 177022680-177081189 on Build GRCh38
Transcripts
NM_001308213.1 NM_001308214.1 NM_001308215.1 NM_001308216.1 NM_001308218.1 NM_001308219.1 NM_001308221.1 NM_001308223.1 NM_012279.3 NR_131773.1 NR_131774.1 XM_005265860.2 XM_005265864.3 XM_011534494.1 XM_011534495.2 XM_011534496.2 XM_017009294.1 XM_017009295.1 XM_017009296.1 XM_017009297.1 XM_017009299.1 XM_017009300.1 XM_017009301.1 XM_017009302.1 XM_017009303.1

SNP Detail

SNP ID Chromosome Location
rs767314383 Chr.5: 177066361-177066361 on Build GRCh38
rs543664386 Chr.5: 177066286-177066286 on Build GRCh38
rs891387584 Chr.5: 177066234-177066234 on Build GRCh38
rs1004927477 Chr.5: 177066104-177066104 on Build GRCh38
rs564667923 Chr.5: 177066204-177066204 on Build GRCh38
rs925673547 Chr.5: 177066395-177066395 on Build GRCh38
rs932883220 Chr.5: 177066134-177066134 on Build GRCh38
rs12655218 Chr.5: 177066481-177066481 on Build GRCh38
rs541954303 Chr.5: 177066078-177066078 on Build GRCh38
rs939746727 Chr.5: 177066464-177066464 on Build GRCh38
rs987310357 Chr.5: 177066251-177066251 on Build GRCh38
rs1022796097 Chr.5: 177066397-177066397 on Build GRCh38
rs561957706 Chr.5: 177066305-177066305 on Build GRCh38
rs898183084 Chr.5: 177066125-177066125 on Build GRCh38
rs547510697 Chr.5: 177066485-177066485 on Build GRCh38
rs6887893 Chr.5: 177066262-177066262 on Build GRCh38
rs527751937 Chr.5: 177066086-177066086 on Build GRCh38
rs571974898 Chr.5: 177066168-177066168 on Build GRCh38
rs1000982957 Chr.5: 177066324-177066324 on Build GRCh38
rs908387547 Chr.5: 177066435-177066435 on Build GRCh38
rs146915315 Chr.5: 177066321-177066321 on Build GRCh38
rs59157655 Chr.5: 177066105-177066105 on Build GRCh38
rs767646107 Chr.5: 177066424-177066424 on Build GRCh38
rs991658467 Chr.5: 177066478-177066478 on Build GRCh38
rs949536742 Chr.5: 177066520-177066521 on Build GRCh38
rs919578308 Chr.5: 177066499-177066499 on Build GRCh38
rs1032746216 Chr.5: 177066331-177066331 on Build GRCh38
rs545769331 Chr.5: 177066195-177066195 on Build GRCh38
rs796251696 Chr.5: 177066353-177066353 on Build GRCh38
rs939798439 Chr.5: 177066089-177066089 on Build GRCh38
rs752842774 Chr.5: 177066434-177066434 on Build GRCh38
rs750153448 Chr.5: 177066367-177066367 on Build GRCh38
rs981211363 Chr.5: 177066432-177066432 on Build GRCh38
rs996395829 Chr.5: 177066132-177066132 on Build GRCh38
rs991365095 Chr.5: 177066388-177066388 on Build GRCh38
rs1008365929 Chr.5: 177066261-177066261 on Build GRCh38

View Full Product Details