Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178122124-178122627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
N4BP3
Entrez Gene ID
23138
Gene Name
NEDD4 binding protein 3
Gene Aliases
LZTS4
UniGene
Hs.101761
Gene Chromosome Location
Chr.5: 178113506-178127034 on Build GRCh38
Transcripts
NM_015111.1 XM_006714834.3 XM_011534473.1 XM_011534474.1

SNP Detail

SNP ID Chromosome Location
rs528240958 Chr.5: 178122156-178122156 on Build GRCh38
rs766023745 Chr.5: 178122539-178122539 on Build GRCh38
rs545381444 Chr.5: 178122418-178122418 on Build GRCh38
rs1041296244 Chr.5: 178122374-178122374 on Build GRCh38
rs368853499 Chr.5: 178122168-178122168 on Build GRCh38
rs557388381 Chr.5: 178122575-178122575 on Build GRCh38
rs1003174135 Chr.5: 178122492-178122492 on Build GRCh38
rs1023248044 Chr.5: 178122251-178122251 on Build GRCh38
rs770160615 Chr.5: 178122239-178122239 on Build GRCh38
rs971910762 Chr.5: 178122328-178122328 on Build GRCh38
rs572879176 Chr.5: 178122579-178122579 on Build GRCh38
rs984586527 Chr.5: 178122190-178122190 on Build GRCh38
rs1000208965 Chr.5: 178122421-178122421 on Build GRCh38
rs957792026 Chr.5: 178122234-178122234 on Build GRCh38
rs532192663 Chr.5: 178122357-178122357 on Build GRCh38
rs1019500676 Chr.5: 178122428-178122428 on Build GRCh38
rs942650913 Chr.5: 178122607-178122607 on Build GRCh38
rs539294064 Chr.5: 178122571-178122571 on Build GRCh38
rs549019796 Chr.5: 178122442-178122442 on Build GRCh38
rs775933212 Chr.5: 178122278-178122278 on Build GRCh38
rs969001372 Chr.5: 178122491-178122491 on Build GRCh38
rs962157649 Chr.5: 178122295-178122295 on Build GRCh38
rs1031613535 Chr.5: 178122173-178122173 on Build GRCh38
rs559506946 Chr.5: 178122331-178122331 on Build GRCh38
rs893997228 Chr.5: 178122470-178122470 on Build GRCh38
rs567419154 Chr.5: 178122443-178122443 on Build GRCh38
rs564762113 Chr.5: 178122275-178122275 on Build GRCh38
rs776149494 Chr.5: 178122522-178122522 on Build GRCh38
rs145590210 Chr.5: 178122324-178122324 on Build GRCh38
rs896068077 Chr.5: 178122311-178122311 on Build GRCh38
rs760281878 Chr.5: 178122532-178122532 on Build GRCh38
rs1034677292 Chr.5: 178122512-178122512 on Build GRCh38
rs927902826 Chr.5: 178122294-178122294 on Build GRCh38
rs183336287 Chr.5: 178122150-178122150 on Build GRCh38
rs965212125 Chr.5: 178122454-178122454 on Build GRCh38
rs754788940 Chr.5: 178122592-178122592 on Build GRCh38
rs961970820 Chr.5: 178122521-178122521 on Build GRCh38
rs879831124 Chr.5: 178122290-178122290 on Build GRCh38
rs952801686 Chr.5: 178122541-178122541 on Build GRCh38
rs753517537 Chr.5: 178122547-178122547 on Build GRCh38
rs3812081 Chr.5: 178122333-178122333 on Build GRCh38
rs3812080 Chr.5: 178122490-178122490 on Build GRCh38
rs370011976 Chr.5: 178122355-178122355 on Build GRCh38
rs1009808257 Chr.5: 178122384-178122384 on Build GRCh38
rs892245769 Chr.5: 178122378-178122378 on Build GRCh38
rs373626232 Chr.5: 178122484-178122484 on Build GRCh38
rs115568608 Chr.5: 178122469-178122469 on Build GRCh38
rs911329977 Chr.5: 178122598-178122598 on Build GRCh38
rs942682226 Chr.5: 178122358-178122358 on Build GRCh38
rs1057263166 Chr.5: 178122287-178122287 on Build GRCh38
rs1048545395 Chr.5: 178122342-178122342 on Build GRCh38
rs1000556218 Chr.5: 178122348-178122348 on Build GRCh38
rs532412456 Chr.5: 178122314-178122314 on Build GRCh38
rs984168195 Chr.5: 178122597-178122597 on Build GRCh38
rs981955093 Chr.5: 178122281-178122281 on Build GRCh38

View Full Product Details