Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178238315-178238814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL23A1
Entrez Gene ID
91522
Gene Name
collagen type XXIII alpha 1 chain
Gene Aliases
-
UniGene
Hs.660026
Gene Chromosome Location
Chr.5: 178237616-178590555 on Build GRCh38
Transcripts
NM_173465.3 XM_006714933.3 XM_006714934.3 XM_006714935.3 XM_006714936.3 XM_011534688.2 XM_011534689.2 XM_011534690.2 XM_011534691.2 XM_011534692.2

SNP Detail

SNP ID Chromosome Location
rs763281163 Chr.5: 178238713-178238713 on Build GRCh38
rs1033987715 Chr.5: 178238363-178238363 on Build GRCh38
rs372951332 Chr.5: 178238783-178238783 on Build GRCh38
rs553580259 Chr.5: 178238452-178238452 on Build GRCh38
rs11740043 Chr.5: 178238531-178238531 on Build GRCh38
rs543726320 Chr.5: 178238545-178238545 on Build GRCh38
rs1033722523 Chr.5: 178238568-178238568 on Build GRCh38
rs937154715 Chr.5: 178238782-178238782 on Build GRCh38
rs775174143 Chr.5: 178238666-178238666 on Build GRCh38
rs541226510 Chr.5: 178238622-178238622 on Build GRCh38
rs1026533065 Chr.5: 178238430-178238430 on Build GRCh38
rs929473976 Chr.5: 178238498-178238498 on Build GRCh38
rs750350339 Chr.5: 178238741-178238741 on Build GRCh38
rs199586986 Chr.5: 178238661-178238661 on Build GRCh38
rs910526895 Chr.5: 178238462-178238462 on Build GRCh38
rs779307547 Chr.5: 178238696-178238696 on Build GRCh38
rs111977623 Chr.5: 178238408-178238408 on Build GRCh38
rs758435503 Chr.5: 178238745-178238745 on Build GRCh38
rs766347497 Chr.5: 178238710-178238715 on Build GRCh38
rs759545126 Chr.5: 178238750-178238751 on Build GRCh38
rs58562702 Chr.5: 178238553-178238553 on Build GRCh38
rs372303892 Chr.5: 178238739-178238739 on Build GRCh38
rs909405325 Chr.5: 178238472-178238472 on Build GRCh38
rs189000256 Chr.5: 178238546-178238546 on Build GRCh38
rs992160034 Chr.5: 178238416-178238416 on Build GRCh38
rs1544926 Chr.5: 178238341-178238341 on Build GRCh38
rs182297389 Chr.5: 178238431-178238431 on Build GRCh38
rs191281464 Chr.5: 178238337-178238337 on Build GRCh38
rs774759018 Chr.5: 178238728-178238728 on Build GRCh38
rs557802075 Chr.5: 178238511-178238511 on Build GRCh38
rs932949484 Chr.5: 178238529-178238529 on Build GRCh38
rs927203297 Chr.5: 178238753-178238753 on Build GRCh38
rs998685117 Chr.5: 178238601-178238601 on Build GRCh38
rs529419384 Chr.5: 178238752-178238752 on Build GRCh38
rs753654863 Chr.5: 178238736-178238736 on Build GRCh38
rs539022515 Chr.5: 178238490-178238490 on Build GRCh38
rs377130455 Chr.5: 178238740-178238740 on Build GRCh38
rs576377093 Chr.5: 178238530-178238530 on Build GRCh38
rs536084994 Chr.5: 178238439-178238439 on Build GRCh38
rs964946083 Chr.5: 178238711-178238711 on Build GRCh38
rs1026335728 Chr.5: 178238638-178238638 on Build GRCh38
rs1047116616 Chr.5: 178238516-178238516 on Build GRCh38
rs1052727370 Chr.5: 178238541-178238541 on Build GRCh38
rs201979754 Chr.5: 178238726-178238726 on Build GRCh38
rs773619714 Chr.5: 178238694-178238694 on Build GRCh38
rs1013562164 Chr.5: 178238611-178238611 on Build GRCh38
rs746614048 Chr.5: 178238671-178238671 on Build GRCh38
rs1002694912 Chr.5: 178238567-178238567 on Build GRCh38
rs1046578675 Chr.5: 178238727-178238727 on Build GRCh38
rs568779477 Chr.5: 178238750-178238750 on Build GRCh38
rs746587057 Chr.5: 178238689-178238690 on Build GRCh38
rs754555214 Chr.5: 178238773-178238773 on Build GRCh38
rs151176062 Chr.5: 178238731-178238731 on Build GRCh38
rs943099941 Chr.5: 178238483-178238483 on Build GRCh38
rs10061163 Chr.5: 178238730-178238730 on Build GRCh38
rs193029360 Chr.5: 178238664-178238664 on Build GRCh38
rs967228679 Chr.5: 178238425-178238425 on Build GRCh38
rs768195464 Chr.5: 178238680-178238680 on Build GRCh38
rs573752756 Chr.5: 178238589-178238589 on Build GRCh38
rs757542798 Chr.5: 178238550-178238550 on Build GRCh38
rs112715515 Chr.5: 178238772-178238772 on Build GRCh38
rs963347887 Chr.5: 178238669-178238669 on Build GRCh38
rs545766417 Chr.5: 178238665-178238665 on Build GRCh38
rs373060611 Chr.5: 178238432-178238432 on Build GRCh38
rs896578448 Chr.5: 178238591-178238591 on Build GRCh38
rs1034086518 Chr.5: 178238401-178238401 on Build GRCh38
rs1544925 Chr.5: 178238485-178238485 on Build GRCh38
rs776202726 Chr.5: 178238721-178238722 on Build GRCh38
rs1030549078 Chr.5: 178238616-178238616 on Build GRCh38
rs369876153 Chr.5: 178238749-178238749 on Build GRCh38
rs973920456 Chr.5: 178238709-178238709 on Build GRCh38
rs1016836886 Chr.5: 178238670-178238670 on Build GRCh38

View Full Product Details