Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 149943481-149943984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTUD7B
Entrez Gene ID
56957
Gene Name
OTU deubiquitinase 7B
Gene Aliases
CEZANNE, ZA20D1
UniGene
Hs.98322
Gene Chromosome Location
Chr.1: 149940330-150010766 on Build GRCh38
Transcripts
NM_020205.3 XM_011509782.1 XM_011509783.2 XM_011509784.2 XM_011509785.2 XM_011509786.1 XM_011509787.1 XM_011509788.1 XM_017001850.1 XM_017001851.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7138347 c.2514C>A p.L838L Chr.1: 149943875-149943875 on Build GRCh38
5032034 c.2469C>A p.Y823* Chr.1: 149943920-149943920 on Build GRCh38
1333764 c.2427C>A p.F809L Chr.1: 149943962-149943962 on Build GRCh38
895433 c.2486G>T p.R829M Chr.1: 149943903-149943903 on Build GRCh38
4186460 c.2467T>A p.Y823N Chr.1: 149943922-149943922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782175859 Chr.1: 149943828-149943828 on Build GRCh38
rs148751820 Chr.1: 149943744-149943744 on Build GRCh38
rs76406948 Chr.1: 149943784-149943784 on Build GRCh38
rs782314227 Chr.1: 149943887-149943887 on Build GRCh38
rs587646908 Chr.1: 149943741-149943741 on Build GRCh38
rs781882769 Chr.1: 149943945-149943945 on Build GRCh38
rs781802267 Chr.1: 149943932-149943932 on Build GRCh38
rs782579072 Chr.1: 149943814-149943814 on Build GRCh38
rs782658260 Chr.1: 149943852-149943852 on Build GRCh38
rs587757032 Chr.1: 149943567-149943567 on Build GRCh38
rs782429516 Chr.1: 149943935-149943935 on Build GRCh38
rs966427167 Chr.1: 149943656-149943656 on Build GRCh38
rs201775933 Chr.1: 149943927-149943927 on Build GRCh38
rs986806578 Chr.1: 149943810-149943810 on Build GRCh38
rs201730149 Chr.1: 149943808-149943808 on Build GRCh38
rs781888285 Chr.1: 149943841-149943842 on Build GRCh38
rs898346457 Chr.1: 149943597-149943597 on Build GRCh38
rs587690807 Chr.1: 149943875-149943875 on Build GRCh38
rs1036859435 Chr.1: 149943578-149943578 on Build GRCh38
rs377196376 Chr.1: 149943963-149943963 on Build GRCh38
rs782804183 Chr.1: 149943938-149943938 on Build GRCh38
rs1055012467 Chr.1: 149943924-149943924 on Build GRCh38
rs931159551 Chr.1: 149943914-149943914 on Build GRCh38
rs587609515 Chr.1: 149943653-149943653 on Build GRCh38
rs781954881 Chr.1: 149943873-149943873 on Build GRCh38
rs1013492701 Chr.1: 149943962-149943962 on Build GRCh38
rs587734358 Chr.1: 149943840-149943840 on Build GRCh38
rs199673670 Chr.1: 149943960-149943960 on Build GRCh38
rs990387060 Chr.1: 149943762-149943762 on Build GRCh38
rs3754041 Chr.1: 149943855-149943855 on Build GRCh38
rs145575715 Chr.1: 149943559-149943559 on Build GRCh38
rs1031818696 Chr.1: 149943709-149943709 on Build GRCh38
rs995423530 Chr.1: 149943599-149943599 on Build GRCh38
rs782091150 Chr.1: 149943894-149943894 on Build GRCh38
rs587647409 Chr.1: 149943627-149943627 on Build GRCh38
rs782036205 Chr.1: 149943888-149943888 on Build GRCh38
rs1013210269 Chr.1: 149943631-149943631 on Build GRCh38
rs891218617 Chr.1: 149943562-149943562 on Build GRCh38
rs782365014 Chr.1: 149943869-149943869 on Build GRCh38
rs200227851 Chr.1: 149943890-149943890 on Build GRCh38
rs1051157303 Chr.1: 149943535-149943535 on Build GRCh38
rs999268008 Chr.1: 149943679-149943679 on Build GRCh38
rs373566777 Chr.1: 149943813-149943813 on Build GRCh38
rs145240430 Chr.1: 149943629-149943629 on Build GRCh38
rs782411716 Chr.1: 149943839-149943839 on Build GRCh38

View Full Product Details