Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150440504-150441006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPRD2
Entrez Gene ID
23248
Gene Name
regulation of nuclear pre-mRNA domain containing 2
Gene Aliases
HSPC099, KIAA0460
UniGene
Hs.213666
Gene Chromosome Location
Chr.1: 150364111-150476566 on Build GRCh38
Transcripts
NM_001297673.1 NM_001297674.1 NM_015203.4 XM_005245033.2 XM_006711240.2 XM_011509344.2 XM_017000763.1 XM_017000764.1 XM_017000765.1 XM_017000766.1 XM_017000767.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4622094 c.390A>G p.R130R Chr.1: 150440977-150440977 on Build GRCh38
7190985 c.367A>G p.I123V Chr.1: 150440954-150440954 on Build GRCh38
895506 c.344C>A p.S115Y Chr.1: 150440931-150440931 on Build GRCh38
4489017 c.341C>T p.P114L Chr.1: 150440928-150440928 on Build GRCh38
895508 c.367A>C p.I123L Chr.1: 150440954-150440954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782474439 Chr.1: 150440923-150440923 on Build GRCh38
rs1038728587 Chr.1: 150440714-150440714 on Build GRCh38
rs1015614340 Chr.1: 150440846-150440846 on Build GRCh38
rs1004228621 Chr.1: 150440843-150440843 on Build GRCh38
rs371380127 Chr.1: 150440893-150440893 on Build GRCh38
rs587684845 Chr.1: 150440685-150440685 on Build GRCh38
rs926519221 Chr.1: 150440552-150440552 on Build GRCh38
rs995904797 Chr.1: 150440898-150440898 on Build GRCh38
rs936305752 Chr.1: 150440656-150440656 on Build GRCh38
rs891304802 Chr.1: 150440823-150440823 on Build GRCh38
rs782414134 Chr.1: 150440886-150440886 on Build GRCh38
rs587758329 Chr.1: 150440586-150440586 on Build GRCh38
rs782749988 Chr.1: 150440602-150440602 on Build GRCh38
rs587764597 Chr.1: 150440732-150440732 on Build GRCh38
rs376591259 Chr.1: 150440556-150440556 on Build GRCh38
rs997109773 Chr.1: 150440736-150440736 on Build GRCh38
rs781889876 Chr.1: 150440885-150440885 on Build GRCh38
rs200304078 Chr.1: 150440897-150440897 on Build GRCh38
rs187992728 Chr.1: 150440980-150440980 on Build GRCh38
rs373603615 Chr.1: 150440911-150440911 on Build GRCh38
rs1051664571 Chr.1: 150440813-150440813 on Build GRCh38
rs587631604 Chr.1: 150440621-150440621 on Build GRCh38
rs781901465 Chr.1: 150440812-150440812 on Build GRCh38
rs924920089 Chr.1: 150440634-150440634 on Build GRCh38
rs917761462 Chr.1: 150440598-150440598 on Build GRCh38

View Full Product Details