Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151000267-151000762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM63A
Entrez Gene ID
55793
Gene Name
family with sequence similarity 63 member A
Gene Aliases
-
UniGene
Hs.743952
Gene Chromosome Location
Chr.1: 150996737-151008378 on Build GRCh38
Transcripts
NM_001040217.2 NM_001163258.1 NM_001163259.1 NM_001163260.1 NM_001319998.1 NM_018379.4 XM_005245321.3 XM_005245325.3 XM_005245327.3 XM_005245328.3 XM_011509757.2 XM_011509759.2 XM_017001770.1 XM_017001771.1 XM_017001772.1 XM_017001773.1 XM_017001774.1 XM_017001775.1 XM_017001776.1 XM_017001777.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6459284 c.577-1G>A p.? Chr.1: 151000616-151000616 on Build GRCh38
5754192 c.879G>T p.Q293H Chr.1: 151000457-151000457 on Build GRCh38
4510165 c.684C>T p.V228V Chr.1: 151000508-151000508 on Build GRCh38
1600779 c.703C>T p.P235S Chr.1: 151000489-151000489 on Build GRCh38
6445299 c.667A>G p.T223A Chr.1: 151000525-151000525 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761131545 Chr.1: 151000405-151000405 on Build GRCh38
rs751605223 Chr.1: 151000467-151000467 on Build GRCh38
rs374000030 Chr.1: 151000502-151000502 on Build GRCh38
rs746474993 Chr.1: 151000487-151000488 on Build GRCh38
rs185287844 Chr.1: 151000435-151000435 on Build GRCh38
rs765512402 Chr.1: 151000442-151000442 on Build GRCh38
rs763180581 Chr.1: 151000554-151000554 on Build GRCh38
rs773669740 Chr.1: 151000411-151000411 on Build GRCh38
rs769593812 Chr.1: 151000543-151000543 on Build GRCh38
rs745734554 Chr.1: 151000739-151000739 on Build GRCh38
rs952973042 Chr.1: 151000402-151000402 on Build GRCh38
rs944178047 Chr.1: 151000348-151000348 on Build GRCh38
rs377369599 Chr.1: 151000600-151000600 on Build GRCh38
rs761639529 Chr.1: 151000423-151000423 on Build GRCh38
rs587683473 Chr.1: 151000625-151000626 on Build GRCh38
rs914153250 Chr.1: 151000672-151000672 on Build GRCh38
rs772931318 Chr.1: 151000380-151000380 on Build GRCh38
rs142093888 Chr.1: 151000520-151000520 on Build GRCh38
rs587607910 Chr.1: 151000388-151000388 on Build GRCh38
rs1057282686 Chr.1: 151000706-151000708 on Build GRCh38
rs982775139 Chr.1: 151000328-151000328 on Build GRCh38
rs776340100 Chr.1: 151000421-151000421 on Build GRCh38
rs912526647 Chr.1: 151000573-151000573 on Build GRCh38
rs587647573 Chr.1: 151000300-151000300 on Build GRCh38
rs369885475 Chr.1: 151000447-151000447 on Build GRCh38
rs905512460 Chr.1: 151000675-151000675 on Build GRCh38
rs778313011 Chr.1: 151000594-151000594 on Build GRCh38
rs200819371 Chr.1: 151000407-151000407 on Build GRCh38
rs751628181 Chr.1: 151000575-151000575 on Build GRCh38
rs965768225 Chr.1: 151000580-151000580 on Build GRCh38
rs587722196 Chr.1: 151000499-151000499 on Build GRCh38
rs1041442919 Chr.1: 151000424-151000424 on Build GRCh38
rs756622366 Chr.1: 151000593-151000593 on Build GRCh38
rs79757708 Chr.1: 151000465-151000465 on Build GRCh38
rs781028977 Chr.1: 151000648-151000648 on Build GRCh38
rs906864823 Chr.1: 151000451-151000451 on Build GRCh38
rs1035473497 Chr.1: 151000351-151000351 on Build GRCh38
rs139033003 Chr.1: 151000471-151000471 on Build GRCh38
rs905588647 Chr.1: 151000323-151000323 on Build GRCh38
rs937045293 Chr.1: 151000686-151000686 on Build GRCh38
rs947304548 Chr.1: 151000641-151000641 on Build GRCh38
rs767964781 Chr.1: 151000579-151000579 on Build GRCh38
rs933131379 Chr.1: 151000606-151000606 on Build GRCh38
rs758561720 Chr.1: 151000450-151000450 on Build GRCh38
rs377400244 Chr.1: 151000413-151000413 on Build GRCh38
rs1040356016 Chr.1: 151000696-151000696 on Build GRCh38
rs181181549 Chr.1: 151000365-151000365 on Build GRCh38
rs753260477 Chr.1: 151000590-151000590 on Build GRCh38
rs201450887 Chr.1: 151000631-151000631 on Build GRCh38
rs773035437 Chr.1: 151000650-151000650 on Build GRCh38
rs778224979 Chr.1: 151000485-151000485 on Build GRCh38
rs748526528 Chr.1: 151000479-151000479 on Build GRCh38
rs1018712012 Chr.1: 151000477-151000477 on Build GRCh38
rs897801950 Chr.1: 151000727-151000727 on Build GRCh38
rs960180009 Chr.1: 151000359-151000359 on Build GRCh38
rs755483762 Chr.1: 151000468-151000468 on Build GRCh38
rs140386498 Chr.1: 151000483-151000483 on Build GRCh38
rs762701457 Chr.1: 151000651-151000651 on Build GRCh38
rs1040196422 Chr.1: 151000356-151000356 on Build GRCh38
rs746508026 Chr.1: 151000510-151000510 on Build GRCh38
rs587716459 Chr.1: 151000398-151000398 on Build GRCh38
rs145467872 Chr.1: 151000537-151000537 on Build GRCh38
rs772928955 Chr.1: 151000544-151000544 on Build GRCh38
rs779727279 Chr.1: 151000610-151000610 on Build GRCh38
rs74899511 Chr.1: 151000626-151000626 on Build GRCh38
rs759655152 Chr.1: 151000577-151000577 on Build GRCh38
rs758175157 Chr.1: 151000607-151000607 on Build GRCh38
rs755192444 Chr.1: 151000338-151000338 on Build GRCh38
rs781213897 Chr.1: 151000626-151000626 on Build GRCh38
rs587758307 Chr.1: 151000680-151000680 on Build GRCh38
rs587665091 Chr.1: 151000389-151000389 on Build GRCh38
rs780512919 Chr.1: 151000636-151000636 on Build GRCh38
rs754553413 Chr.1: 151000586-151000588 on Build GRCh38
rs768518414 Chr.1: 151000519-151000519 on Build GRCh38
rs945571524 Chr.1: 151000715-151000715 on Build GRCh38
rs375127079 Chr.1: 151000647-151000647 on Build GRCh38
rs779453457 Chr.1: 151000504-151000504 on Build GRCh38
rs766455637 Chr.1: 151000453-151000453 on Build GRCh38
rs994335605 Chr.1: 151000396-151000396 on Build GRCh38
rs766598183 Chr.1: 151000569-151000569 on Build GRCh38
rs774997624 Chr.1: 151000410-151000410 on Build GRCh38

View Full Product Details