Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151165940-151166445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYSMD1
Entrez Gene ID
388695
Gene Name
LysM domain containing 1
Gene Aliases
SB145
UniGene
Hs.591482
Gene Chromosome Location
Chr.1: 151148496-151165948 on Build GRCh38
Transcripts
NM_001136543.1 NM_001204856.1 NM_024041.3 NR_037937.1 NM_001204848.1

Gene Symbol
SCNM1
Entrez Gene ID
79005
Gene Name
sodium channel modifier 1
Gene Aliases
-
UniGene
Hs.732060
Gene Chromosome Location
Chr.1: 151166022-151170297 on Build GRCh38
Transcripts
NM_001136543.1 NM_001204856.1 NM_024041.3 NR_037937.1 NM_001204848.1

Gene Symbol
TNFAIP8L2-SCNM1
Entrez Gene ID
100534012
Gene Name
TNFAIP8L2-SCNM1 readthrough
Gene Aliases
-
UniGene
Hs.732060
Gene Chromosome Location
Chr.1: 151156629-151170297 on Build GRCh38
Transcripts
NM_001136543.1 NM_001204856.1 NM_024041.3 NR_037937.1 NM_001204848.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
83926 c.34C>T p.Q12* Chr.1: 151166186-151166186 on Build GRCh38
423807 c.16G>A p.E6K Chr.1: 151166168-151166168 on Build GRCh38
6057871 c.33T>A p.S11R Chr.1: 151166185-151166185 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs587672161 Chr.1: 151166165-151166165 on Build GRCh38
rs774292599 Chr.1: 151166166-151166166 on Build GRCh38
rs149471504 Chr.1: 151166193-151166193 on Build GRCh38
rs41268542 Chr.1: 151166091-151166091 on Build GRCh38
rs988130024 Chr.1: 151165991-151165991 on Build GRCh38
rs902941042 Chr.1: 151166071-151166071 on Build GRCh38
rs937952961 Chr.1: 151166373-151166373 on Build GRCh38
rs757615674 Chr.1: 151166131-151166131 on Build GRCh38
rs1026495614 Chr.1: 151165978-151165978 on Build GRCh38
rs892856303 Chr.1: 151166296-151166296 on Build GRCh38
rs777729875 Chr.1: 151166260-151166260 on Build GRCh38
rs746946216 Chr.1: 151166315-151166315 on Build GRCh38
rs749078327 Chr.1: 151166164-151166164 on Build GRCh38
rs1038448236 Chr.1: 151166027-151166027 on Build GRCh38
rs1033560001 Chr.1: 151165972-151165972 on Build GRCh38
rs747407031 Chr.1: 151166262-151166262 on Build GRCh38
rs757014601 Chr.1: 151166227-151166227 on Build GRCh38
rs188622485 Chr.1: 151166133-151166133 on Build GRCh38
rs995030137 Chr.1: 151165977-151165977 on Build GRCh38
rs920594885 Chr.1: 151166419-151166419 on Build GRCh38
rs1054641156 Chr.1: 151166290-151166290 on Build GRCh38
rs781316998 Chr.1: 151166163-151166163 on Build GRCh38
rs1013095207 Chr.1: 151166307-151166307 on Build GRCh38
rs760321684 Chr.1: 151166182-151166182 on Build GRCh38
rs368258231 Chr.1: 151166207-151166207 on Build GRCh38
rs776190593 Chr.1: 151166189-151166189 on Build GRCh38
rs41268538 Chr.1: 151165971-151165971 on Build GRCh38
rs921036122 Chr.1: 151166043-151166043 on Build GRCh38
rs587706791 Chr.1: 151165984-151165984 on Build GRCh38
rs971550225 Chr.1: 151166351-151166351 on Build GRCh38
rs763782707 Chr.1: 151166184-151166184 on Build GRCh38
rs908573838 Chr.1: 151166122-151166122 on Build GRCh38
rs587711423 Chr.1: 151166360-151166360 on Build GRCh38
rs765955714 Chr.1: 151166133-151166133 on Build GRCh38
rs927897498 Chr.1: 151166093-151166093 on Build GRCh38
rs773024654 Chr.1: 151166330-151166330 on Build GRCh38
rs371542102 Chr.1: 151166126-151166126 on Build GRCh38
rs762743617 Chr.1: 151166109-151166109 on Build GRCh38
rs981418912 Chr.1: 151165998-151165998 on Build GRCh38
rs767153800 Chr.1: 151166113-151166113 on Build GRCh38
rs755178616 Chr.1: 151166154-151166154 on Build GRCh38
rs145204847 Chr.1: 151166179-151166179 on Build GRCh38
rs587607911 Chr.1: 151166015-151166015 on Build GRCh38
rs1003168458 Chr.1: 151166414-151166414 on Build GRCh38
rs759073059 Chr.1: 151166107-151166107 on Build GRCh38
rs886613170 Chr.1: 151166170-151166170 on Build GRCh38
rs908632300 Chr.1: 151166004-151166004 on Build GRCh38
rs778187403 Chr.1: 151166035-151166035 on Build GRCh38
rs974586993 Chr.1: 151166141-151166141 on Build GRCh38
rs768804837 Chr.1: 151166230-151166230 on Build GRCh38
rs368840640 Chr.1: 151166044-151166044 on Build GRCh38
rs745809770 Chr.1: 151166174-151166174 on Build GRCh38
rs376521754 Chr.1: 151166030-151166030 on Build GRCh38
rs587720532 Chr.1: 151166070-151166070 on Build GRCh38
rs913756092 Chr.1: 151166062-151166062 on Build GRCh38
rs375496414 Chr.1: 151166033-151166033 on Build GRCh38
rs758912220 Chr.1: 151166201-151166201 on Build GRCh38
rs1019653280 Chr.1: 151166188-151166188 on Build GRCh38
rs758106461 Chr.1: 151166239-151166239 on Build GRCh38
rs765748237 Chr.1: 151166132-151166132 on Build GRCh38
rs587647574 Chr.1: 151165983-151165983 on Build GRCh38
rs775651034 Chr.1: 151166078-151166078 on Build GRCh38
rs368171990 Chr.1: 151166114-151166114 on Build GRCh38
rs1015829238 Chr.1: 151166418-151166418 on Build GRCh38
rs1055022814 Chr.1: 151166395-151166395 on Build GRCh38
rs758424650 Chr.1: 151166138-151166138 on Build GRCh38
rs752885739 Chr.1: 151166216-151166216 on Build GRCh38
rs1018524605 Chr.1: 151166061-151166062 on Build GRCh38
rs138492136 Chr.1: 151166178-151166178 on Build GRCh38
rs994939087 Chr.1: 151166151-151166151 on Build GRCh38
rs766940557 Chr.1: 151166204-151166204 on Build GRCh38
rs750924428 Chr.1: 151166135-151166135 on Build GRCh38
rs758055242 Chr.1: 151166022-151166022 on Build GRCh38
rs781637566 Chr.1: 151166191-151166191 on Build GRCh38
rs772124955 Chr.1: 151166252-151166252 on Build GRCh38
rs587670289 Chr.1: 151166118-151166118 on Build GRCh38
rs763221375 Chr.1: 151166218-151166218 on Build GRCh38
rs1032758066 Chr.1: 151166077-151166077 on Build GRCh38
rs61819224 Chr.1: 151166145-151166145 on Build GRCh38
rs945174799 Chr.1: 151166075-151166075 on Build GRCh38
rs989162001 Chr.1: 151166049-151166049 on Build GRCh38
rs989193299 Chr.1: 151166209-151166209 on Build GRCh38
rs587756598 Chr.1: 151166074-151166075 on Build GRCh38
rs373468565 Chr.1: 151166214-151166214 on Build GRCh38
rs750872334 Chr.1: 151166200-151166200 on Build GRCh38
rs957550955 Chr.1: 151165974-151165974 on Build GRCh38
rs587766258 Chr.1: 151166423-151166423 on Build GRCh38

View Full Product Details