Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151171292-151171790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMOD4
Entrez Gene ID
29765
Gene Name
tropomodulin 4
Gene Aliases
SK-TMOD
UniGene
Hs.709681
Gene Chromosome Location
Chr.1: 151169986-151176071 on Build GRCh38
Transcripts
NM_013353.2 XM_011509449.1 XM_017001089.1 XM_017001090.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3801804 c.702G>A p.T234T Chr.1: 151171457-151171457 on Build GRCh38
3704966 c.719T>A p.I240N Chr.1: 151171440-151171440 on Build GRCh38
4552623 c.564G>C p.K188N Chr.1: 151171687-151171687 on Build GRCh38
76768 c.710G>T p.G237V Chr.1: 151171449-151171449 on Build GRCh38
3360326 c.722C>T p.A241V Chr.1: 151171437-151171437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762604847 Chr.1: 151171391-151171391 on Build GRCh38
rs1038618363 Chr.1: 151171368-151171368 on Build GRCh38
rs1049846538 Chr.1: 151171654-151171654 on Build GRCh38
rs771592520 Chr.1: 151171571-151171571 on Build GRCh38
rs752407659 Chr.1: 151171550-151171550 on Build GRCh38
rs753754893 Chr.1: 151171647-151171647 on Build GRCh38
rs587748712 Chr.1: 151171707-151171707 on Build GRCh38
rs35954676 Chr.1: 151171484-151171484 on Build GRCh38
rs771590010 Chr.1: 151171653-151171653 on Build GRCh38
rs746686213 Chr.1: 151171679-151171679 on Build GRCh38
rs191901748 Chr.1: 151171712-151171712 on Build GRCh38
rs745314320 Chr.1: 151171661-151171661 on Build GRCh38
rs775238840 Chr.1: 151171576-151171576 on Build GRCh38
rs1008215105 Chr.1: 151171697-151171697 on Build GRCh38
rs749238997 Chr.1: 151171557-151171557 on Build GRCh38
rs772838854 Chr.1: 151171389-151171389 on Build GRCh38
rs905609538 Chr.1: 151171317-151171317 on Build GRCh38
rs764544131 Chr.1: 151171505-151171505 on Build GRCh38
rs774166003 Chr.1: 151171399-151171399 on Build GRCh38
rs770575570 Chr.1: 151171468-151171468 on Build GRCh38
rs587719901 Chr.1: 151171551-151171551 on Build GRCh38
rs757023935 Chr.1: 151171562-151171562 on Build GRCh38
rs377541747 Chr.1: 151171729-151171729 on Build GRCh38
rs779506311 Chr.1: 151171668-151171668 on Build GRCh38
rs756096828 Chr.1: 151171440-151171441 on Build GRCh38
rs201060059 Chr.1: 151171730-151171730 on Build GRCh38
rs779601964 Chr.1: 151171744-151171744 on Build GRCh38
rs750379295 Chr.1: 151171732-151171732 on Build GRCh38
rs772792859 Chr.1: 151171722-151171722 on Build GRCh38
rs587632353 Chr.1: 151171592-151171592 on Build GRCh38
rs779044470 Chr.1: 151171651-151171651 on Build GRCh38
rs751815869 Chr.1: 151171631-151171631 on Build GRCh38
rs1018352182 Chr.1: 151171476-151171476 on Build GRCh38
rs765307311 Chr.1: 151171543-151171543 on Build GRCh38
rs587610354 Chr.1: 151171607-151171607 on Build GRCh38
rs937193128 Chr.1: 151171331-151171331 on Build GRCh38
rs761071797 Chr.1: 151171407-151171407 on Build GRCh38
rs201200128 Chr.1: 151171728-151171728 on Build GRCh38
rs774078060 Chr.1: 151171479-151171479 on Build GRCh38
rs764709604 Chr.1: 151171408-151171408 on Build GRCh38
rs1001373081 Chr.1: 151171734-151171734 on Build GRCh38
rs757957617 Chr.1: 151171666-151171666 on Build GRCh38
rs761140788 Chr.1: 151171501-151171501 on Build GRCh38
rs765046154 Chr.1: 151171731-151171731 on Build GRCh38
rs376286863 Chr.1: 151171395-151171395 on Build GRCh38
rs757787722 Chr.1: 151171736-151171736 on Build GRCh38
rs757231199 Chr.1: 151171648-151171648 on Build GRCh38
rs189151097 Chr.1: 151171600-151171600 on Build GRCh38
rs937075562 Chr.1: 151171588-151171588 on Build GRCh38
rs761824810 Chr.1: 151171541-151171541 on Build GRCh38
rs757358819 Chr.1: 151171433-151171433 on Build GRCh38
rs759836935 Chr.1: 151171724-151171724 on Build GRCh38
rs753884241 Chr.1: 151171425-151171425 on Build GRCh38
rs777145671 Chr.1: 151171522-151171522 on Build GRCh38
rs769600169 Chr.1: 151171458-151171458 on Build GRCh38
rs373235083 Chr.1: 151171680-151171680 on Build GRCh38
rs74423006 Chr.1: 151171610-151171610 on Build GRCh38
rs771059810 Chr.1: 151171718-151171718 on Build GRCh38
rs1032280831 Chr.1: 151171766-151171766 on Build GRCh38
rs201393339 Chr.1: 151171545-151171545 on Build GRCh38
rs767775802 Chr.1: 151171403-151171403 on Build GRCh38
rs751690480 Chr.1: 151171525-151171525 on Build GRCh38
rs750659321 Chr.1: 151171440-151171440 on Build GRCh38
rs748754198 Chr.1: 151171465-151171465 on Build GRCh38
rs373463476 Chr.1: 151171552-151171552 on Build GRCh38
rs759103510 Chr.1: 151171480-151171480 on Build GRCh38
rs748548849 Chr.1: 151171685-151171685 on Build GRCh38
rs755918408 Chr.1: 151171632-151171632 on Build GRCh38
rs148768361 Chr.1: 151171561-151171561 on Build GRCh38
rs28730722 Chr.1: 151171494-151171494 on Build GRCh38
rs773787438 Chr.1: 151171692-151171692 on Build GRCh38
rs780847706 Chr.1: 151171454-151171454 on Build GRCh38
rs754737684 Chr.1: 151171452-151171452 on Build GRCh38
rs898288011 Chr.1: 151171388-151171388 on Build GRCh38
rs763970307 Chr.1: 151171634-151171634 on Build GRCh38
rs751134490 Chr.1: 151171751-151171751 on Build GRCh38
rs770261287 Chr.1: 151171687-151171687 on Build GRCh38
rs28730723 Chr.1: 151171581-151171581 on Build GRCh38
rs777902772 Chr.1: 151171464-151171464 on Build GRCh38
rs142974975 Chr.1: 151171764-151171764 on Build GRCh38
rs753170644 Chr.1: 151171404-151171404 on Build GRCh38
rs748036938 Chr.1: 151171457-151171457 on Build GRCh38
rs779059293 Chr.1: 151171434-151171434 on Build GRCh38
rs766675922 Chr.1: 151171546-151171546 on Build GRCh38
rs587732992 Chr.1: 151171582-151171582 on Build GRCh38
rs199965373 Chr.1: 151171673-151171673 on Build GRCh38
rs200980248 Chr.1: 151171599-151171599 on Build GRCh38
rs754591355 Chr.1: 151171756-151171756 on Build GRCh38

View Full Product Details