Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151579527-151580028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUFT1
Entrez Gene ID
7286
Gene Name
tuftelin 1
Gene Aliases
-
UniGene
Hs.489922
Gene Chromosome Location
Chr.1: 151540294-151583583 on Build GRCh38
Transcripts
NM_001126337.1 NM_001301317.1 NM_020127.2 XM_011509961.2 XM_017002223.1 XM_017002224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
206032 c.961G>A p.A321T Chr.1: 151579685-151579685 on Build GRCh38
5559249 c.927C>G p.L309L Chr.1: 151579651-151579651 on Build GRCh38
5962497 c.967A>C p.I323L Chr.1: 151579691-151579691 on Build GRCh38
3984223 c.939A>G p.K313K Chr.1: 151579663-151579663 on Build GRCh38
895876 c.990C>T p.I330I Chr.1: 151579714-151579714 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs893762911 Chr.1: 151579797-151579797 on Build GRCh38
rs201062061 Chr.1: 151579663-151579663 on Build GRCh38
rs753135518 Chr.1: 151579624-151579624 on Build GRCh38
rs966841474 Chr.1: 151579564-151579564 on Build GRCh38
rs908019563 Chr.1: 151579692-151579692 on Build GRCh38
rs920851271 Chr.1: 151579653-151579653 on Build GRCh38
rs966632153 Chr.1: 151580004-151580004 on Build GRCh38
rs79895644 Chr.1: 151579948-151579948 on Build GRCh38
rs764501647 Chr.1: 151579597-151579597 on Build GRCh38
rs775968818 Chr.1: 151579782-151579782 on Build GRCh38
rs551302442 Chr.1: 151579840-151579840 on Build GRCh38
rs372102721 Chr.1: 151579746-151579746 on Build GRCh38
rs766087803 Chr.1: 151579598-151579598 on Build GRCh38
rs781081327 Chr.1: 151579689-151579689 on Build GRCh38
rs751260466 Chr.1: 151579611-151579611 on Build GRCh38
rs566967029 Chr.1: 151579812-151579812 on Build GRCh38
rs1027607411 Chr.1: 151579961-151579961 on Build GRCh38
rs748737870 Chr.1: 151579735-151579735 on Build GRCh38
rs760966802 Chr.1: 151579784-151579784 on Build GRCh38
rs767738977 Chr.1: 151579781-151579781 on Build GRCh38
rs888685988 Chr.1: 151579846-151579846 on Build GRCh38
rs964488012 Chr.1: 151579929-151579929 on Build GRCh38
rs1021200327 Chr.1: 151579555-151579555 on Build GRCh38
rs3748606 Chr.1: 151579896-151579896 on Build GRCh38
rs757143636 Chr.1: 151579570-151579570 on Build GRCh38
rs762643211 Chr.1: 151579825-151579826 on Build GRCh38
rs756680826 Chr.1: 151579627-151579627 on Build GRCh38
rs952353084 Chr.1: 151579971-151579971 on Build GRCh38
rs913148716 Chr.1: 151579589-151579589 on Build GRCh38
rs114673006 Chr.1: 151579806-151579806 on Build GRCh38
rs533732809 Chr.1: 151579982-151579982 on Build GRCh38
rs3833527 Chr.1: 151579826-151579826 on Build GRCh38
rs1049389838 Chr.1: 151579788-151579788 on Build GRCh38
rs188445385 Chr.1: 151579622-151579622 on Build GRCh38
rs773056626 Chr.1: 151579717-151579717 on Build GRCh38
rs902356799 Chr.1: 151579810-151579810 on Build GRCh38
rs998246614 Chr.1: 151579943-151579943 on Build GRCh38
rs1035039028 Chr.1: 151579593-151579593 on Build GRCh38
rs3748607 Chr.1: 151579816-151579816 on Build GRCh38
rs764012537 Chr.1: 151579762-151579766 on Build GRCh38
rs757336286 Chr.1: 151579668-151579668 on Build GRCh38
rs6587604 Chr.1: 151580003-151580003 on Build GRCh38
rs150367520 Chr.1: 151579618-151579618 on Build GRCh38
rs1018419479 Chr.1: 151579898-151579898 on Build GRCh38
rs959412375 Chr.1: 151579939-151579939 on Build GRCh38
rs182383164 Chr.1: 151579798-151579798 on Build GRCh38
rs199983603 Chr.1: 151579826-151579826 on Build GRCh38
rs759327891 Chr.1: 151579760-151579760 on Build GRCh38
rs754728864 Chr.1: 151579685-151579685 on Build GRCh38
rs775416644 Chr.1: 151579603-151579604 on Build GRCh38
rs746150897 Chr.1: 151579684-151579684 on Build GRCh38
rs900901907 Chr.1: 151579750-151579750 on Build GRCh38
rs748013747 Chr.1: 151579696-151579696 on Build GRCh38
rs369587294 Chr.1: 151579628-151579628 on Build GRCh38
rs779053056 Chr.1: 151579670-151579670 on Build GRCh38
rs984097469 Chr.1: 151579973-151579973 on Build GRCh38
rs770577589 Chr.1: 151579741-151579741 on Build GRCh38
rs535598564 Chr.1: 151579716-151579716 on Build GRCh38
rs956744267 Chr.1: 151579970-151579970 on Build GRCh38
rs760371144 Chr.1: 151579666-151579667 on Build GRCh38
rs988648942 Chr.1: 151579584-151579584 on Build GRCh38
rs769723174 Chr.1: 151579715-151579715 on Build GRCh38
rs968591508 Chr.1: 151579999-151579999 on Build GRCh38

View Full Product Details