Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180848594-180849095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP62
Entrez Gene ID
643836
Gene Name
ZFP62 zinc finger protein
Gene Aliases
ZET, ZNF755
UniGene
Hs.509227
Gene Chromosome Location
Chr.5: 180843580-180861327 on Build GRCh38
Transcripts
NM_001172638.1 NM_152283.4 XM_017009710.1 XM_017009711.1 XM_017009712.1 XM_017009713.1 XM_017009714.1 XM_017009715.1 XM_017009716.1 XM_017009717.1 XM_017009718.1 XM_017009719.1 XM_017009720.1 XM_017009721.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6203131 c.2514A>G p.I838M Chr.5: 180848882-180848882 on Build GRCh38
3941278 c.2430T>A p.N810K Chr.5: 180848966-180848966 on Build GRCh38
3012692 c.2302A>G p.I768V Chr.5: 180849094-180849094 on Build GRCh38
6439398 c.2522G>A p.G841E Chr.5: 180848874-180848874 on Build GRCh38
7398562 c.2422C>G p.R808G Chr.5: 180848974-180848974 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867727058 Chr.5: 180848859-180848859 on Build GRCh38
rs1054733436 Chr.5: 180849062-180849062 on Build GRCh38
rs893729028 Chr.5: 180848993-180848993 on Build GRCh38
rs748384863 Chr.5: 180848953-180848953 on Build GRCh38
rs895569688 Chr.5: 180848620-180848620 on Build GRCh38
rs375571790 Chr.5: 180849016-180849016 on Build GRCh38
rs763774336 Chr.5: 180849068-180849068 on Build GRCh38
rs199749521 Chr.5: 180848946-180848946 on Build GRCh38
rs928838368 Chr.5: 180848826-180848826 on Build GRCh38
rs537351323 Chr.5: 180848751-180848751 on Build GRCh38
rs943813461 Chr.5: 180848967-180848967 on Build GRCh38
rs754806051 Chr.5: 180848981-180848983 on Build GRCh38
rs755768339 Chr.5: 180848867-180848867 on Build GRCh38
rs138860378 Chr.5: 180848913-180848913 on Build GRCh38
rs1023350121 Chr.5: 180848767-180848767 on Build GRCh38
rs747120287 Chr.5: 180848937-180848937 on Build GRCh38
rs190457380 Chr.5: 180848825-180848825 on Build GRCh38
rs749086741 Chr.5: 180848976-180848976 on Build GRCh38
rs568957381 Chr.5: 180848841-180848841 on Build GRCh38
rs187156768 Chr.5: 180848792-180848792 on Build GRCh38
rs182004779 Chr.5: 180848791-180848791 on Build GRCh38
rs765716971 Chr.5: 180848892-180848892 on Build GRCh38
rs774337643 Chr.5: 180848988-180848988 on Build GRCh38
rs566298652 Chr.5: 180848783-180848783 on Build GRCh38
rs957933720 Chr.5: 180848749-180848749 on Build GRCh38
rs1053550453 Chr.5: 180848983-180848983 on Build GRCh38
rs557910310 Chr.5: 180848719-180848719 on Build GRCh38
rs1041166952 Chr.5: 180848968-180848968 on Build GRCh38
rs920208733 Chr.5: 180848876-180848876 on Build GRCh38
rs770832115 Chr.5: 180848984-180848984 on Build GRCh38
rs1019450932 Chr.5: 180848701-180848701 on Build GRCh38
rs865885441 Chr.5: 180848897-180848897 on Build GRCh38
rs974825533 Chr.5: 180848824-180848824 on Build GRCh38
rs149065023 Chr.5: 180848973-180848973 on Build GRCh38
rs779704548 Chr.5: 180848768-180848769 on Build GRCh38
rs767025559 Chr.5: 180848794-180848795 on Build GRCh38
rs372196874 Chr.5: 180848637-180848637 on Build GRCh38
rs999415327 Chr.5: 180848726-180848726 on Build GRCh38
rs755047865 Chr.5: 180848943-180848943 on Build GRCh38
rs759476785 Chr.5: 180848990-180848990 on Build GRCh38
rs150247342 Chr.5: 180848896-180848896 on Build GRCh38
rs376564487 Chr.5: 180848815-180848815 on Build GRCh38
rs752398826 Chr.5: 180848864-180848864 on Build GRCh38
rs972883870 Chr.5: 180848844-180848844 on Build GRCh38
rs905636872 Chr.5: 180849057-180849057 on Build GRCh38
rs930996365 Chr.5: 180848634-180848634 on Build GRCh38
rs775088620 Chr.5: 180848998-180848998 on Build GRCh38
rs778533584 Chr.5: 180848975-180848975 on Build GRCh38
rs1012545889 Chr.5: 180849005-180849005 on Build GRCh38
rs762827722 Chr.5: 180848813-180848813 on Build GRCh38
rs751079925 Chr.5: 180848817-180848817 on Build GRCh38
rs767145674 Chr.5: 180848991-180848991 on Build GRCh38
rs940981889 Chr.5: 180848917-180848917 on Build GRCh38
rs571491304 Chr.5: 180848652-180848652 on Build GRCh38
rs1000490947 Chr.5: 180848709-180848709 on Build GRCh38
rs749068291 Chr.5: 180848885-180848886 on Build GRCh38
rs190771865 Chr.5: 180848721-180848721 on Build GRCh38
rs931551063 Chr.5: 180848882-180848882 on Build GRCh38
rs568543524 Chr.5: 180848957-180848957 on Build GRCh38
rs1008351962 Chr.5: 180848692-180848692 on Build GRCh38
rs780592915 Chr.5: 180848915-180848915 on Build GRCh38
rs1000013042 Chr.5: 180849058-180849058 on Build GRCh38
rs544019288 Chr.5: 180848683-180848683 on Build GRCh38
rs966083431 Chr.5: 180848779-180848779 on Build GRCh38
rs760438009 Chr.5: 180849056-180849056 on Build GRCh38
rs1044136825 Chr.5: 180849052-180849052 on Build GRCh38
rs1022980700 Chr.5: 180849020-180849020 on Build GRCh38
rs773004018 Chr.5: 180848766-180848766 on Build GRCh38
rs571317705 Chr.5: 180848737-180848737 on Build GRCh38
rs767341735 Chr.5: 180848851-180848851 on Build GRCh38
rs902251157 Chr.5: 180848969-180848969 on Build GRCh38
rs530519938 Chr.5: 180848656-180848656 on Build GRCh38
rs898730291 Chr.5: 180848625-180848625 on Build GRCh38

View Full Product Details