Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181199769-181200275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM7
Entrez Gene ID
81786
Gene Name
tripartite motif containing 7
Gene Aliases
GNIP, RNF90
UniGene
Hs.487412
Gene Chromosome Location
Chr.5: 181193924-181206000 on Build GRCh38
Transcripts
NM_203293.2 NM_203294.1 NM_203295.1 NM_203296.1 NM_203297.1 XM_017009903.1 XM_017009904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3715054 c.728G>A p.R243Q Chr.5: 181199972-181199972 on Build GRCh38
6808561 c.151C>T p.R51W Chr.5: 181200003-181200003 on Build GRCh38
1066920 c.709C>A p.R237S Chr.5: 181199991-181199991 on Build GRCh38
4874312 c.163C>A p.R55S Chr.5: 181199991-181199991 on Build GRCh38
6535848 c.279A>C p.Q93H Chr.5: 181199875-181199875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745457733 Chr.5: 181199935-181199935 on Build GRCh38
rs766622255 Chr.5: 181200132-181200132 on Build GRCh38
rs150528571 Chr.5: 181200020-181200020 on Build GRCh38
rs115623830 Chr.5: 181200223-181200223 on Build GRCh38
rs777718999 Chr.5: 181199930-181199930 on Build GRCh38
rs761813342 Chr.5: 181200167-181200167 on Build GRCh38
rs2546397 Chr.5: 181200224-181200224 on Build GRCh38
rs910433771 Chr.5: 181199847-181199847 on Build GRCh38
rs61743103 Chr.5: 181200048-181200048 on Build GRCh38
rs565643389 Chr.5: 181199813-181199813 on Build GRCh38
rs1000549430 Chr.5: 181199910-181199910 on Build GRCh38
rs756137955 Chr.5: 181199990-181199990 on Build GRCh38
rs773469993 Chr.5: 181200110-181200111 on Build GRCh38
rs767964081 Chr.5: 181200172-181200172 on Build GRCh38
rs569307799 Chr.5: 181199972-181199972 on Build GRCh38
rs760908037 Chr.5: 181200032-181200032 on Build GRCh38
rs751236589 Chr.5: 181200179-181200179 on Build GRCh38
rs770411661 Chr.5: 181199826-181199826 on Build GRCh38
rs199628505 Chr.5: 181200111-181200111 on Build GRCh38
rs774408562 Chr.5: 181199950-181199951 on Build GRCh38
rs368145514 Chr.5: 181200067-181200067 on Build GRCh38
rs774647409 Chr.5: 181199969-181199969 on Build GRCh38
rs202024254 Chr.5: 181200171-181200171 on Build GRCh38
rs776389750 Chr.5: 181200154-181200154 on Build GRCh38
rs973459789 Chr.5: 181200207-181200207 on Build GRCh38
rs61737740 Chr.5: 181199899-181199899 on Build GRCh38
rs764254563 Chr.5: 181200149-181200149 on Build GRCh38
rs753573355 Chr.5: 181199991-181199991 on Build GRCh38
rs200756291 Chr.5: 181200010-181200010 on Build GRCh38
rs750602893 Chr.5: 181199866-181199866 on Build GRCh38
rs752619688 Chr.5: 181199979-181199979 on Build GRCh38
rs200292025 Chr.5: 181199814-181199814 on Build GRCh38
rs748983180 Chr.5: 181199801-181199801 on Build GRCh38
rs778690711 Chr.5: 181199932-181199932 on Build GRCh38
rs769447717 Chr.5: 181199944-181199944 on Build GRCh38
rs770269863 Chr.5: 181200012-181200012 on Build GRCh38
rs368389149 Chr.5: 181200204-181200204 on Build GRCh38
rs78866624 Chr.5: 181200174-181200174 on Build GRCh38
rs445864 Chr.5: 181200250-181200250 on Build GRCh38
rs1032862038 Chr.5: 181199917-181199917 on Build GRCh38
rs958548562 Chr.5: 181199918-181199918 on Build GRCh38
rs929371157 Chr.5: 181199941-181199941 on Build GRCh38
rs199763714 Chr.5: 181199829-181199829 on Build GRCh38
rs751589361 Chr.5: 181199890-181199890 on Build GRCh38
rs781254469 Chr.5: 181199897-181199897 on Build GRCh38
rs765108080 Chr.5: 181200054-181200054 on Build GRCh38
rs141070470 Chr.5: 181200023-181200023 on Build GRCh38
rs756949813 Chr.5: 181200049-181200049 on Build GRCh38
rs141530115 Chr.5: 181200002-181200002 on Build GRCh38
rs763206244 Chr.5: 181200015-181200015 on Build GRCh38
rs762023403 Chr.5: 181200075-181200076 on Build GRCh38
rs760812156 Chr.5: 181200123-181200123 on Build GRCh38
rs749704803 Chr.5: 181200194-181200194 on Build GRCh38
rs780758669 Chr.5: 181200191-181200191 on Build GRCh38
rs775701997 Chr.5: 181200028-181200028 on Build GRCh38
rs775898229 Chr.5: 181200119-181200119 on Build GRCh38
rs779543676 Chr.5: 181200007-181200007 on Build GRCh38
rs373704693 Chr.5: 181200022-181200022 on Build GRCh38
rs771134825 Chr.5: 181200087-181200087 on Build GRCh38
rs368553710 Chr.5: 181199836-181199836 on Build GRCh38
rs771530729 Chr.5: 181200005-181200005 on Build GRCh38
rs867856489 Chr.5: 181199895-181199895 on Build GRCh38
rs373948376 Chr.5: 181199834-181199834 on Build GRCh38
rs761059110 Chr.5: 181200112-181200113 on Build GRCh38
rs749975908 Chr.5: 181200055-181200055 on Build GRCh38
rs61739211 Chr.5: 181199973-181199973 on Build GRCh38
rs560728652 Chr.5: 181200110-181200110 on Build GRCh38
rs767720176 Chr.5: 181200089-181200091 on Build GRCh38
rs755127201 Chr.5: 181199896-181199896 on Build GRCh38
rs745551797 Chr.5: 181200004-181200004 on Build GRCh38
rs920588278 Chr.5: 181200233-181200233 on Build GRCh38
rs766481173 Chr.5: 181199960-181199960 on Build GRCh38
rs764088743 Chr.5: 181200042-181200042 on Build GRCh38
rs777861379 Chr.5: 181200216-181200216 on Build GRCh38
rs999840662 Chr.5: 181200117-181200117 on Build GRCh38
rs777070029 Chr.5: 181199943-181199943 on Build GRCh38
rs916866561 Chr.5: 181200251-181200251 on Build GRCh38
rs748916038 Chr.5: 181199931-181199931 on Build GRCh38
rs559432527 Chr.5: 181200013-181200013 on Build GRCh38
rs1014988367 Chr.5: 181200217-181200217 on Build GRCh38
rs939462356 Chr.5: 181199974-181199974 on Build GRCh38
rs544149971 Chr.5: 181200076-181200076 on Build GRCh38
rs754533784 Chr.5: 181200074-181200074 on Build GRCh38
rs774795162 Chr.5: 181200092-181200092 on Build GRCh38
rs1047928151 Chr.5: 181199964-181199964 on Build GRCh38
rs754445437 Chr.5: 181200183-181200183 on Build GRCh38
rs546493756 Chr.5: 181200112-181200112 on Build GRCh38
rs416574 Chr.5: 181199927-181199927 on Build GRCh38
rs1031341541 Chr.5: 181200151-181200151 on Build GRCh38
rs61743096 Chr.5: 181200027-181200027 on Build GRCh38
rs768777214 Chr.5: 181199940-181199940 on Build GRCh38
rs757113095 Chr.5: 181199992-181199992 on Build GRCh38
rs749832431 Chr.5: 181200085-181200085 on Build GRCh38
rs890176791 Chr.5: 181199872-181199872 on Build GRCh38
rs999006865 Chr.5: 181200090-181200090 on Build GRCh38
rs763431044 Chr.5: 181199954-181199954 on Build GRCh38
rs188706920 Chr.5: 181200213-181200213 on Build GRCh38
rs376856007 Chr.5: 181199885-181199885 on Build GRCh38
rs765612355 Chr.5: 181199861-181199861 on Build GRCh38
rs758060160 Chr.5: 181200060-181200060 on Build GRCh38
rs767692247 Chr.5: 181200212-181200212 on Build GRCh38
rs201559417 Chr.5: 181200068-181200068 on Build GRCh38
rs777159857 Chr.5: 181199835-181199835 on Build GRCh38

View Full Product Details