Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 485399-485940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC2
Entrez Gene ID
55770
Gene Name
exocyst complex component 2
Gene Aliases
SEC5, SEC5L1, Sec5p
UniGene
Hs.484412
Gene Chromosome Location
Chr.6: 485138-693141 on Build GRCh38
Transcripts
NM_018303.5 NR_073064.1 XM_017011018.1 XM_017011020.1 XM_017011021.1

SNP Detail

SNP ID Chromosome Location
rs975243159 Chr.6: 485915-485915 on Build GRCh38
rs7765038 Chr.6: 485896-485896 on Build GRCh38
rs781443235 Chr.6: 485667-485667 on Build GRCh38
rs1034551293 Chr.6: 485744-485744 on Build GRCh38
rs374927446 Chr.6: 485686-485686 on Build GRCh38
rs1002217226 Chr.6: 485742-485742 on Build GRCh38
rs927277477 Chr.6: 485446-485446 on Build GRCh38
rs978175452 Chr.6: 485511-485511 on Build GRCh38
rs904661425 Chr.6: 485725-485725 on Build GRCh38
rs962573571 Chr.6: 485773-485773 on Build GRCh38
rs1021917782 Chr.6: 485585-485585 on Build GRCh38
rs960063395 Chr.6: 485750-485750 on Build GRCh38
rs541451527 Chr.6: 485464-485464 on Build GRCh38
rs1043625150 Chr.6: 485709-485709 on Build GRCh38
rs754314108 Chr.6: 485767-485767 on Build GRCh38
rs370392852 Chr.6: 485625-485625 on Build GRCh38
rs964256419 Chr.6: 485913-485913 on Build GRCh38
rs535363278 Chr.6: 485885-485885 on Build GRCh38
rs761412896 Chr.6: 485608-485608 on Build GRCh38
rs537683856 Chr.6: 485830-485830 on Build GRCh38
rs957516144 Chr.6: 485468-485468 on Build GRCh38
rs1022037487 Chr.6: 485469-485470 on Build GRCh38
rs999716764 Chr.6: 485881-485881 on Build GRCh38
rs938776614 Chr.6: 485456-485456 on Build GRCh38
rs960011577 Chr.6: 485597-485597 on Build GRCh38
rs753277758 Chr.6: 485855-485855 on Build GRCh38
rs557109729 Chr.6: 485831-485831 on Build GRCh38
rs571359953 Chr.6: 485600-485600 on Build GRCh38
rs918443357 Chr.6: 485478-485478 on Build GRCh38
rs980509174 Chr.6: 485434-485434 on Build GRCh38
rs929797231 Chr.6: 485483-485483 on Build GRCh38
rs918662617 Chr.6: 485607-485607 on Build GRCh38
rs892219927 Chr.6: 485671-485671 on Build GRCh38
rs968784317 Chr.6: 485429-485429 on Build GRCh38
rs145201339 Chr.6: 485803-485803 on Build GRCh38
rs933634793 Chr.6: 485666-485666 on Build GRCh38
rs534671709 Chr.6: 485786-485786 on Build GRCh38
rs911124484 Chr.6: 485737-485737 on Build GRCh38
rs947596715 Chr.6: 485854-485854 on Build GRCh38
rs1039598277 Chr.6: 485634-485634 on Build GRCh38
rs946471534 Chr.6: 485836-485836 on Build GRCh38
rs543056880 Chr.6: 485516-485516 on Build GRCh38
rs11242914 Chr.6: 485586-485586 on Build GRCh38
rs984191040 Chr.6: 485780-485780 on Build GRCh38
rs909723426 Chr.6: 485791-485791 on Build GRCh38
rs376878223 Chr.6: 485822-485822 on Build GRCh38
rs549447612 Chr.6: 485644-485644 on Build GRCh38
rs62390392 Chr.6: 485789-485789 on Build GRCh38
rs796837286 Chr.6: 485826-485826 on Build GRCh38
rs951387805 Chr.6: 485890-485890 on Build GRCh38
rs896074345 Chr.6: 485875-485875 on Build GRCh38
rs572542146 Chr.6: 485483-485484 on Build GRCh38
rs922506853 Chr.6: 485815-485815 on Build GRCh38
rs777750806 Chr.6: 485893-485893 on Build GRCh38
rs922270835 Chr.6: 485519-485519 on Build GRCh38
rs1034778074 Chr.6: 485596-485596 on Build GRCh38
rs569979436 Chr.6: 485858-485858 on Build GRCh38
rs985366916 Chr.6: 485656-485656 on Build GRCh38
rs115301227 Chr.6: 485814-485814 on Build GRCh38
rs1025610233 Chr.6: 485886-485886 on Build GRCh38
rs1022282591 Chr.6: 485424-485424 on Build GRCh38
rs922289205 Chr.6: 485654-485654 on Build GRCh38
rs942591365 Chr.6: 485533-485533 on Build GRCh38
rs369179892 Chr.6: 485526-485526 on Build GRCh38
rs1020437560 Chr.6: 485615-485615 on Build GRCh38
rs140825671 Chr.6: 485839-485839 on Build GRCh38
rs1048339793 Chr.6: 485521-485521 on Build GRCh38
rs527620707 Chr.6: 485560-485560 on Build GRCh38
rs114884914 Chr.6: 485849-485849 on Build GRCh38
rs181349916 Chr.6: 485561-485561 on Build GRCh38
rs1025559260 Chr.6: 485768-485768 on Build GRCh38
rs748178731 Chr.6: 485810-485810 on Build GRCh38
rs1039782691 Chr.6: 485536-485536 on Build GRCh38
rs140409368 Chr.6: 485563-485563 on Build GRCh38
rs6597054 Chr.6: 485620-485620 on Build GRCh38
rs1029750492 Chr.6: 485626-485626 on Build GRCh38
rs1001818633 Chr.6: 485594-485594 on Build GRCh38
rs984362287 Chr.6: 485613-485613 on Build GRCh38
rs888357277 Chr.6: 485523-485523 on Build GRCh38
rs185040459 Chr.6: 485587-485587 on Build GRCh38
rs535168530 Chr.6: 485847-485847 on Build GRCh38
rs951553195 Chr.6: 485775-485775 on Build GRCh38
rs1010815227 Chr.6: 485573-485573 on Build GRCh38
rs566073051 Chr.6: 485697-485697 on Build GRCh38
rs561343010 Chr.6: 485538-485538 on Build GRCh38
rs1014423165 Chr.6: 485880-485880 on Build GRCh38
rs779405645 Chr.6: 485475-485476 on Build GRCh38

View Full Product Details