Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 3152680-3153214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPHL
Entrez Gene ID
670
Gene Name
biphenyl hydrolase-like (serine hydrolase)
Gene Aliases
BPH-RP, MCNAA, VACVASE
UniGene
Hs.10136
Gene Chromosome Location
Chr.6: 3118376-3153198 on Build GRCh38
Transcripts
NM_001302777.1 NM_004332.3 NR_026648.2 NR_026649.2

SNP Detail

SNP ID Chromosome Location
rs1018774484 Chr.6: 3152872-3152872 on Build GRCh38
rs575215749 Chr.6: 3152736-3152736 on Build GRCh38
rs960748157 Chr.6: 3153056-3153056 on Build GRCh38
rs1017079359 Chr.6: 3152961-3152961 on Build GRCh38
rs983431939 Chr.6: 3152972-3152972 on Build GRCh38
rs925192176 Chr.6: 3153165-3153165 on Build GRCh38
rs564060258 Chr.6: 3152903-3152903 on Build GRCh38
rs140628685 Chr.6: 3153079-3153079 on Build GRCh38
rs112913728 Chr.6: 3152892-3152892 on Build GRCh38
rs3088000 Chr.6: 3153149-3153149 on Build GRCh38
rs962873566 Chr.6: 3152981-3152981 on Build GRCh38
rs146883942 Chr.6: 3152856-3152856 on Build GRCh38
rs58044191 Chr.6: 3152958-3152958 on Build GRCh38
rs1014896300 Chr.6: 3152994-3152994 on Build GRCh38
rs552750387 Chr.6: 3153024-3153024 on Build GRCh38
rs947854613 Chr.6: 3153046-3153047 on Build GRCh38
rs935215054 Chr.6: 3153066-3153066 on Build GRCh38
rs1046097003 Chr.6: 3152759-3152759 on Build GRCh38
rs879600976 Chr.6: 3153059-3153059 on Build GRCh38
rs1043632500 Chr.6: 3152784-3152784 on Build GRCh38
rs1027896645 Chr.6: 3152948-3152948 on Build GRCh38
rs11553273 Chr.6: 3152768-3152768 on Build GRCh38
rs915200450 Chr.6: 3153088-3153088 on Build GRCh38
rs1010908268 Chr.6: 3152863-3152864 on Build GRCh38
rs767894571 Chr.6: 3153139-3153139 on Build GRCh38
rs1137422 Chr.6: 3152947-3152947 on Build GRCh38
rs916687727 Chr.6: 3153097-3153097 on Build GRCh38
rs376268563 Chr.6: 3152729-3152730 on Build GRCh38
rs916307692 Chr.6: 3153034-3153034 on Build GRCh38
rs771013886 Chr.6: 3152893-3152893 on Build GRCh38
rs941729774 Chr.6: 3152738-3152738 on Build GRCh38
rs569377016 Chr.6: 3153046-3153046 on Build GRCh38
rs1056201352 Chr.6: 3152755-3152755 on Build GRCh38
rs982162667 Chr.6: 3153047-3153047 on Build GRCh38
rs1052541030 Chr.6: 3152826-3152826 on Build GRCh38
rs568298251 Chr.6: 3153000-3153000 on Build GRCh38
rs532628238 Chr.6: 3153020-3153020 on Build GRCh38
rs1137421 Chr.6: 3152919-3152919 on Build GRCh38
rs570675712 Chr.6: 3153167-3153167 on Build GRCh38
rs973157735 Chr.6: 3152743-3152743 on Build GRCh38
rs545529419 Chr.6: 3152708-3152708 on Build GRCh38
rs927665276 Chr.6: 3153064-3153064 on Build GRCh38
rs541428793 Chr.6: 3152795-3152795 on Build GRCh38
rs998297463 Chr.6: 3153164-3153164 on Build GRCh38
rs531255452 Chr.6: 3152761-3152761 on Build GRCh38
rs935007575 Chr.6: 3152814-3152814 on Build GRCh38
rs1018064394 Chr.6: 3152902-3152902 on Build GRCh38
rs991694994 Chr.6: 3153028-3153029 on Build GRCh38
rs1039570060 Chr.6: 3153104-3153104 on Build GRCh38
rs1060332 Chr.6: 3153143-3153143 on Build GRCh38
rs570588189 Chr.6: 3152967-3152967 on Build GRCh38
rs1052191786 Chr.6: 3152855-3152855 on Build GRCh38
rs887112376 Chr.6: 3153187-3153187 on Build GRCh38
rs112175280 Chr.6: 3152854-3152854 on Build GRCh38
rs939440030 Chr.6: 3152751-3152751 on Build GRCh38
rs1052085510 Chr.6: 3153173-3153173 on Build GRCh38
rs947919484 Chr.6: 3152769-3152769 on Build GRCh38
rs996785588 Chr.6: 3152936-3152936 on Build GRCh38

View Full Product Details