Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 10694868-10695381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAK1IP1
Entrez Gene ID
55003
Gene Name
PAK1 interacting protein 1
Gene Aliases
MAK11, PIP1, WDR84, bA421M1.5, hPIP1
UniGene
Hs.310231
Gene Chromosome Location
Chr.6: 10690865-10709782 on Build GRCh38
Transcripts
NM_017906.2 XM_005249204.2 XM_011514720.1 XM_011514721.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7428028 c.12C>G p.V4V Chr.6: 10694997-10694997 on Build GRCh38
4593449 c.71G>T p.C24F Chr.6: 10695056-10695056 on Build GRCh38
7382254 c.23A>T p.Y8F Chr.6: 10695008-10695008 on Build GRCh38
5615579 c.50T>C p.V17A Chr.6: 10695035-10695035 on Build GRCh38
4160100 c.36C>T p.L12L Chr.6: 10695021-10695021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984230461 Chr.6: 10695187-10695187 on Build GRCh38
rs981597737 Chr.6: 10695071-10695071 on Build GRCh38
rs757088413 Chr.6: 10694998-10694998 on Build GRCh38
rs774651899 Chr.6: 10695047-10695047 on Build GRCh38
rs969865451 Chr.6: 10695136-10695136 on Build GRCh38
rs750289338 Chr.6: 10695101-10695101 on Build GRCh38
rs1005060014 Chr.6: 10695247-10695247 on Build GRCh38
rs973806996 Chr.6: 10695270-10695270 on Build GRCh38
rs976956303 Chr.6: 10695200-10695201 on Build GRCh38
rs7745856 Chr.6: 10694925-10694925 on Build GRCh38
rs915603855 Chr.6: 10695271-10695271 on Build GRCh38
rs1052911299 Chr.6: 10695274-10695274 on Build GRCh38
rs576058557 Chr.6: 10694909-10694909 on Build GRCh38
rs879133674 Chr.6: 10694910-10694910 on Build GRCh38
rs373902982 Chr.6: 10694903-10694903 on Build GRCh38
rs948622352 Chr.6: 10694904-10694904 on Build GRCh38
rs756176747 Chr.6: 10695013-10695013 on Build GRCh38
rs367606250 Chr.6: 10694983-10694983 on Build GRCh38
rs907018391 Chr.6: 10694907-10694907 on Build GRCh38
rs941000440 Chr.6: 10694900-10694900 on Build GRCh38
rs187280543 Chr.6: 10695293-10695293 on Build GRCh38
rs893224839 Chr.6: 10695145-10695145 on Build GRCh38
rs542686333 Chr.6: 10695085-10695085 on Build GRCh38
rs934487641 Chr.6: 10695118-10695118 on Build GRCh38
rs900487670 Chr.6: 10695215-10695215 on Build GRCh38
rs776433994 Chr.6: 10694943-10694944 on Build GRCh38
rs759418854 Chr.6: 10695083-10695086 on Build GRCh38
rs753916583 Chr.6: 10695106-10695106 on Build GRCh38
rs201605878 Chr.6: 10695081-10695081 on Build GRCh38
rs1017519858 Chr.6: 10694913-10694913 on Build GRCh38
rs566669973 Chr.6: 10694908-10694908 on Build GRCh38
rs932934447 Chr.6: 10695220-10695220 on Build GRCh38
rs747048551 Chr.6: 10695092-10695093 on Build GRCh38
rs752263745 Chr.6: 10694981-10694981 on Build GRCh38
rs764725295 Chr.6: 10694976-10694976 on Build GRCh38
rs182635087 Chr.6: 10694955-10694955 on Build GRCh38
rs749505397 Chr.6: 10694938-10694938 on Build GRCh38
rs1011540721 Chr.6: 10694911-10694911 on Build GRCh38
rs761160420 Chr.6: 10695056-10695056 on Build GRCh38
rs751272379 Chr.6: 10694997-10694997 on Build GRCh38
rs1053163750 Chr.6: 10695087-10695087 on Build GRCh38
rs767866626 Chr.6: 10694956-10694956 on Build GRCh38
rs371781516 Chr.6: 10695010-10695010 on Build GRCh38
rs908543408 Chr.6: 10695194-10695195 on Build GRCh38
rs368640685 Chr.6: 10695078-10695078 on Build GRCh38
rs775778036 Chr.6: 10695241-10695241 on Build GRCh38
rs922887716 Chr.6: 10695218-10695218 on Build GRCh38
rs922773533 Chr.6: 10694973-10694973 on Build GRCh38
rs894397474 Chr.6: 10694903-10694903 on Build GRCh38
rs376490072 Chr.6: 10694923-10694924 on Build GRCh38
rs1011593206 Chr.6: 10695152-10695152 on Build GRCh38
rs749417108 Chr.6: 10695037-10695037 on Build GRCh38
rs369312896 Chr.6: 10695053-10695054 on Build GRCh38
rs145531831 Chr.6: 10695234-10695234 on Build GRCh38
rs758025593 Chr.6: 10694984-10694984 on Build GRCh38
rs928552156 Chr.6: 10695304-10695304 on Build GRCh38
rs952868903 Chr.6: 10695162-10695162 on Build GRCh38
rs775777762 Chr.6: 10694936-10694936 on Build GRCh38
rs908230315 Chr.6: 10694892-10694892 on Build GRCh38
rs543412749 Chr.6: 10694912-10694912 on Build GRCh38
rs1038327513 Chr.6: 10694901-10694901 on Build GRCh38
rs997800121 Chr.6: 10695227-10695227 on Build GRCh38
rs905755169 Chr.6: 10695298-10695298 on Build GRCh38
rs376060945 Chr.6: 10695015-10695015 on Build GRCh38
rs878881986 Chr.6: 10694904-10694905 on Build GRCh38
rs1030526585 Chr.6: 10694948-10694948 on Build GRCh38
rs7759621 Chr.6: 10695255-10695255 on Build GRCh38
rs202109461 Chr.6: 10694923-10694923 on Build GRCh38
rs891450127 Chr.6: 10695280-10695280 on Build GRCh38
rs368480648 Chr.6: 10695076-10695076 on Build GRCh38
rs755102835 Chr.6: 10695115-10695115 on Build GRCh38
rs915535382 Chr.6: 10695023-10695023 on Build GRCh38
rs1027508145 Chr.6: 10695151-10695151 on Build GRCh38
rs965007610 Chr.6: 10694934-10694934 on Build GRCh38
rs767552712 Chr.6: 10695109-10695110 on Build GRCh38
rs774544435 Chr.6: 10694945-10694945 on Build GRCh38
rs199615995 Chr.6: 10694944-10694944 on Build GRCh38
rs778939360 Chr.6: 10695116-10695116 on Build GRCh38
rs772493979 Chr.6: 10695051-10695051 on Build GRCh38
rs1039022820 Chr.6: 10695198-10695198 on Build GRCh38
rs550189154 Chr.6: 10695178-10695178 on Build GRCh38
rs867273616 Chr.6: 10695030-10695030 on Build GRCh38
rs751228814 Chr.6: 10695083-10695083 on Build GRCh38
rs1020499762 Chr.6: 10694924-10694924 on Build GRCh38
rs982565372 Chr.6: 10694957-10694957 on Build GRCh38
rs1045438594 Chr.6: 10695295-10695295 on Build GRCh38
rs767328103 Chr.6: 10695090-10695090 on Build GRCh38
rs970126475 Chr.6: 10695280-10695280 on Build GRCh38
rs772452069 Chr.6: 10695156-10695156 on Build GRCh38
rs768716813 Chr.6: 10695039-10695039 on Build GRCh38
rs763848894 Chr.6: 10694995-10694995 on Build GRCh38
rs780256684 Chr.6: 10695022-10695022 on Build GRCh38
rs758983576 Chr.6: 10694975-10694975 on Build GRCh38
rs879200696 Chr.6: 10694904-10694904 on Build GRCh38
rs112419636 Chr.6: 10694965-10694965 on Build GRCh38
rs765167483 Chr.6: 10695103-10695104 on Build GRCh38
rs565846191 Chr.6: 10695285-10695285 on Build GRCh38
rs112998972 Chr.6: 10695048-10695048 on Build GRCh38
rs531567144 Chr.6: 10695139-10695139 on Build GRCh38
rs746254482 Chr.6: 10695351-10695351 on Build GRCh38
rs368645069 Chr.6: 10694950-10694950 on Build GRCh38
rs762481701 Chr.6: 10695079-10695079 on Build GRCh38
rs201065929 Chr.6: 10695104-10695104 on Build GRCh38
rs1045584006 Chr.6: 10694906-10694906 on Build GRCh38
rs200871118 Chr.6: 10695001-10695001 on Build GRCh38
rs7742220 Chr.6: 10695258-10695258 on Build GRCh38

View Full Product Details