Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 14117717-14118210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD83
Entrez Gene ID
9308
Gene Name
CD83 molecule
Gene Aliases
BL11, HB15
UniGene
Hs.595133
Gene Chromosome Location
Chr.6: 14117256-14136918 on Build GRCh38
Transcripts
NM_001040280.1 NM_001251901.1 NM_004233.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6212479 c.153+4G>A p.? Chr.6: 14118069-14118069 on Build GRCh38
1196923 c.61C>G p.P21A Chr.6: 14117973-14117973 on Build GRCh38
450594 c.101C>T p.P34L Chr.6: 14118013-14118013 on Build GRCh38
6206749 c.153+8C>T p.? Chr.6: 14118073-14118073 on Build GRCh38
1161590 c.126G>A p.Q42Q Chr.6: 14118038-14118038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772197208 Chr.6: 14118019-14118019 on Build GRCh38
rs774672304 Chr.6: 14118001-14118001 on Build GRCh38
rs868306014 Chr.6: 14117824-14117824 on Build GRCh38
rs995123984 Chr.6: 14118016-14118016 on Build GRCh38
rs886476751 Chr.6: 14118037-14118037 on Build GRCh38
rs191995750 Chr.6: 14117951-14117951 on Build GRCh38
rs1028820386 Chr.6: 14117736-14117736 on Build GRCh38
rs201481435 Chr.6: 14117952-14117952 on Build GRCh38
rs776014831 Chr.6: 14118090-14118090 on Build GRCh38
rs771547901 Chr.6: 14117880-14117880 on Build GRCh38
rs748196931 Chr.6: 14118015-14118015 on Build GRCh38
rs775333580 Chr.6: 14117897-14117897 on Build GRCh38
rs557629646 Chr.6: 14117830-14117830 on Build GRCh38
rs769547541 Chr.6: 14117946-14117946 on Build GRCh38
rs551026480 Chr.6: 14118081-14118081 on Build GRCh38
rs764732620 Chr.6: 14118093-14118093 on Build GRCh38
rs754842313 Chr.6: 14118073-14118073 on Build GRCh38
rs543401049 Chr.6: 14117976-14117976 on Build GRCh38
rs749015030 Chr.6: 14118070-14118070 on Build GRCh38
rs756818537 Chr.6: 14118067-14118067 on Build GRCh38
rs765804909 Chr.6: 14118064-14118064 on Build GRCh38
rs762995685 Chr.6: 14117845-14117845 on Build GRCh38
rs377582590 Chr.6: 14117820-14117820 on Build GRCh38
rs955926639 Chr.6: 14117767-14117767 on Build GRCh38
rs571092171 Chr.6: 14118107-14118107 on Build GRCh38
rs770300340 Chr.6: 14117764-14117764 on Build GRCh38
rs757608513 Chr.6: 14117862-14117862 on Build GRCh38
rs377641266 Chr.6: 14117910-14117910 on Build GRCh38
rs745578592 Chr.6: 14118002-14118002 on Build GRCh38
rs780605284 Chr.6: 14117929-14117929 on Build GRCh38
rs772779206 Chr.6: 14117794-14117794 on Build GRCh38
rs573347554 Chr.6: 14117939-14117939 on Build GRCh38
rs199841901 Chr.6: 14118038-14118038 on Build GRCh38
rs372948996 Chr.6: 14117920-14117920 on Build GRCh38
rs995258989 Chr.6: 14117790-14117790 on Build GRCh38
rs978523250 Chr.6: 14117761-14117761 on Build GRCh38
rs924487316 Chr.6: 14118173-14118173 on Build GRCh38
rs752769812 Chr.6: 14117837-14117837 on Build GRCh38
rs761904490 Chr.6: 14117963-14117963 on Build GRCh38
rs773292889 Chr.6: 14118046-14118046 on Build GRCh38
rs370839800 Chr.6: 14118182-14118182 on Build GRCh38
rs760513345 Chr.6: 14117797-14117797 on Build GRCh38
rs756574697 Chr.6: 14117877-14117877 on Build GRCh38
rs547871356 Chr.6: 14118069-14118069 on Build GRCh38
rs779650210 Chr.6: 14118004-14118004 on Build GRCh38
rs757961559 Chr.6: 14118100-14118101 on Build GRCh38
rs749854064 Chr.6: 14118097-14118097 on Build GRCh38
rs899399503 Chr.6: 14117781-14117781 on Build GRCh38
rs1028325351 Chr.6: 14117914-14117914 on Build GRCh38
rs568423910 Chr.6: 14117769-14117769 on Build GRCh38
rs763491762 Chr.6: 14118092-14118092 on Build GRCh38
rs199812564 Chr.6: 14117816-14117816 on Build GRCh38
rs750737307 Chr.6: 14117926-14117926 on Build GRCh38
rs575580132 Chr.6: 14118142-14118142 on Build GRCh38
rs773489465 Chr.6: 14117967-14117967 on Build GRCh38
rs756519906 Chr.6: 14117928-14117928 on Build GRCh38
rs759423526 Chr.6: 14117888-14117888 on Build GRCh38
rs749141648 Chr.6: 14118008-14118008 on Build GRCh38
rs766653554 Chr.6: 14118056-14118056 on Build GRCh38
rs1048445037 Chr.6: 14118017-14118017 on Build GRCh38
rs369115814 Chr.6: 14117886-14117886 on Build GRCh38
rs780449249 Chr.6: 14117878-14117878 on Build GRCh38
rs781479969 Chr.6: 14117868-14117868 on Build GRCh38
rs373931910 Chr.6: 14117937-14117937 on Build GRCh38
rs780990603 Chr.6: 14117996-14117996 on Build GRCh38
rs940166585 Chr.6: 14118133-14118133 on Build GRCh38
rs555897798 Chr.6: 14118132-14118132 on Build GRCh38
rs778807997 Chr.6: 14118076-14118076 on Build GRCh38
rs751798123 Chr.6: 14117859-14117859 on Build GRCh38
rs568010456 Chr.6: 14118078-14118078 on Build GRCh38
rs771707547 Chr.6: 14117786-14117786 on Build GRCh38
rs759812378 Chr.6: 14118088-14118088 on Build GRCh38
rs968940738 Chr.6: 14118150-14118150 on Build GRCh38
rs745623724 Chr.6: 14117941-14117941 on Build GRCh38
rs111804875 Chr.6: 14117803-14117803 on Build GRCh38
rs747662904 Chr.6: 14117879-14117879 on Build GRCh38
rs773846710 Chr.6: 14117766-14117766 on Build GRCh38
rs138051430 Chr.6: 14118080-14118080 on Build GRCh38
rs889842486 Chr.6: 14117930-14117930 on Build GRCh38
rs112782255 Chr.6: 14117932-14117932 on Build GRCh38
rs547736250 Chr.6: 14117765-14117765 on Build GRCh38
rs975205432 Chr.6: 14117847-14117847 on Build GRCh38
rs776808603 Chr.6: 14118059-14118059 on Build GRCh38
rs200320680 Chr.6: 14117993-14117993 on Build GRCh38
rs769745422 Chr.6: 14117889-14117889 on Build GRCh38
rs1009279800 Chr.6: 14118051-14118051 on Build GRCh38
rs778609947 Chr.6: 14118115-14118115 on Build GRCh38
rs765918140 Chr.6: 14118106-14118106 on Build GRCh38
rs767080229 Chr.6: 14118068-14118068 on Build GRCh38
rs528134602 Chr.6: 14118065-14118065 on Build GRCh38
rs755396165 Chr.6: 14117987-14117987 on Build GRCh38
rs771034512 Chr.6: 14118084-14118084 on Build GRCh38
rs777078872 Chr.6: 14117935-14117935 on Build GRCh38
rs184937521 Chr.6: 14118123-14118123 on Build GRCh38
rs1035671854 Chr.6: 14118059-14118068 on Build GRCh38
rs774322730 Chr.6: 14118014-14118014 on Build GRCh38
rs544558198 Chr.6: 14118146-14118146 on Build GRCh38
rs559024173 Chr.6: 14118063-14118063 on Build GRCh38
rs979826291 Chr.6: 14117858-14117858 on Build GRCh38
rs955853795 Chr.6: 14117992-14117992 on Build GRCh38
rs577018663 Chr.6: 14118178-14118178 on Build GRCh38
rs370407778 Chr.6: 14118116-14118116 on Build GRCh38
rs545562141 Chr.6: 14118049-14118049 on Build GRCh38
rs756650749 Chr.6: 14117764-14117765 on Build GRCh38
rs761087447 Chr.6: 14117969-14117969 on Build GRCh38
rs535995254 Chr.6: 14118127-14118127 on Build GRCh38
rs376874262 Chr.6: 14117945-14117945 on Build GRCh38
rs556994325 Chr.6: 14118152-14118152 on Build GRCh38
rs767705052 Chr.6: 14117965-14117965 on Build GRCh38
rs368118535 Chr.6: 14118086-14118086 on Build GRCh38
rs764350777 Chr.6: 14117846-14117846 on Build GRCh38
rs776525262 Chr.6: 14117887-14117887 on Build GRCh38
rs756863673 Chr.6: 14117995-14117995 on Build GRCh38
rs201670981 Chr.6: 14118103-14118103 on Build GRCh38
rs774376193 Chr.6: 14117962-14117962 on Build GRCh38
rs762243383 Chr.6: 14118089-14118089 on Build GRCh38
rs553558881 Chr.6: 14118119-14118119 on Build GRCh38
rs766593353 Chr.6: 14117972-14117972 on Build GRCh38
rs780511827 Chr.6: 14117790-14117797 on Build GRCh38
rs370454590 Chr.6: 14118079-14118079 on Build GRCh38
rs754246369 Chr.6: 14117949-14117966 on Build GRCh38
rs2235368 Chr.6: 14117907-14117907 on Build GRCh38
rs369912153 Chr.6: 14117883-14117883 on Build GRCh38
rs761957759 Chr.6: 14117913-14117913 on Build GRCh38
rs187657576 Chr.6: 14117950-14117950 on Build GRCh38
rs563581725 Chr.6: 14117989-14117989 on Build GRCh38
rs575587222 Chr.6: 14117958-14117958 on Build GRCh38
rs978743215 Chr.6: 14118171-14118171 on Build GRCh38
rs534203052 Chr.6: 14117872-14117872 on Build GRCh38
rs539667595 Chr.6: 14118108-14118108 on Build GRCh38
rs559162537 Chr.6: 14118188-14118188 on Build GRCh38
rs1019032246 Chr.6: 14117819-14117819 on Build GRCh38
rs760282071 Chr.6: 14118104-14118104 on Build GRCh38
rs532220475 Chr.6: 14118009-14118009 on Build GRCh38
rs954161700 Chr.6: 14117751-14117751 on Build GRCh38

View Full Product Details