Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 16299033-16299543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN1
Entrez Gene ID
6310
Gene Name
ataxin 1
Gene Aliases
ATX1, D6S504E, SCA1
UniGene
Hs.434961
Gene Chromosome Location
Chr.6: 16299112-16761490 on Build GRCh38
Transcripts
NM_000332.3 NM_001128164.1

SNP Detail

SNP ID Chromosome Location
rs946612122 Chr.6: 16299505-16299505 on Build GRCh38
rs934719497 Chr.6: 16299162-16299162 on Build GRCh38
rs888100332 Chr.6: 16299357-16299357 on Build GRCh38
rs1044928142 Chr.6: 16299507-16299507 on Build GRCh38
rs1015384961 Chr.6: 16299084-16299084 on Build GRCh38
rs953917572 Chr.6: 16299096-16299096 on Build GRCh38
rs534321633 Chr.6: 16299214-16299215 on Build GRCh38
rs1044513316 Chr.6: 16299310-16299310 on Build GRCh38
rs779175890 Chr.6: 16299140-16299140 on Build GRCh38
rs775022351 Chr.6: 16299517-16299517 on Build GRCh38
rs1028149662 Chr.6: 16299492-16299492 on Build GRCh38
rs975638498 Chr.6: 16299354-16299354 on Build GRCh38
rs1003846359 Chr.6: 16299143-16299143 on Build GRCh38
rs750092828 Chr.6: 16299460-16299461 on Build GRCh38
rs146557770 Chr.6: 16299376-16299376 on Build GRCh38
rs934665763 Chr.6: 16299461-16299461 on Build GRCh38
rs377662674 Chr.6: 16299181-16299181 on Build GRCh38
rs548708943 Chr.6: 16299249-16299249 on Build GRCh38
rs915176616 Chr.6: 16299488-16299488 on Build GRCh38
rs953929846 Chr.6: 16299504-16299504 on Build GRCh38
rs764897144 Chr.6: 16299155-16299155 on Build GRCh38
rs778435564 Chr.6: 16299284-16299284 on Build GRCh38
rs1049358634 Chr.6: 16299339-16299339 on Build GRCh38
rs761347972 Chr.6: 16299062-16299063 on Build GRCh38
rs568962977 Chr.6: 16299338-16299338 on Build GRCh38
rs997950319 Chr.6: 16299469-16299469 on Build GRCh38
rs528834173 Chr.6: 16299234-16299234 on Build GRCh38
rs924664992 Chr.6: 16299446-16299446 on Build GRCh38
rs906069683 Chr.6: 16299327-16299327 on Build GRCh38
rs978733319 Chr.6: 16299114-16299114 on Build GRCh38
rs1028894064 Chr.6: 16299357-16299358 on Build GRCh38
rs878885817 Chr.6: 16299456-16299458 on Build GRCh38
rs976540310 Chr.6: 16299090-16299090 on Build GRCh38
rs772210812 Chr.6: 16299255-16299255 on Build GRCh38
rs1017198857 Chr.6: 16299180-16299183 on Build GRCh38
rs545792650 Chr.6: 16299181-16299184 on Build GRCh38
rs540555244 Chr.6: 16299086-16299086 on Build GRCh38
rs900897133 Chr.6: 16299463-16299463 on Build GRCh38
rs950141201 Chr.6: 16299309-16299309 on Build GRCh38
rs150423599 Chr.6: 16299161-16299161 on Build GRCh38
rs140524018 Chr.6: 16299074-16299074 on Build GRCh38
rs748199778 Chr.6: 16299200-16299200 on Build GRCh38
rs761138435 Chr.6: 16299479-16299479 on Build GRCh38
rs3834854 Chr.6: 16299220-16299221 on Build GRCh38
rs567194830 Chr.6: 16299150-16299151 on Build GRCh38
rs1034722220 Chr.6: 16299087-16299087 on Build GRCh38
rs955524429 Chr.6: 16299375-16299375 on Build GRCh38
rs1003846905 Chr.6: 16299373-16299373 on Build GRCh38
rs1053605895 Chr.6: 16299199-16299199 on Build GRCh38
rs921115071 Chr.6: 16299094-16299094 on Build GRCh38
rs963262591 Chr.6: 16299180-16299180 on Build GRCh38
rs531477936 Chr.6: 16299359-16299359 on Build GRCh38

View Full Product Details