Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 25800798-25801309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC17A1
Entrez Gene ID
6568
Gene Name
solute carrier family 17 member 1
Gene Aliases
NAPI-1, NPT-1, NPT1
UniGene
Hs.205816
Gene Chromosome Location
Chr.6: 25723744-25832108 on Build GRCh38
Transcripts
NM_005074.3 XM_011514818.2 XM_011514819.2 XM_011514820.2 XM_011514821.2 XM_017011199.1 XM_017011200.1 XM_017011201.1 XM_017011202.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3366408 c.1221G>T p.M407I Chr.6: 25800938-25800938 on Build GRCh38
1442544 c.1235T>C p.I412T Chr.6: 25800924-25800924 on Build GRCh38
3623363 c.1257G>A p.L419L Chr.6: 25800902-25800902 on Build GRCh38
1696980 c.1241C>T p.S414F Chr.6: 25800918-25800918 on Build GRCh38
5690806 c.1215T>A p.T405T Chr.6: 25800944-25800944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021492024 Chr.6: 25801185-25801185 on Build GRCh38
rs1004538487 Chr.6: 25801123-25801123 on Build GRCh38
rs748590582 Chr.6: 25800891-25800891 on Build GRCh38
rs564282932 Chr.6: 25801223-25801223 on Build GRCh38
rs775152924 Chr.6: 25800972-25800972 on Build GRCh38
rs761085335 Chr.6: 25800963-25800963 on Build GRCh38
rs1055435857 Chr.6: 25801154-25801154 on Build GRCh38
rs906762320 Chr.6: 25801137-25801137 on Build GRCh38
rs768797088 Chr.6: 25800898-25800898 on Build GRCh38
rs760083178 Chr.6: 25801228-25801228 on Build GRCh38
rs377729986 Chr.6: 25800854-25800854 on Build GRCh38
rs1034119053 Chr.6: 25800824-25800824 on Build GRCh38
rs186125927 Chr.6: 25800904-25800904 on Build GRCh38
rs763558837 Chr.6: 25801001-25801001 on Build GRCh38
rs370892770 Chr.6: 25800885-25800885 on Build GRCh38
rs762365919 Chr.6: 25800915-25800915 on Build GRCh38
rs780650351 Chr.6: 25800855-25800855 on Build GRCh38
rs915212572 Chr.6: 25800866-25800866 on Build GRCh38
rs750827664 Chr.6: 25800843-25800843 on Build GRCh38
rs757861616 Chr.6: 25800856-25800856 on Build GRCh38
rs1002553274 Chr.6: 25801142-25801142 on Build GRCh38
rs762619579 Chr.6: 25801000-25801000 on Build GRCh38
rs141225053 Chr.6: 25801201-25801201 on Build GRCh38
rs751453328 Chr.6: 25800869-25800869 on Build GRCh38
rs1048555955 Chr.6: 25801062-25801062 on Build GRCh38
rs772312084 Chr.6: 25800929-25800931 on Build GRCh38
rs767477277 Chr.6: 25801019-25801019 on Build GRCh38
rs375130789 Chr.6: 25800991-25800991 on Build GRCh38
rs1165209 Chr.6: 25801091-25801091 on Build GRCh38
rs1045444022 Chr.6: 25801184-25801184 on Build GRCh38
rs757218059 Chr.6: 25800872-25800872 on Build GRCh38
rs751038351 Chr.6: 25801013-25801013 on Build GRCh38
rs975389894 Chr.6: 25801281-25801281 on Build GRCh38
rs150311726 Chr.6: 25800830-25800830 on Build GRCh38
rs565643872 Chr.6: 25801176-25801176 on Build GRCh38
rs370346678 Chr.6: 25800853-25800853 on Build GRCh38
rs541124639 Chr.6: 25801200-25801200 on Build GRCh38
rs944849731 Chr.6: 25800899-25800899 on Build GRCh38
rs908697867 Chr.6: 25801066-25801066 on Build GRCh38
rs769536619 Chr.6: 25800891-25800891 on Build GRCh38
rs1011461667 Chr.6: 25801274-25801274 on Build GRCh38
rs1047069211 Chr.6: 25801097-25801097 on Build GRCh38
rs766659170 Chr.6: 25800966-25800966 on Build GRCh38
rs750396381 Chr.6: 25801027-25801027 on Build GRCh38
rs143142456 Chr.6: 25800923-25800923 on Build GRCh38
rs781149209 Chr.6: 25800873-25800873 on Build GRCh38
rs773637782 Chr.6: 25800925-25800925 on Build GRCh38
rs975360187 Chr.6: 25800938-25800938 on Build GRCh38
rs921360008 Chr.6: 25800942-25800942 on Build GRCh38
rs957441590 Chr.6: 25801240-25801240 on Build GRCh38
rs774863938 Chr.6: 25801183-25801183 on Build GRCh38
rs545607257 Chr.6: 25801018-25801018 on Build GRCh38
rs773394681 Chr.6: 25800893-25800893 on Build GRCh38
rs749475831 Chr.6: 25800895-25800895 on Build GRCh38
rs1165210 Chr.6: 25800829-25800829 on Build GRCh38
rs555716758 Chr.6: 25801174-25801174 on Build GRCh38
rs965237024 Chr.6: 25801263-25801263 on Build GRCh38
rs773568937 Chr.6: 25800989-25800990 on Build GRCh38
rs906974917 Chr.6: 25801198-25801198 on Build GRCh38
rs868164347 Chr.6: 25800906-25800906 on Build GRCh38
rs967602302 Chr.6: 25801220-25801220 on Build GRCh38
rs572136518 Chr.6: 25801175-25801175 on Build GRCh38
rs777282526 Chr.6: 25800868-25800868 on Build GRCh38
rs199907051 Chr.6: 25800926-25800926 on Build GRCh38

View Full Product Details