Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 25862096-25862641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC17A3
Entrez Gene ID
10786
Gene Name
solute carrier family 17 member 3
Gene Aliases
GOUT4, NPT4, UAQTL4
UniGene
Hs.327179
Gene Chromosome Location
Chr.6: 25845100-25874243 on Build GRCh38
Transcripts
NM_001098486.1 NM_006632.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3761733 c.298G>A p.A100T Chr.6: 25862238-25862238 on Build GRCh38
6173479 c.264T>A p.F88L Chr.6: 25862272-25862272 on Build GRCh38
3623390 c.221C>T p.S74F Chr.6: 25862315-25862315 on Build GRCh38
3761732 c.298G>A p.A100T Chr.6: 25862238-25862238 on Build GRCh38
6034931 c.295C>T p.P99S Chr.6: 25862241-25862241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866508354 Chr.6: 25862442-25862442 on Build GRCh38
rs889595703 Chr.6: 25862570-25862570 on Build GRCh38
rs775274504 Chr.6: 25862562-25862562 on Build GRCh38
rs776504543 Chr.6: 25862205-25862205 on Build GRCh38
rs993617566 Chr.6: 25862496-25862496 on Build GRCh38
rs147686677 Chr.6: 25862355-25862355 on Build GRCh38
rs777359199 Chr.6: 25862208-25862208 on Build GRCh38
rs151168556 Chr.6: 25862539-25862539 on Build GRCh38
rs770455457 Chr.6: 25862443-25862443 on Build GRCh38
rs1025748444 Chr.6: 25862618-25862618 on Build GRCh38
rs1052902922 Chr.6: 25862530-25862530 on Build GRCh38
rs762844729 Chr.6: 25862493-25862493 on Build GRCh38
rs779208858 Chr.6: 25862460-25862460 on Build GRCh38
rs1016956075 Chr.6: 25862317-25862317 on Build GRCh38
rs934419238 Chr.6: 25862593-25862593 on Build GRCh38
rs766365403 Chr.6: 25862233-25862233 on Build GRCh38
rs79826932 Chr.6: 25862611-25862611 on Build GRCh38
rs139869471 Chr.6: 25862423-25862423 on Build GRCh38
rs181724461 Chr.6: 25862606-25862606 on Build GRCh38
rs916877785 Chr.6: 25862445-25862445 on Build GRCh38
rs758625183 Chr.6: 25862361-25862361 on Build GRCh38
rs545706751 Chr.6: 25862304-25862304 on Build GRCh38
rs78287505 Chr.6: 25862607-25862607 on Build GRCh38
rs757424881 Chr.6: 25862379-25862379 on Build GRCh38
rs963475923 Chr.6: 25862471-25862471 on Build GRCh38
rs372711885 Chr.6: 25862327-25862327 on Build GRCh38
rs387907257 Chr.6: 25862334-25862334 on Build GRCh38
rs556407051 Chr.6: 25862321-25862321 on Build GRCh38
rs777970636 Chr.6: 25862263-25862263 on Build GRCh38
rs146348149 Chr.6: 25862410-25862410 on Build GRCh38
rs1165165 Chr.6: 25862238-25862238 on Build GRCh38
rs780441613 Chr.6: 25862412-25862412 on Build GRCh38
rs752475287 Chr.6: 25862354-25862354 on Build GRCh38
rs1026484200 Chr.6: 25862507-25862507 on Build GRCh38
rs370851319 Chr.6: 25862280-25862280 on Build GRCh38
rs576305771 Chr.6: 25862378-25862378 on Build GRCh38
rs761175840 Chr.6: 25862182-25862182 on Build GRCh38
rs756988365 Chr.6: 25862381-25862381 on Build GRCh38
rs904217075 Chr.6: 25862486-25862486 on Build GRCh38
rs548410321 Chr.6: 25862346-25862346 on Build GRCh38
rs866082995 Chr.6: 25862553-25862553 on Build GRCh38
rs370136832 Chr.6: 25862216-25862216 on Build GRCh38
rs142438811 Chr.6: 25862406-25862406 on Build GRCh38
rs770215693 Chr.6: 25862295-25862295 on Build GRCh38
rs765112090 Chr.6: 25862336-25862336 on Build GRCh38
rs752861068 Chr.6: 25862472-25862473 on Build GRCh38
rs377331591 Chr.6: 25862262-25862262 on Build GRCh38
rs115225744 Chr.6: 25862424-25862424 on Build GRCh38
rs916137774 Chr.6: 25862537-25862537 on Build GRCh38
rs765322317 Chr.6: 25862248-25862248 on Build GRCh38
rs904313884 Chr.6: 25862132-25862132 on Build GRCh38
rs771757955 Chr.6: 25862314-25862314 on Build GRCh38
rs775343623 Chr.6: 25862485-25862485 on Build GRCh38
rs747135425 Chr.6: 25862276-25862276 on Build GRCh38
rs998070204 Chr.6: 25862542-25862542 on Build GRCh38
rs925703501 Chr.6: 25862316-25862316 on Build GRCh38
rs759579287 Chr.6: 25862246-25862247 on Build GRCh38
rs199802947 Chr.6: 25862217-25862217 on Build GRCh38
rs375972210 Chr.6: 25862329-25862329 on Build GRCh38
rs1044037520 Chr.6: 25862127-25862127 on Build GRCh38
rs776392198 Chr.6: 25862456-25862456 on Build GRCh38
rs752986681 Chr.6: 25862254-25862254 on Build GRCh38
rs781745859 Chr.6: 25862380-25862380 on Build GRCh38
rs566130121 Chr.6: 25862126-25862126 on Build GRCh38
rs747801489 Chr.6: 25862313-25862313 on Build GRCh38
rs746160411 Chr.6: 25862290-25862290 on Build GRCh38
rs535131207 Chr.6: 25862183-25862183 on Build GRCh38
rs754811305 Chr.6: 25862239-25862239 on Build GRCh38
rs370057568 Chr.6: 25862330-25862330 on Build GRCh38
rs1043921454 Chr.6: 25862468-25862468 on Build GRCh38
rs564056592 Chr.6: 25862595-25862595 on Build GRCh38
rs17851584 Chr.6: 25862275-25862275 on Build GRCh38
rs970734306 Chr.6: 25862540-25862540 on Build GRCh38
rs757833943 Chr.6: 25862277-25862277 on Build GRCh38
rs765241486 Chr.6: 25862407-25862410 on Build GRCh38
rs960272969 Chr.6: 25862155-25862155 on Build GRCh38
rs764470766 Chr.6: 25862372-25862372 on Build GRCh38
rs985494834 Chr.6: 25862286-25862286 on Build GRCh38
rs1052869894 Chr.6: 25862335-25862335 on Build GRCh38
rs1006816870 Chr.6: 25862575-25862575 on Build GRCh38
rs891521518 Chr.6: 25862532-25862532 on Build GRCh38
rs1016695070 Chr.6: 25862429-25862429 on Build GRCh38
rs765138165 Chr.6: 25862463-25862463 on Build GRCh38
rs34508088 Chr.6: 25862190-25862190 on Build GRCh38
rs527353361 Chr.6: 25862439-25862439 on Build GRCh38
rs935641254 Chr.6: 25862505-25862505 on Build GRCh38

View Full Product Details