Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26390674-26391200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN2A2
Entrez Gene ID
10385
Gene Name
butyrophilin subfamily 2 member A2
Gene Aliases
BT2.2, BTF2, BTN2.2
UniGene
Hs.373938
Gene Chromosome Location
Chr.6: 26382891-26394874 on Build GRCh38
Transcripts
NM_001197237.1 NM_001197238.1 NM_001197239.1 NM_001197240.1 NM_006995.4 NM_181531.2 XM_005248797.3 XM_005248798.4 XM_006714953.3 XM_006714955.3 XM_011514228.2 XM_011514229.2 XM_011514231.2 XM_017010166.1 XM_017010167.1 XM_017010168.1 XM_017010169.1 XM_011514268.2

Gene Symbol
BTN3A2
Entrez Gene ID
11118
Gene Name
butyrophilin subfamily 3 member A2
Gene Aliases
BT3.2, BTF4, BTN3.2, CD277
UniGene
Hs.376046
Gene Chromosome Location
Chr.6: 26365159-26451697 on Build GRCh38
Transcripts
NM_001197237.1 NM_001197238.1 NM_001197239.1 NM_001197240.1 NM_006995.4 NM_181531.2 XM_005248797.3 XM_005248798.4 XM_006714953.3 XM_006714955.3 XM_011514228.2 XM_011514229.2 XM_011514231.2 XM_017010166.1 XM_017010167.1 XM_017010168.1 XM_017010169.1 XM_011514268.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5975921 c.952+1G>T p.? Chr.6: 26390708-26390708 on Build GRCh38
4666392 c.671-1G>T p.? Chr.6: 26390802-26390802 on Build GRCh38
4666391 c.953-1G>T p.? Chr.6: 26390802-26390802 on Build GRCh38
5975922 c.952+1G>T p.? Chr.6: 26390708-26390708 on Build GRCh38
6613537 c.953-1G>T p.? Chr.6: 26390802-26390802 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373288008 Chr.6: 26390718-26390718 on Build GRCh38
rs761743719 Chr.6: 26390705-26390705 on Build GRCh38
rs891389083 Chr.6: 26391055-26391055 on Build GRCh38
rs770796723 Chr.6: 26390827-26390827 on Build GRCh38
rs928351935 Chr.6: 26391137-26391137 on Build GRCh38
rs758886447 Chr.6: 26390753-26390753 on Build GRCh38
rs759563161 Chr.6: 26390708-26390708 on Build GRCh38
rs200502017 Chr.6: 26390797-26390797 on Build GRCh38
rs569106839 Chr.6: 26391139-26391139 on Build GRCh38
rs757588311 Chr.6: 26390773-26390773 on Build GRCh38
rs538053564 Chr.6: 26391149-26391149 on Build GRCh38
rs775703148 Chr.6: 26390740-26390740 on Build GRCh38
rs764703476 Chr.6: 26390866-26390866 on Build GRCh38
rs192311792 Chr.6: 26391000-26391000 on Build GRCh38
rs759770098 Chr.6: 26390836-26390836 on Build GRCh38
rs756212182 Chr.6: 26390927-26390927 on Build GRCh38
rs201272391 Chr.6: 26390872-26390872 on Build GRCh38
rs971820674 Chr.6: 26391078-26391078 on Build GRCh38
rs754717188 Chr.6: 26390886-26390886 on Build GRCh38
rs778132036 Chr.6: 26390765-26390765 on Build GRCh38
rs386698235 Chr.6: 26391166-26391167 on Build GRCh38
rs71544681 Chr.6: 26391163-26391164 on Build GRCh38
rs925085705 Chr.6: 26390946-26390946 on Build GRCh38
rs562989262 Chr.6: 26390792-26390792 on Build GRCh38
rs200463511 Chr.6: 26390708-26390708 on Build GRCh38
rs752040796 Chr.6: 26390869-26390869 on Build GRCh38
rs11756444 Chr.6: 26391163-26391163 on Build GRCh38
rs765068708 Chr.6: 26390708-26390709 on Build GRCh38
rs997274512 Chr.6: 26390944-26390944 on Build GRCh38
rs984386673 Chr.6: 26390731-26390731 on Build GRCh38
rs531770086 Chr.6: 26390824-26390824 on Build GRCh38
rs17537465 Chr.6: 26390777-26390777 on Build GRCh38
rs34582313 Chr.6: 26391066-26391067 on Build GRCh38
rs540093983 Chr.6: 26390971-26390971 on Build GRCh38
rs550225952 Chr.6: 26391106-26391106 on Build GRCh38
rs757641440 Chr.6: 26390871-26390871 on Build GRCh38
rs371166406 Chr.6: 26390772-26390772 on Build GRCh38
rs767598841 Chr.6: 26390791-26390791 on Build GRCh38
rs1028791185 Chr.6: 26390958-26390958 on Build GRCh38
rs765248834 Chr.6: 26390846-26390846 on Build GRCh38
rs9467748 Chr.6: 26390925-26390925 on Build GRCh38
rs779732237 Chr.6: 26390775-26390775 on Build GRCh38
rs576875639 Chr.6: 26390736-26390736 on Build GRCh38
rs199636222 Chr.6: 26390832-26390832 on Build GRCh38
rs754067995 Chr.6: 26390997-26390997 on Build GRCh38
rs759714221 Chr.6: 26390728-26390728 on Build GRCh38
rs962042003 Chr.6: 26391012-26391012 on Build GRCh38
rs374432830 Chr.6: 26390936-26390936 on Build GRCh38
rs370266526 Chr.6: 26390715-26390715 on Build GRCh38
rs12205476 Chr.6: 26391157-26391157 on Build GRCh38
rs778424261 Chr.6: 26390887-26390887 on Build GRCh38
rs2237234 Chr.6: 26391166-26391166 on Build GRCh38
rs894702818 Chr.6: 26390825-26390825 on Build GRCh38
rs760544320 Chr.6: 26390711-26390711 on Build GRCh38
rs183275760 Chr.6: 26391004-26391004 on Build GRCh38
rs747732083 Chr.6: 26390889-26390889 on Build GRCh38
rs2237235 Chr.6: 26391167-26391167 on Build GRCh38
rs2072804 Chr.6: 26390962-26390962 on Build GRCh38
rs753502566 Chr.6: 26390880-26390880 on Build GRCh38
rs41266835 Chr.6: 26390735-26390735 on Build GRCh38
rs775700940 Chr.6: 26390849-26390849 on Build GRCh38
rs929626467 Chr.6: 26390829-26390829 on Build GRCh38
rs771778516 Chr.6: 26390707-26390707 on Build GRCh38
rs762939985 Chr.6: 26390850-26390850 on Build GRCh38
rs768159236 Chr.6: 26390783-26390783 on Build GRCh38
rs371030792 Chr.6: 26390987-26390987 on Build GRCh38
rs891821303 Chr.6: 26391003-26391003 on Build GRCh38

View Full Product Details