Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 39190411-39190912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK5
Entrez Gene ID
8645
Gene Name
potassium two pore domain channel subfamily K member 5
Gene Aliases
K2p5.1, KCNK5b, TASK-2, TASK2
UniGene
Hs.444448
Gene Chromosome Location
Chr.6: 39188971-39229475 on Build GRCh38
Transcripts
NM_003740.3 XM_005249456.1 XM_006715235.2

SNP Detail

SNP ID Chromosome Location
rs959479790 Chr.6: 39190484-39190484 on Build GRCh38
rs79407244 Chr.6: 39190655-39190655 on Build GRCh38
rs1044837169 Chr.6: 39190439-39190439 on Build GRCh38
rs998684389 Chr.6: 39190830-39190830 on Build GRCh38
rs574893131 Chr.6: 39190499-39190499 on Build GRCh38
rs1021903819 Chr.6: 39190785-39190785 on Build GRCh38
rs973438090 Chr.6: 39190750-39190750 on Build GRCh38
rs141588229 Chr.6: 39190708-39190708 on Build GRCh38
rs1053556700 Chr.6: 39190820-39190820 on Build GRCh38
rs1025570617 Chr.6: 39190835-39190835 on Build GRCh38
rs4714222 Chr.6: 39190563-39190563 on Build GRCh38
rs1031120957 Chr.6: 39190485-39190485 on Build GRCh38
rs994954830 Chr.6: 39190881-39190881 on Build GRCh38
rs776155806 Chr.6: 39190624-39190624 on Build GRCh38
rs1052063051 Chr.6: 39190726-39190726 on Build GRCh38
rs1053462808 Chr.6: 39190672-39190672 on Build GRCh38
rs762012701 Chr.6: 39190846-39190846 on Build GRCh38
rs939139362 Chr.6: 39190721-39190721 on Build GRCh38
rs1042071581 Chr.6: 39190844-39190845 on Build GRCh38
rs754994351 Chr.6: 39190684-39190684 on Build GRCh38
rs947776264 Chr.6: 39190436-39190436 on Build GRCh38
rs906779590 Chr.6: 39190845-39190845 on Build GRCh38
rs892051725 Chr.6: 39190487-39190487 on Build GRCh38
rs554626998 Chr.6: 39190483-39190483 on Build GRCh38
rs970995107 Chr.6: 39190696-39190696 on Build GRCh38
rs569424725 Chr.6: 39190678-39190678 on Build GRCh38
rs1021245565 Chr.6: 39190691-39190691 on Build GRCh38
rs148788474 Chr.6: 39190596-39190596 on Build GRCh38
rs965880381 Chr.6: 39190555-39190555 on Build GRCh38
rs570127869 Chr.6: 39190857-39190857 on Build GRCh38
rs985926029 Chr.6: 39190539-39190539 on Build GRCh38
rs746011992 Chr.6: 39190540-39190540 on Build GRCh38
rs543910179 Chr.6: 39190505-39190505 on Build GRCh38
rs765987521 Chr.6: 39190827-39190827 on Build GRCh38
rs1034274605 Chr.6: 39190860-39190860 on Build GRCh38
rs577056853 Chr.6: 39190813-39190813 on Build GRCh38
rs920663841 Chr.6: 39190752-39190752 on Build GRCh38
rs780821412 Chr.6: 39190720-39190720 on Build GRCh38
rs899584152 Chr.6: 39190617-39190617 on Build GRCh38
rs947892405 Chr.6: 39190706-39190706 on Build GRCh38
rs552306774 Chr.6: 39190814-39190814 on Build GRCh38
rs767790920 Chr.6: 39190879-39190879 on Build GRCh38
rs750184368 Chr.6: 39190880-39190880 on Build GRCh38
rs918589185 Chr.6: 39190585-39190585 on Build GRCh38
rs549499682 Chr.6: 39190773-39190773 on Build GRCh38
rs908495487 Chr.6: 39190786-39190786 on Build GRCh38
rs184396902 Chr.6: 39190766-39190766 on Build GRCh38
rs965451550 Chr.6: 39190828-39190828 on Build GRCh38
rs929860966 Chr.6: 39190626-39190626 on Build GRCh38
rs566238772 Chr.6: 39190443-39190443 on Build GRCh38
rs932411505 Chr.6: 39190816-39190816 on Build GRCh38
rs920558835 Chr.6: 39190500-39190500 on Build GRCh38
rs527550171 Chr.6: 39190689-39190689 on Build GRCh38
rs374965520 Chr.6: 39190799-39190799 on Build GRCh38
rs3734615 Chr.6: 39190525-39190525 on Build GRCh38
rs943931192 Chr.6: 39190798-39190798 on Build GRCh38
rs535022407 Chr.6: 39190444-39190444 on Build GRCh38
rs984015107 Chr.6: 39190629-39190629 on Build GRCh38
rs3734616 Chr.6: 39190738-39190738 on Build GRCh38

View Full Product Details