Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152085515-152086013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCHHL1
Entrez Gene ID
126637
Gene Name
trichohyalin like 1
Gene Aliases
S100A17, THHL1
UniGene
Hs.546477
Gene Chromosome Location
Chr.1: 152084144-152089064 on Build GRCh38
Transcripts
NM_001008536.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5589131 c.2107G>A p.G703S Chr.1: 152085575-152085575 on Build GRCh38
1491726 c.2115T>A p.S705R Chr.1: 152085567-152085567 on Build GRCh38
3473779 c.2132G>A p.G711E Chr.1: 152085550-152085550 on Build GRCh38
528895 c.1775C>A p.P592Q Chr.1: 152085907-152085907 on Build GRCh38
4600146 c.1894G>C p.E632Q Chr.1: 152085788-152085788 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759436797 Chr.1: 152085921-152085921 on Build GRCh38
rs748919984 Chr.1: 152085751-152085751 on Build GRCh38
rs150014958 Chr.1: 152085716-152085716 on Build GRCh38
rs770050163 Chr.1: 152085938-152085938 on Build GRCh38
rs754027296 Chr.1: 152085720-152085720 on Build GRCh38
rs967297441 Chr.1: 152085947-152085947 on Build GRCh38
rs752517226 Chr.1: 152085968-152085968 on Build GRCh38
rs372763917 Chr.1: 152085564-152085564 on Build GRCh38
rs143917611 Chr.1: 152085963-152085963 on Build GRCh38
rs772697835 Chr.1: 152085771-152085771 on Build GRCh38
rs867167639 Chr.1: 152085806-152085806 on Build GRCh38
rs780640667 Chr.1: 152085636-152085636 on Build GRCh38
rs996143524 Chr.1: 152085974-152085974 on Build GRCh38
rs777276330 Chr.1: 152085750-152085750 on Build GRCh38
rs764213395 Chr.1: 152085785-152085785 on Build GRCh38
rs768500320 Chr.1: 152085631-152085631 on Build GRCh38
rs563845269 Chr.1: 152085865-152085865 on Build GRCh38
rs577347116 Chr.1: 152085717-152085717 on Build GRCh38
rs746271861 Chr.1: 152085762-152085762 on Build GRCh38
rs752034291 Chr.1: 152085549-152085549 on Build GRCh38
rs566602576 Chr.1: 152085950-152085950 on Build GRCh38
rs967458296 Chr.1: 152085602-152085602 on Build GRCh38
rs41266086 Chr.1: 152085737-152085737 on Build GRCh38
rs779820997 Chr.1: 152085769-152085769 on Build GRCh38
rs1016149764 Chr.1: 152085658-152085658 on Build GRCh38
rs991651066 Chr.1: 152085694-152085694 on Build GRCh38
rs886312669 Chr.1: 152085606-152085606 on Build GRCh38
rs779719742 Chr.1: 152085617-152085617 on Build GRCh38
rs189336449 Chr.1: 152085809-152085809 on Build GRCh38
rs375684500 Chr.1: 152085734-152085734 on Build GRCh38
rs756377255 Chr.1: 152085578-152085578 on Build GRCh38
rs1035354884 Chr.1: 152085965-152085965 on Build GRCh38
rs377058550 Chr.1: 152085699-152085699 on Build GRCh38
rs1047476705 Chr.1: 152085835-152085835 on Build GRCh38
rs761856474 Chr.1: 152085932-152085932 on Build GRCh38
rs1055769468 Chr.1: 152085777-152085777 on Build GRCh38
rs748896386 Chr.1: 152085666-152085666 on Build GRCh38
rs199816622 Chr.1: 152085748-152085748 on Build GRCh38
rs554946548 Chr.1: 152085568-152085568 on Build GRCh38
rs768213971 Chr.1: 152085839-152085840 on Build GRCh38
rs763984666 Chr.1: 152085986-152085986 on Build GRCh38
rs908762414 Chr.1: 152085727-152085727 on Build GRCh38
rs996934668 Chr.1: 152085765-152085765 on Build GRCh38
rs748784319 Chr.1: 152085885-152085885 on Build GRCh38
rs756908094 Chr.1: 152085901-152085901 on Build GRCh38
rs761272378 Chr.1: 152085783-152085783 on Build GRCh38
rs548538132 Chr.1: 152085983-152085983 on Build GRCh38
rs746722954 Chr.1: 152085835-152085835 on Build GRCh38
rs41266084 Chr.1: 152085668-152085668 on Build GRCh38
rs767302326 Chr.1: 152085853-152085853 on Build GRCh38
rs770517414 Chr.1: 152085752-152085752 on Build GRCh38
rs201733774 Chr.1: 152085760-152085760 on Build GRCh38
rs776742015 Chr.1: 152085923-152085923 on Build GRCh38
rs554977997 Chr.1: 152085585-152085585 on Build GRCh38
rs149143221 Chr.1: 152085664-152085664 on Build GRCh38
rs776207691 Chr.1: 152085781-152085781 on Build GRCh38
rs760045070 Chr.1: 152085681-152085681 on Build GRCh38
rs181883123 Chr.1: 152085948-152085948 on Build GRCh38
rs868196788 Chr.1: 152085628-152085628 on Build GRCh38
rs754834315 Chr.1: 152085851-152085851 on Build GRCh38
rs143095130 Chr.1: 152085909-152085909 on Build GRCh38
rs747705881 Chr.1: 152085646-152085646 on Build GRCh38
rs149878459 Chr.1: 152085922-152085922 on Build GRCh38
rs772501534 Chr.1: 152085918-152085918 on Build GRCh38
rs1021565520 Chr.1: 152085935-152085935 on Build GRCh38
rs546414179 Chr.1: 152085721-152085721 on Build GRCh38
rs762056212 Chr.1: 152085796-152085796 on Build GRCh38
rs888955730 Chr.1: 152085828-152085828 on Build GRCh38
rs777294470 Chr.1: 152085884-152085884 on Build GRCh38
rs752572108 Chr.1: 152085869-152085869 on Build GRCh38
rs191248783 Chr.1: 152085706-152085706 on Build GRCh38
rs542201601 Chr.1: 152085788-152085788 on Build GRCh38
rs1021514165 Chr.1: 152085595-152085595 on Build GRCh38
rs375554341 Chr.1: 152085847-152085847 on Build GRCh38
rs200319700 Chr.1: 152085733-152085733 on Build GRCh38
rs750233685 Chr.1: 152085600-152085600 on Build GRCh38
rs766149904 Chr.1: 152085728-152085728 on Build GRCh38
rs765419076 Chr.1: 152085836-152085836 on Build GRCh38
rs756065725 Chr.1: 152085878-152085878 on Build GRCh38
rs916076191 Chr.1: 152085696-152085696 on Build GRCh38
rs773238747 Chr.1: 152085942-152085942 on Build GRCh38
rs758282713 Chr.1: 152085604-152085604 on Build GRCh38
rs778491647 Chr.1: 152085903-152085903 on Build GRCh38
rs780711414 Chr.1: 152085967-152085967 on Build GRCh38
rs1005970153 Chr.1: 152085639-152085639 on Build GRCh38
rs746908725 Chr.1: 152085620-152085620 on Build GRCh38
rs532974821 Chr.1: 152085937-152085937 on Build GRCh38
rs769422808 Chr.1: 152085650-152085650 on Build GRCh38
rs183880729 Chr.1: 152085793-152085793 on Build GRCh38
rs774597573 Chr.1: 152085677-152085677 on Build GRCh38
rs138170974 Chr.1: 152085566-152085566 on Build GRCh38

View Full Product Details