Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153001654-153002196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRR3
Entrez Gene ID
6707
Gene Name
small proline rich protein 3
Gene Aliases
-
UniGene
Hs.139322
Gene Chromosome Location
Chr.1: 153001747-153003856 on Build GRCh38
Transcripts
NM_001097589.1 NM_005416.2

SNP Detail

SNP ID Chromosome Location
rs889407359 Chr.1: 153001854-153001854 on Build GRCh38
rs149789358 Chr.1: 153002073-153002073 on Build GRCh38
rs1012910278 Chr.1: 153001961-153001962 on Build GRCh38
rs909696690 Chr.1: 153002084-153002084 on Build GRCh38
rs4845329 Chr.1: 153001978-153001978 on Build GRCh38
rs1024137044 Chr.1: 153002060-153002060 on Build GRCh38
rs777599406 Chr.1: 153001729-153001729 on Build GRCh38
rs1040519541 Chr.1: 153002124-153002124 on Build GRCh38
rs758260784 Chr.1: 153001720-153001720 on Build GRCh38
rs560721241 Chr.1: 153001980-153001980 on Build GRCh38
rs6684188 Chr.1: 153001809-153001809 on Build GRCh38
rs1039231238 Chr.1: 153002004-153002004 on Build GRCh38
rs969830373 Chr.1: 153001898-153001898 on Build GRCh38
rs1013883623 Chr.1: 153002053-153002053 on Build GRCh38
rs562877179 Chr.1: 153001810-153001810 on Build GRCh38
rs919261149 Chr.1: 153001961-153001961 on Build GRCh38
rs903359137 Chr.1: 153002129-153002129 on Build GRCh38
rs1054467189 Chr.1: 153002164-153002164 on Build GRCh38
rs538354321 Chr.1: 153001692-153001692 on Build GRCh38
rs930840670 Chr.1: 153001799-153001799 on Build GRCh38
rs991374691 Chr.1: 153001737-153001737 on Build GRCh38
rs967898371 Chr.1: 153001699-153001699 on Build GRCh38
rs777377971 Chr.1: 153001951-153001951 on Build GRCh38
rs1040967624 Chr.1: 153001868-153001868 on Build GRCh38
rs1030627341 Chr.1: 153001726-153001726 on Build GRCh38
rs978803399 Chr.1: 153001803-153001803 on Build GRCh38
rs926677810 Chr.1: 153001906-153001906 on Build GRCh38
rs929325345 Chr.1: 153001977-153001977 on Build GRCh38
rs566358522 Chr.1: 153001756-153001756 on Build GRCh38
rs189037234 Chr.1: 153001757-153001757 on Build GRCh38
rs992704068 Chr.1: 153001840-153001840 on Build GRCh38
rs193126653 Chr.1: 153001887-153001887 on Build GRCh38
rs999932117 Chr.1: 153002021-153002021 on Build GRCh38
rs769631523 Chr.1: 153001939-153001939 on Build GRCh38
rs969967631 Chr.1: 153002062-153002062 on Build GRCh38
rs955726873 Chr.1: 153002040-153002040 on Build GRCh38
rs957370497 Chr.1: 153001943-153001943 on Build GRCh38
rs879055352 Chr.1: 153001707-153001707 on Build GRCh38
rs1009056649 Chr.1: 153001863-153001863 on Build GRCh38
rs28924721 Chr.1: 153001778-153001778 on Build GRCh38
rs184504624 Chr.1: 153001686-153001686 on Build GRCh38
rs955923746 Chr.1: 153001821-153001821 on Build GRCh38
rs556339256 Chr.1: 153001827-153001827 on Build GRCh38
rs903245526 Chr.1: 153001899-153001899 on Build GRCh38
rs754798002 Chr.1: 153001834-153001834 on Build GRCh38
rs865904369 Chr.1: 153001775-153001775 on Build GRCh38
rs549568006 Chr.1: 153002009-153002009 on Build GRCh38
rs971311515 Chr.1: 153001970-153001970 on Build GRCh38
rs6671524 Chr.1: 153001771-153001771 on Build GRCh38
rs139800473 Chr.1: 153001904-153001904 on Build GRCh38
rs573597674 Chr.1: 153001804-153001804 on Build GRCh38
rs957257233 Chr.1: 153001721-153001721 on Build GRCh38
rs17629427 Chr.1: 153001780-153001780 on Build GRCh38
rs981710756 Chr.1: 153001763-153001763 on Build GRCh38
rs1031880291 Chr.1: 153002024-153002024 on Build GRCh38
rs919598443 Chr.1: 153001787-153001787 on Build GRCh38
rs527324317 Chr.1: 153001955-153001955 on Build GRCh38
rs557852660 Chr.1: 153001715-153001715 on Build GRCh38
rs576040835 Chr.1: 153001865-153001865 on Build GRCh38
rs34942291 Chr.1: 153001753-153001754 on Build GRCh38
rs1037726487 Chr.1: 153001690-153001690 on Build GRCh38
rs529639504 Chr.1: 153002001-153002001 on Build GRCh38
rs145727242 Chr.1: 153002146-153002146 on Build GRCh38
rs148923996 Chr.1: 153002159-153002159 on Build GRCh38

View Full Product Details