Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153375461-153375967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
S100A12
Entrez Gene ID
6283
Gene Name
S100 calcium binding protein A12
Gene Aliases
CAAF1, CAGC, CGRP, ENRAGE, MRP-6, MRP6, p6
UniGene
Hs.19413
Gene Chromosome Location
Chr.1: 153373708-153375599 on Build GRCh38
Transcripts
NM_005621.1

SNP Detail

SNP ID Chromosome Location
rs753716947 Chr.1: 153375774-153375774 on Build GRCh38
rs1043292816 Chr.1: 153375593-153375593 on Build GRCh38
rs981473934 Chr.1: 153375537-153375537 on Build GRCh38
rs552934106 Chr.1: 153375850-153375850 on Build GRCh38
rs891972330 Chr.1: 153375570-153375570 on Build GRCh38
rs1004757084 Chr.1: 153375769-153375769 on Build GRCh38
rs187616709 Chr.1: 153375532-153375532 on Build GRCh38
rs569206892 Chr.1: 153375834-153375834 on Build GRCh38
rs566910357 Chr.1: 153375763-153375763 on Build GRCh38
rs922000819 Chr.1: 153375504-153375504 on Build GRCh38
rs904824899 Chr.1: 153375652-153375652 on Build GRCh38
rs1033091938 Chr.1: 153375933-153375933 on Build GRCh38
rs974725436 Chr.1: 153375830-153375830 on Build GRCh38
rs944980826 Chr.1: 153375758-153375758 on Build GRCh38
rs1016180109 Chr.1: 153375789-153375789 on Build GRCh38
rs529327442 Chr.1: 153375794-153375794 on Build GRCh38
rs1040754250 Chr.1: 153375829-153375829 on Build GRCh38
rs544438617 Chr.1: 153375520-153375520 on Build GRCh38
rs930813089 Chr.1: 153375703-153375703 on Build GRCh38
rs548071788 Chr.1: 153375664-153375664 on Build GRCh38
rs1033230258 Chr.1: 153375486-153375486 on Build GRCh38
rs1047919013 Chr.1: 153375712-153375712 on Build GRCh38
rs533517429 Chr.1: 153375530-153375530 on Build GRCh38
rs1051918039 Chr.1: 153375559-153375559 on Build GRCh38
rs549360720 Chr.1: 153375814-153375814 on Build GRCh38
rs946286230 Chr.1: 153375576-153375576 on Build GRCh38
rs765034402 Chr.1: 153375909-153375909 on Build GRCh38
rs937056119 Chr.1: 153375565-153375565 on Build GRCh38
rs926995021 Chr.1: 153375557-153375557 on Build GRCh38
rs4323654 Chr.1: 153375857-153375857 on Build GRCh38
rs761875920 Chr.1: 153375713-153375713 on Build GRCh38
rs988683450 Chr.1: 153375501-153375501 on Build GRCh38
rs919703433 Chr.1: 153375613-153375613 on Build GRCh38
rs754799468 Chr.1: 153375844-153375844 on Build GRCh38
rs1024773943 Chr.1: 153375731-153375731 on Build GRCh38
rs973834355 Chr.1: 153375603-153375603 on Build GRCh38
rs957174154 Chr.1: 153375488-153375488 on Build GRCh38
rs766361390 Chr.1: 153375756-153375756 on Build GRCh38
rs1000474084 Chr.1: 153375690-153375690 on Build GRCh38
rs868070292 Chr.1: 153375848-153375848 on Build GRCh38
rs564500835 Chr.1: 153375521-153375521 on Build GRCh38

View Full Product Details