Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153947568-153948069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTC2
Entrez Gene ID
200186
Gene Name
CREB regulated transcription coactivator 2
Gene Aliases
TORC-2, TORC2
UniGene
Hs.406392
Gene Chromosome Location
Chr.1: 153947672-153958656 on Build GRCh38
Transcripts
NM_181715.2 XM_005244946.1 XM_005244949.2 XM_017000575.1 XM_017000576.1 XM_017000577.1 XM_017000578.1 XM_017000579.1

SNP Detail

SNP ID Chromosome Location
rs1030485580 Chr.1: 153947656-153947659 on Build GRCh38
rs777887046 Chr.1: 153947601-153947601 on Build GRCh38
rs879219938 Chr.1: 153948010-153948010 on Build GRCh38
rs41312716 Chr.1: 153947716-153947716 on Build GRCh38
rs1018232459 Chr.1: 153947608-153947608 on Build GRCh38
rs564104982 Chr.1: 153947876-153947876 on Build GRCh38
rs576072816 Chr.1: 153947930-153947930 on Build GRCh38
rs373365245 Chr.1: 153947595-153947595 on Build GRCh38
rs548651195 Chr.1: 153947685-153947685 on Build GRCh38
rs866788240 Chr.1: 153947804-153947804 on Build GRCh38
rs1017443166 Chr.1: 153947787-153947787 on Build GRCh38
rs569695534 Chr.1: 153947603-153947603 on Build GRCh38
rs892072398 Chr.1: 153947709-153947709 on Build GRCh38
rs999608733 Chr.1: 153947669-153947669 on Build GRCh38
rs866473467 Chr.1: 153947749-153947749 on Build GRCh38
rs561719140 Chr.1: 153948030-153948030 on Build GRCh38
rs979117990 Chr.1: 153947723-153947723 on Build GRCh38
rs780768791 Chr.1: 153947883-153947883 on Build GRCh38
rs887777658 Chr.1: 153947855-153947855 on Build GRCh38
rs555409369 Chr.1: 153947707-153947707 on Build GRCh38
rs987826225 Chr.1: 153947691-153947691 on Build GRCh38
rs937931325 Chr.1: 153947752-153947752 on Build GRCh38
rs925040756 Chr.1: 153947958-153947958 on Build GRCh38
rs962715227 Chr.1: 153947803-153947803 on Build GRCh38
rs769440799 Chr.1: 153947863-153947864 on Build GRCh38
rs1053274722 Chr.1: 153947697-153947697 on Build GRCh38
rs8450 Chr.1: 153947810-153947810 on Build GRCh38
rs543562633 Chr.1: 153947981-153947981 on Build GRCh38
rs578217139 Chr.1: 153947825-153947825 on Build GRCh38
rs537186102 Chr.1: 153947647-153947647 on Build GRCh38
rs992426700 Chr.1: 153947761-153947761 on Build GRCh38
rs934992802 Chr.1: 153947991-153947991 on Build GRCh38
rs1030476295 Chr.1: 153947959-153947959 on Build GRCh38
rs545681447 Chr.1: 153947871-153947871 on Build GRCh38
rs534860862 Chr.1: 153947740-153947740 on Build GRCh38
rs1001588165 Chr.1: 153947776-153947776 on Build GRCh38
rs571177423 Chr.1: 153948007-153948007 on Build GRCh38
rs964986276 Chr.1: 153947638-153947643 on Build GRCh38
rs942534244 Chr.1: 153947931-153947931 on Build GRCh38
rs979173512 Chr.1: 153947954-153947954 on Build GRCh38
rs913582711 Chr.1: 153947703-153947707 on Build GRCh38
rs1047812479 Chr.1: 153947812-153947812 on Build GRCh38
rs986628820 Chr.1: 153947880-153947880 on Build GRCh38
rs942063934 Chr.1: 153947872-153947872 on Build GRCh38
rs998125034 Chr.1: 153947650-153947650 on Build GRCh38

View Full Product Details