Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41935959-41936466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCND3
Entrez Gene ID
896
Gene Name
cyclin D3
Gene Aliases
-
UniGene
Hs.534307
Gene Chromosome Location
Chr.6: 41934933-42048894 on Build GRCh38
Transcripts
NM_001136017.3 NM_001136125.2 NM_001136126.2 NM_001287427.1 NM_001287434.1 NM_001760.4 XM_011514971.1 XM_011514972.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6205892 c.798_819del22 p.P267fs*>19 Chr.6: 41936000-41936021 on Build GRCh38
220538 c.826C>T p.Q276* Chr.6: 41935993-41935993 on Build GRCh38
6211076 c.766_776del11 p.R256fs*>34 Chr.6: 41936043-41936053 on Build GRCh38
5019335 c.759G>T p.E253D Chr.6: 41936060-41936060 on Build GRCh38
220540 c.860T>A p.V287D Chr.6: 41935959-41935959 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759741362 Chr.6: 41935995-41935995 on Build GRCh38
rs752366531 Chr.6: 41936144-41936144 on Build GRCh38
rs369861674 Chr.6: 41936063-41936063 on Build GRCh38
rs565876880 Chr.6: 41936185-41936185 on Build GRCh38
rs1007313936 Chr.6: 41936182-41936182 on Build GRCh38
rs33966734 Chr.6: 41936060-41936060 on Build GRCh38
rs200046302 Chr.6: 41936008-41936008 on Build GRCh38
rs3218102 Chr.6: 41936045-41936045 on Build GRCh38
rs928317031 Chr.6: 41936318-41936318 on Build GRCh38
rs972515253 Chr.6: 41936334-41936335 on Build GRCh38
rs147958536 Chr.6: 41936078-41936078 on Build GRCh38
rs775643743 Chr.6: 41936062-41936062 on Build GRCh38
rs956805243 Chr.6: 41936013-41936013 on Build GRCh38
rs202197675 Chr.6: 41936162-41936162 on Build GRCh38
rs748692236 Chr.6: 41936034-41936034 on Build GRCh38
rs3218101 Chr.6: 41936146-41936146 on Build GRCh38
rs1018563511 Chr.6: 41936238-41936238 on Build GRCh38
rs760616733 Chr.6: 41936111-41936111 on Build GRCh38
rs568279708 Chr.6: 41936239-41936239 on Build GRCh38
rs549926134 Chr.6: 41936121-41936121 on Build GRCh38
rs140718705 Chr.6: 41936225-41936225 on Build GRCh38
rs201507759 Chr.6: 41936116-41936116 on Build GRCh38
rs1000782967 Chr.6: 41936288-41936288 on Build GRCh38
rs375843578 Chr.6: 41936021-41936021 on Build GRCh38
rs749415129 Chr.6: 41936420-41936420 on Build GRCh38
rs148137451 Chr.6: 41936245-41936245 on Build GRCh38
rs777265467 Chr.6: 41935982-41935982 on Build GRCh38
rs760760322 Chr.6: 41936012-41936017 on Build GRCh38
rs764155094 Chr.6: 41936007-41936007 on Build GRCh38
rs527923415 Chr.6: 41936012-41936012 on Build GRCh38
rs760015142 Chr.6: 41936000-41936000 on Build GRCh38
rs781708411 Chr.6: 41936147-41936147 on Build GRCh38
rs1048515528 Chr.6: 41936401-41936401 on Build GRCh38
rs956774949 Chr.6: 41936306-41936306 on Build GRCh38
rs1025224393 Chr.6: 41936389-41936389 on Build GRCh38
rs891319807 Chr.6: 41936192-41936192 on Build GRCh38
rs999610686 Chr.6: 41936208-41936208 on Build GRCh38
rs1031472507 Chr.6: 41936234-41936234 on Build GRCh38
rs1051130 Chr.6: 41936044-41936044 on Build GRCh38
rs545840762 Chr.6: 41936297-41936297 on Build GRCh38
rs563728371 Chr.6: 41936143-41936143 on Build GRCh38
rs540997482 Chr.6: 41936098-41936098 on Build GRCh38
rs1021567233 Chr.6: 41936257-41936257 on Build GRCh38
rs761632994 Chr.6: 41936081-41936081 on Build GRCh38
rs377286500 Chr.6: 41936148-41936148 on Build GRCh38
rs113738889 Chr.6: 41936320-41936320 on Build GRCh38
rs779634736 Chr.6: 41936033-41936033 on Build GRCh38
rs386700585 Chr.6: 41936044-41936045 on Build GRCh38
rs990076942 Chr.6: 41936024-41936024 on Build GRCh38
rs372911419 Chr.6: 41936003-41936003 on Build GRCh38
rs539178499 Chr.6: 41936396-41936396 on Build GRCh38
rs373093172 Chr.6: 41936156-41936156 on Build GRCh38
rs376278514 Chr.6: 41936413-41936413 on Build GRCh38
rs754436542 Chr.6: 41936125-41936125 on Build GRCh38
rs767328984 Chr.6: 41936097-41936097 on Build GRCh38
rs34598973 Chr.6: 41936204-41936205 on Build GRCh38
rs762030113 Chr.6: 41936443-41936443 on Build GRCh38
rs777320361 Chr.6: 41936046-41936046 on Build GRCh38
rs746409401 Chr.6: 41936056-41936056 on Build GRCh38
rs775393073 Chr.6: 41936002-41936002 on Build GRCh38
rs145920031 Chr.6: 41936226-41936226 on Build GRCh38
rs756023182 Chr.6: 41936173-41936173 on Build GRCh38
rs763029528 Chr.6: 41936074-41936074 on Build GRCh38
rs142862109 Chr.6: 41936022-41936022 on Build GRCh38
rs756000746 Chr.6: 41936028-41936028 on Build GRCh38
rs754668264 Chr.6: 41936036-41936036 on Build GRCh38
rs879501541 Chr.6: 41936044-41936045 on Build GRCh38
rs970619720 Chr.6: 41936442-41936442 on Build GRCh38
rs989960939 Chr.6: 41936240-41936240 on Build GRCh38

View Full Product Details