Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42207928-42208436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPS10
Entrez Gene ID
55173
Gene Name
mitochondrial ribosomal protein S10
Gene Aliases
MRP-S10, PNAS-122
UniGene
Hs.380887
Gene Chromosome Location
Chr.6: 42206801-42217907 on Build GRCh38
Transcripts
NM_018141.3 XM_005249206.2 XM_011514724.2 XM_011514725.2 XM_017010996.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4393318 c.602C>T p.S201L Chr.6: 42208293-42208293 on Build GRCh38
6715437 c.523-3delT p.? Chr.6: 42208375-42208375 on Build GRCh38
6216299 c.592G>T p.E198* Chr.6: 42208303-42208303 on Build GRCh38
7258227 c.574C>A p.L192I Chr.6: 42208321-42208321 on Build GRCh38
6251315 c.564C>A p.I188I Chr.6: 42208331-42208331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773553706 Chr.6: 42208386-42208386 on Build GRCh38
rs150834133 Chr.6: 42208031-42208031 on Build GRCh38
rs550757486 Chr.6: 42208337-42208337 on Build GRCh38
rs756763607 Chr.6: 42208371-42208371 on Build GRCh38
rs752557282 Chr.6: 42208405-42208405 on Build GRCh38
rs372365869 Chr.6: 42208077-42208077 on Build GRCh38
rs757528246 Chr.6: 42208359-42208360 on Build GRCh38
rs777302702 Chr.6: 42208239-42208239 on Build GRCh38
rs776904279 Chr.6: 42208397-42208397 on Build GRCh38
rs758331610 Chr.6: 42208359-42208359 on Build GRCh38
rs9471805 Chr.6: 42208172-42208172 on Build GRCh38
rs746687007 Chr.6: 42208241-42208241 on Build GRCh38
rs1008490669 Chr.6: 42208125-42208125 on Build GRCh38
rs767535033 Chr.6: 42208302-42208302 on Build GRCh38
rs746247324 Chr.6: 42208034-42208034 on Build GRCh38
rs772655733 Chr.6: 42208384-42208384 on Build GRCh38
rs771457195 Chr.6: 42208284-42208290 on Build GRCh38
rs1045653402 Chr.6: 42208283-42208283 on Build GRCh38
rs535925150 Chr.6: 42208010-42208010 on Build GRCh38
rs1038132454 Chr.6: 42208052-42208052 on Build GRCh38
rs959472833 Chr.6: 42208216-42208216 on Build GRCh38
rs1046031989 Chr.6: 42207995-42207995 on Build GRCh38
rs2295278 Chr.6: 42208382-42208382 on Build GRCh38
rs746544710 Chr.6: 42208370-42208370 on Build GRCh38
rs754001754 Chr.6: 42208333-42208333 on Build GRCh38
rs149751323 Chr.6: 42208336-42208336 on Build GRCh38
rs776380099 Chr.6: 42208273-42208273 on Build GRCh38
rs188762459 Chr.6: 42208285-42208285 on Build GRCh38
rs781009007 Chr.6: 42208374-42208374 on Build GRCh38
rs577849219 Chr.6: 42208112-42208112 on Build GRCh38
rs766433117 Chr.6: 42208396-42208396 on Build GRCh38
rs764986997 Chr.6: 42208403-42208403 on Build GRCh38
rs979605498 Chr.6: 42208182-42208182 on Build GRCh38
rs113113702 Chr.6: 42208318-42208318 on Build GRCh38
rs746229043 Chr.6: 42208369-42208370 on Build GRCh38
rs533661682 Chr.6: 42208380-42208380 on Build GRCh38
rs769089129 Chr.6: 42208276-42208276 on Build GRCh38
rs950027105 Chr.6: 42207958-42207958 on Build GRCh38
rs575469841 Chr.6: 42208014-42208014 on Build GRCh38
rs11618 Chr.6: 42208189-42208189 on Build GRCh38
rs140242341 Chr.6: 42208345-42208345 on Build GRCh38
rs768337922 Chr.6: 42208401-42208401 on Build GRCh38
rs955198448 Chr.6: 42208094-42208094 on Build GRCh38
rs766619745 Chr.6: 42208327-42208327 on Build GRCh38
rs139604808 Chr.6: 42208056-42208056 on Build GRCh38
rs566812257 Chr.6: 42208392-42208392 on Build GRCh38
rs555605966 Chr.6: 42208013-42208013 on Build GRCh38
rs547456048 Chr.6: 42208066-42208066 on Build GRCh38
rs1021587683 Chr.6: 42208166-42208166 on Build GRCh38
rs764350885 Chr.6: 42208299-42208302 on Build GRCh38
rs564652372 Chr.6: 42208353-42208353 on Build GRCh38
rs199933095 Chr.6: 42208319-42208319 on Build GRCh38
rs770726801 Chr.6: 42208243-42208243 on Build GRCh38
rs542266910 Chr.6: 42208195-42208195 on Build GRCh38
rs762380857 Chr.6: 42208287-42208287 on Build GRCh38
rs560061668 Chr.6: 42208176-42208176 on Build GRCh38
rs917938048 Chr.6: 42208258-42208258 on Build GRCh38
rs375259938 Chr.6: 42208367-42208367 on Build GRCh38
rs370468409 Chr.6: 42208400-42208400 on Build GRCh38

View Full Product Details