Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43171152-43171676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRF
Entrez Gene ID
6722
Gene Name
serum response factor
Gene Aliases
MCM1
UniGene
Hs.520140
Gene Chromosome Location
Chr.6: 43171295-43181506 on Build GRCh38
Transcripts
NM_003131.3

SNP Detail

SNP ID Chromosome Location
rs1010856336 Chr.6: 43171291-43171291 on Build GRCh38
rs994876410 Chr.6: 43171442-43171442 on Build GRCh38
rs906012078 Chr.6: 43171329-43171329 on Build GRCh38
rs895586554 Chr.6: 43171569-43171569 on Build GRCh38
rs532892350 Chr.6: 43171305-43171305 on Build GRCh38
rs765957223 Chr.6: 43171251-43171251 on Build GRCh38
rs970185564 Chr.6: 43171309-43171309 on Build GRCh38
rs950668549 Chr.6: 43171486-43171486 on Build GRCh38
rs970428621 Chr.6: 43171297-43171297 on Build GRCh38
rs973339692 Chr.6: 43171317-43171317 on Build GRCh38
rs922055283 Chr.6: 43171539-43171539 on Build GRCh38
rs1027322729 Chr.6: 43171485-43171485 on Build GRCh38
rs1039123220 Chr.6: 43171351-43171351 on Build GRCh38
rs942533108 Chr.6: 43171377-43171378 on Build GRCh38
rs959731428 Chr.6: 43171228-43171228 on Build GRCh38
rs550547236 Chr.6: 43171296-43171296 on Build GRCh38
rs760651185 Chr.6: 43171356-43171356 on Build GRCh38
rs911743660 Chr.6: 43171341-43171341 on Build GRCh38
rs936039233 Chr.6: 43171357-43171357 on Build GRCh38
rs1047797712 Chr.6: 43171200-43171200 on Build GRCh38
rs973095419 Chr.6: 43171244-43171244 on Build GRCh38
rs564257131 Chr.6: 43171299-43171299 on Build GRCh38
rs891761645 Chr.6: 43171383-43171383 on Build GRCh38
rs983400815 Chr.6: 43171487-43171487 on Build GRCh38
rs894803627 Chr.6: 43171404-43171404 on Build GRCh38
rs952646463 Chr.6: 43171348-43171348 on Build GRCh38
rs944763871 Chr.6: 43171347-43171347 on Build GRCh38
rs373311819 Chr.6: 43171334-43171334 on Build GRCh38
rs994295909 Chr.6: 43171597-43171597 on Build GRCh38
rs546251468 Chr.6: 43171339-43171339 on Build GRCh38
rs369845610 Chr.6: 43171256-43171256 on Build GRCh38
rs771615419 Chr.6: 43171652-43171652 on Build GRCh38
rs1046297949 Chr.6: 43171384-43171384 on Build GRCh38
rs1018719817 Chr.6: 43171500-43171500 on Build GRCh38
rs767122177 Chr.6: 43171289-43171289 on Build GRCh38
rs535118405 Chr.6: 43171609-43171609 on Build GRCh38
rs959048580 Chr.6: 43171307-43171307 on Build GRCh38
rs566210658 Chr.6: 43171397-43171397 on Build GRCh38
rs960122456 Chr.6: 43171345-43171345 on Build GRCh38
rs892009945 Chr.6: 43171378-43171378 on Build GRCh38
rs759053881 Chr.6: 43171282-43171282 on Build GRCh38
rs965873714 Chr.6: 43171505-43171505 on Build GRCh38
rs1001596910 Chr.6: 43171313-43171313 on Build GRCh38
rs946324848 Chr.6: 43171381-43171381 on Build GRCh38
rs1055140164 Chr.6: 43171276-43171276 on Build GRCh38
rs911929393 Chr.6: 43171223-43171223 on Build GRCh38
rs929168660 Chr.6: 43171192-43171192 on Build GRCh38
rs530822769 Chr.6: 43171261-43171261 on Build GRCh38
rs1033319937 Chr.6: 43171303-43171303 on Build GRCh38
rs900523229 Chr.6: 43171257-43171257 on Build GRCh38
rs1021761811 Chr.6: 43171195-43171195 on Build GRCh38
rs1003273382 Chr.6: 43171391-43171391 on Build GRCh38
rs1053493770 Chr.6: 43171304-43171304 on Build GRCh38
rs1053143217 Chr.6: 43171382-43171382 on Build GRCh38
rs979836683 Chr.6: 43171209-43171209 on Build GRCh38
rs1011812044 Chr.6: 43171310-43171310 on Build GRCh38
rs747601402 Chr.6: 43171650-43171650 on Build GRCh38
rs920540577 Chr.6: 43171325-43171325 on Build GRCh38
rs542221739 Chr.6: 43171226-43171226 on Build GRCh38
rs1024486253 Chr.6: 43171292-43171292 on Build GRCh38
rs532949406 Chr.6: 43171179-43171179 on Build GRCh38
rs1054707466 Chr.6: 43171375-43171375 on Build GRCh38
rs907385747 Chr.6: 43171388-43171388 on Build GRCh38
rs972430268 Chr.6: 43171343-43171343 on Build GRCh38
rs943483544 Chr.6: 43171289-43171290 on Build GRCh38
rs1000391147 Chr.6: 43171272-43171272 on Build GRCh38
rs561868372 Chr.6: 43171258-43171258 on Build GRCh38
rs1035755335 Chr.6: 43171342-43171342 on Build GRCh38
rs947641797 Chr.6: 43171396-43171396 on Build GRCh38
rs974796628 Chr.6: 43171510-43171510 on Build GRCh38
rs1057474299 Chr.6: 43171514-43171514 on Build GRCh38

View Full Product Details