Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43519787-43520302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR1C
Entrez Gene ID
9533
Gene Name
RNA polymerase I subunit C
Gene Aliases
AC40, HLD11, RPA39, RPA40, RPA5, RPAC1, RPC40, TCS3
UniGene
Hs.743509
Gene Chromosome Location
Chr.6: 43517024-43562404 on Build GRCh38
Transcripts
NM_001318876.1 NM_203290.3 XM_011515000.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5897596 c.503-7C>T p.? Chr.6: 43520268-43520268 on Build GRCh38
5996552 c.359G>A p.R120H Chr.6: 43519815-43519815 on Build GRCh38
7099577 c.400G>C p.E134Q Chr.6: 43520083-43520083 on Build GRCh38
246708 c.490G>A p.V164M Chr.6: 43520173-43520173 on Build GRCh38
7283510 c.528delC p.L178fs*76 Chr.6: 43520300-43520300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956091160 Chr.6: 43520217-43520217 on Build GRCh38
rs925937062 Chr.6: 43520214-43520214 on Build GRCh38
rs964946378 Chr.6: 43519958-43519958 on Build GRCh38
rs533111548 Chr.6: 43520108-43520108 on Build GRCh38
rs753326212 Chr.6: 43519815-43519815 on Build GRCh38
rs750727358 Chr.6: 43520105-43520105 on Build GRCh38
rs745325916 Chr.6: 43520163-43520163 on Build GRCh38
rs989283512 Chr.6: 43520235-43520235 on Build GRCh38
rs1017737027 Chr.6: 43520169-43520169 on Build GRCh38
rs746689926 Chr.6: 43520152-43520152 on Build GRCh38
rs374405229 Chr.6: 43519823-43519823 on Build GRCh38
rs143381942 Chr.6: 43520029-43520029 on Build GRCh38
rs773388338 Chr.6: 43520185-43520185 on Build GRCh38
rs184442171 Chr.6: 43519995-43519995 on Build GRCh38
rs757588820 Chr.6: 43519843-43519843 on Build GRCh38
rs796052125 Chr.6: 43520119-43520119 on Build GRCh38
rs114815047 Chr.6: 43519938-43519938 on Build GRCh38
rs141582886 Chr.6: 43520259-43520259 on Build GRCh38
rs553478354 Chr.6: 43520136-43520138 on Build GRCh38
rs779991362 Chr.6: 43520020-43520020 on Build GRCh38
rs954903590 Chr.6: 43519914-43519914 on Build GRCh38
rs755099052 Chr.6: 43520202-43520202 on Build GRCh38
rs987743903 Chr.6: 43519926-43519927 on Build GRCh38
rs202138719 Chr.6: 43520222-43520222 on Build GRCh38
rs374806421 Chr.6: 43520073-43520073 on Build GRCh38
rs146162408 Chr.6: 43520135-43520135 on Build GRCh38
rs756279219 Chr.6: 43520024-43520025 on Build GRCh38
rs767689208 Chr.6: 43520133-43520133 on Build GRCh38
rs765044240 Chr.6: 43520046-43520046 on Build GRCh38
rs987667455 Chr.6: 43519928-43519928 on Build GRCh38
rs778679379 Chr.6: 43520043-43520043 on Build GRCh38
rs200847882 Chr.6: 43520209-43520209 on Build GRCh38
rs144195949 Chr.6: 43520126-43520126 on Build GRCh38
rs187997427 Chr.6: 43520172-43520172 on Build GRCh38
rs751906432 Chr.6: 43519829-43519829 on Build GRCh38
rs762290929 Chr.6: 43520097-43520097 on Build GRCh38
rs148385032 Chr.6: 43520104-43520104 on Build GRCh38
rs545889680 Chr.6: 43520092-43520092 on Build GRCh38
rs751274069 Chr.6: 43520226-43520226 on Build GRCh38
rs766490023 Chr.6: 43520115-43520115 on Build GRCh38
rs950125476 Chr.6: 43520271-43520271 on Build GRCh38
rs763542757 Chr.6: 43519825-43519825 on Build GRCh38
rs940947633 Chr.6: 43520089-43520089 on Build GRCh38
rs758655268 Chr.6: 43520204-43520204 on Build GRCh38
rs760951790 Chr.6: 43520189-43520189 on Build GRCh38
rs543544898 Chr.6: 43519974-43519974 on Build GRCh38
rs146332364 Chr.6: 43519814-43519814 on Build GRCh38
rs771262971 Chr.6: 43520278-43520278 on Build GRCh38
rs779700234 Chr.6: 43520254-43520254 on Build GRCh38
rs1008942133 Chr.6: 43520186-43520186 on Build GRCh38
rs774882134 Chr.6: 43520095-43520095 on Build GRCh38
rs746639153 Chr.6: 43520260-43520261 on Build GRCh38
rs752745376 Chr.6: 43520210-43520210 on Build GRCh38
rs776311990 Chr.6: 43520158-43520158 on Build GRCh38
rs567744785 Chr.6: 43520268-43520268 on Build GRCh38
rs199534486 Chr.6: 43520231-43520231 on Build GRCh38
rs767980525 Chr.6: 43520177-43520177 on Build GRCh38
rs1002131319 Chr.6: 43520040-43520040 on Build GRCh38
rs548176283 Chr.6: 43520260-43520260 on Build GRCh38
rs979075107 Chr.6: 43520213-43520213 on Build GRCh38
rs994826443 Chr.6: 43520166-43520166 on Build GRCh38
rs750595496 Chr.6: 43520153-43520153 on Build GRCh38
rs911725106 Chr.6: 43519927-43519927 on Build GRCh38
rs369805980 Chr.6: 43520200-43520200 on Build GRCh38
rs778773104 Chr.6: 43520132-43520132 on Build GRCh38
rs913724338 Chr.6: 43520003-43520003 on Build GRCh38
rs781430489 Chr.6: 43519865-43519865 on Build GRCh38
rs374732135 Chr.6: 43520028-43520028 on Build GRCh38
rs745488555 Chr.6: 43520241-43520241 on Build GRCh38
rs1055452309 Chr.6: 43520025-43520025 on Build GRCh38
rs769498204 Chr.6: 43520252-43520252 on Build GRCh38
rs377163139 Chr.6: 43520192-43520192 on Build GRCh38
rs573140963 Chr.6: 43520265-43520265 on Build GRCh38
rs760899351 Chr.6: 43520275-43520275 on Build GRCh38
rs756957074 Chr.6: 43520229-43520229 on Build GRCh38
rs201320592 Chr.6: 43520078-43520078 on Build GRCh38
rs776224077 Chr.6: 43520081-43520081 on Build GRCh38
rs906664643 Chr.6: 43520080-43520080 on Build GRCh38
rs753845398 Chr.6: 43520121-43520121 on Build GRCh38
rs764567148 Chr.6: 43519827-43519827 on Build GRCh38
rs993300804 Chr.6: 43519811-43519811 on Build GRCh38
rs769410222 Chr.6: 43520164-43520164 on Build GRCh38
rs777418804 Chr.6: 43520146-43520146 on Build GRCh38
rs979138638 Chr.6: 43519963-43519963 on Build GRCh38
rs777630577 Chr.6: 43520218-43520218 on Build GRCh38
rs949775448 Chr.6: 43520064-43520064 on Build GRCh38
rs1053240800 Chr.6: 43519996-43519996 on Build GRCh38
rs776258335 Chr.6: 43520008-43520008 on Build GRCh38
rs953450571 Chr.6: 43519887-43519887 on Build GRCh38
rs748490341 Chr.6: 43519812-43519812 on Build GRCh38
rs200525225 Chr.6: 43520173-43520173 on Build GRCh38
rs756260977 Chr.6: 43520016-43520016 on Build GRCh38
rs369733205 Chr.6: 43519885-43519885 on Build GRCh38
rs755323652 Chr.6: 43520173-43520185 on Build GRCh38
rs771643954 Chr.6: 43520072-43520072 on Build GRCh38
rs779191389 Chr.6: 43520194-43520195 on Build GRCh38
rs758212801 Chr.6: 43520216-43520216 on Build GRCh38
rs371870708 Chr.6: 43520086-43520086 on Build GRCh38
rs770857344 Chr.6: 43520143-43520144 on Build GRCh38

View Full Product Details