Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44139345-44139849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM63B
Entrez Gene ID
55362
Gene Name
transmembrane protein 63B
Gene Aliases
C6orf110
UniGene
Hs.414473
Gene Chromosome Location
Chr.6: 44126628-44155519 on Build GRCh38
Transcripts
NM_001318792.1 NM_018426.2 XM_005249211.3 XM_005249213.4 XM_005249214.4 XM_005249215.4 XM_005249217.1 XM_006715135.1 XM_017010999.1 XM_017011000.1 XM_017011001.1 XM_017011002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5890249 c.506C>T p.S169F Chr.6: 44139565-44139565 on Build GRCh38
40701 c.437G>A p.G146D Chr.6: 44139496-44139496 on Build GRCh38
6262502 c.478G>T p.G160W Chr.6: 44139537-44139537 on Build GRCh38
1621702 c.602+10G>A p.? Chr.6: 44139769-44139769 on Build GRCh38
6107036 c.463C>T p.Q155* Chr.6: 44139522-44139522 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs72858118 Chr.6: 44139822-44139822 on Build GRCh38
rs771850110 Chr.6: 44139764-44139764 on Build GRCh38
rs759062304 Chr.6: 44139795-44139795 on Build GRCh38
rs758969385 Chr.6: 44139573-44139573 on Build GRCh38
rs762999319 Chr.6: 44139537-44139537 on Build GRCh38
rs370893644 Chr.6: 44139526-44139526 on Build GRCh38
rs778328775 Chr.6: 44139593-44139593 on Build GRCh38
rs752080550 Chr.6: 44139797-44139797 on Build GRCh38
rs771116222 Chr.6: 44139509-44139509 on Build GRCh38
rs774083434 Chr.6: 44139545-44139545 on Build GRCh38
rs770073031 Chr.6: 44139618-44139618 on Build GRCh38
rs367570916 Chr.6: 44139687-44139687 on Build GRCh38
rs754453969 Chr.6: 44139676-44139676 on Build GRCh38
rs765312141 Chr.6: 44139658-44139659 on Build GRCh38
rs749415382 Chr.6: 44139715-44139715 on Build GRCh38
rs778002704 Chr.6: 44139505-44139505 on Build GRCh38
rs756460767 Chr.6: 44139707-44139707 on Build GRCh38
rs371639085 Chr.6: 44139693-44139693 on Build GRCh38
rs764833657 Chr.6: 44139796-44139796 on Build GRCh38
rs200839192 Chr.6: 44139557-44139557 on Build GRCh38
rs201583875 Chr.6: 44139670-44139670 on Build GRCh38
rs146291875 Chr.6: 44139467-44139467 on Build GRCh38
rs376492794 Chr.6: 44139515-44139515 on Build GRCh38
rs752129994 Chr.6: 44139685-44139685 on Build GRCh38
rs775889969 Chr.6: 44139666-44139666 on Build GRCh38
rs746122920 Chr.6: 44139606-44139606 on Build GRCh38
rs764059807 Chr.6: 44139540-44139540 on Build GRCh38
rs35174285 Chr.6: 44139452-44139452 on Build GRCh38
rs747373916 Chr.6: 44139596-44139596 on Build GRCh38
rs748124949 Chr.6: 44139741-44139741 on Build GRCh38
rs767380854 Chr.6: 44139656-44139656 on Build GRCh38
rs181880322 Chr.6: 44139414-44139414 on Build GRCh38
rs751312777 Chr.6: 44139457-44139457 on Build GRCh38
rs41282654 Chr.6: 44139628-44139628 on Build GRCh38
rs773051560 Chr.6: 44139765-44139765 on Build GRCh38
rs764557964 Chr.6: 44139682-44139682 on Build GRCh38
rs776640775 Chr.6: 44139446-44139446 on Build GRCh38
rs780776879 Chr.6: 44139769-44139769 on Build GRCh38
rs577510167 Chr.6: 44139390-44139390 on Build GRCh38
rs1039982875 Chr.6: 44139409-44139409 on Build GRCh38
rs755585313 Chr.6: 44139469-44139469 on Build GRCh38
rs199973630 Chr.6: 44139458-44139458 on Build GRCh38
rs575197570 Chr.6: 44139413-44139413 on Build GRCh38
rs780416151 Chr.6: 44139709-44139709 on Build GRCh38
rs774309333 Chr.6: 44139634-44139634 on Build GRCh38
rs1052230253 Chr.6: 44139818-44139818 on Build GRCh38
rs549073978 Chr.6: 44139635-44139635 on Build GRCh38
rs981025839 Chr.6: 44139524-44139524 on Build GRCh38
rs187298481 Chr.6: 44139622-44139622 on Build GRCh38
rs374498822 Chr.6: 44139730-44139730 on Build GRCh38
rs757664730 Chr.6: 44139691-44139691 on Build GRCh38
rs771116731 Chr.6: 44139445-44139445 on Build GRCh38
rs11967633 Chr.6: 44139463-44139463 on Build GRCh38
rs761659717 Chr.6: 44139546-44139546 on Build GRCh38
rs139340075 Chr.6: 44139768-44139768 on Build GRCh38
rs1003871005 Chr.6: 44139400-44139400 on Build GRCh38
rs200560167 Chr.6: 44139583-44139583 on Build GRCh38
rs757620173 Chr.6: 44139599-44139599 on Build GRCh38
rs750117918 Chr.6: 44139558-44139558 on Build GRCh38
rs766705588 Chr.6: 44139406-44139406 on Build GRCh38
rs747393854 Chr.6: 44139506-44139506 on Build GRCh38
rs1052232690 Chr.6: 44139668-44139668 on Build GRCh38
rs145356402 Chr.6: 44139566-44139566 on Build GRCh38
rs781361985 Chr.6: 44139605-44139605 on Build GRCh38
rs144454929 Chr.6: 44139525-44139525 on Build GRCh38
rs202216036 Chr.6: 44139677-44139677 on Build GRCh38
rs766005257 Chr.6: 44139672-44139672 on Build GRCh38
rs753414791 Chr.6: 44139675-44139675 on Build GRCh38
rs756345908 Chr.6: 44139481-44139481 on Build GRCh38
rs1015295133 Chr.6: 44139437-44139437 on Build GRCh38
rs73733839 Chr.6: 44139455-44139455 on Build GRCh38
rs199984650 Chr.6: 44139497-44139497 on Build GRCh38
rs745312021 Chr.6: 44139779-44139779 on Build GRCh38
rs201619627 Chr.6: 44139483-44139483 on Build GRCh38
rs746175796 Chr.6: 44139704-44139704 on Build GRCh38
rs373121259 Chr.6: 44139448-44139448 on Build GRCh38
rs891044582 Chr.6: 44139380-44139380 on Build GRCh38
rs201285466 Chr.6: 44139778-44139778 on Build GRCh38
rs749362302 Chr.6: 44139627-44139627 on Build GRCh38
rs62401767 Chr.6: 44139527-44139527 on Build GRCh38
rs117623146 Chr.6: 44139454-44139454 on Build GRCh38
rs768754994 Chr.6: 44139722-44139722 on Build GRCh38
rs765884785 Chr.6: 44139567-44139567 on Build GRCh38

View Full Product Details