Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44139629-44140134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM63B
Entrez Gene ID
55362
Gene Name
transmembrane protein 63B
Gene Aliases
C6orf110
UniGene
Hs.414473
Gene Chromosome Location
Chr.6: 44126628-44155519 on Build GRCh38
Transcripts
NM_001318792.1 NM_018426.2 XM_005249211.3 XM_005249213.4 XM_005249214.4 XM_005249215.4 XM_005249217.1 XM_006715135.1 XM_017010999.1 XM_017011000.1 XM_017011001.1 XM_017011002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6027242 c.602+9C>T p.? Chr.6: 44139768-44139768 on Build GRCh38
1621702 c.602+10G>A p.? Chr.6: 44139769-44139769 on Build GRCh38
742837 c.552G>C p.E184D Chr.6: 44139709-44139709 on Build GRCh38
4842459 c.575G>C p.R192T Chr.6: 44139732-44139732 on Build GRCh38
6804186 c.587C>T p.A196V Chr.6: 44139744-44139744 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs72858118 Chr.6: 44139822-44139822 on Build GRCh38
rs771850110 Chr.6: 44139764-44139764 on Build GRCh38
rs191872389 Chr.6: 44139865-44139865 on Build GRCh38
rs759062304 Chr.6: 44139795-44139795 on Build GRCh38
rs34358384 Chr.6: 44139984-44139985 on Build GRCh38
rs775350758 Chr.6: 44139835-44139835 on Build GRCh38
rs752080550 Chr.6: 44139797-44139797 on Build GRCh38
rs963556671 Chr.6: 44139932-44139932 on Build GRCh38
rs555157399 Chr.6: 44140077-44140077 on Build GRCh38
rs949526635 Chr.6: 44140000-44140000 on Build GRCh38
rs925217403 Chr.6: 44139951-44139951 on Build GRCh38
rs367570916 Chr.6: 44139687-44139687 on Build GRCh38
rs776782906 Chr.6: 44140068-44140068 on Build GRCh38
rs1025083516 Chr.6: 44139861-44139861 on Build GRCh38
rs754453969 Chr.6: 44139676-44139676 on Build GRCh38
rs765312141 Chr.6: 44139658-44139659 on Build GRCh38
rs756460767 Chr.6: 44139707-44139707 on Build GRCh38
rs749415382 Chr.6: 44139715-44139715 on Build GRCh38
rs371639085 Chr.6: 44139693-44139693 on Build GRCh38
rs940851976 Chr.6: 44140021-44140021 on Build GRCh38
rs764833657 Chr.6: 44139796-44139796 on Build GRCh38
rs919417464 Chr.6: 44139970-44139970 on Build GRCh38
rs966582882 Chr.6: 44139890-44139890 on Build GRCh38
rs759465489 Chr.6: 44140069-44140069 on Build GRCh38
rs936654142 Chr.6: 44139962-44139962 on Build GRCh38
rs201583875 Chr.6: 44139670-44139670 on Build GRCh38
rs10807295 Chr.6: 44139903-44139903 on Build GRCh38
rs949183719 Chr.6: 44140109-44140109 on Build GRCh38
rs150548237 Chr.6: 44140073-44140073 on Build GRCh38
rs752129994 Chr.6: 44139685-44139685 on Build GRCh38
rs532129913 Chr.6: 44140031-44140031 on Build GRCh38
rs10755724 Chr.6: 44139875-44139875 on Build GRCh38
rs765290853 Chr.6: 44140084-44140084 on Build GRCh38
rs775889969 Chr.6: 44139666-44139666 on Build GRCh38
rs916660694 Chr.6: 44139995-44139995 on Build GRCh38
rs748124949 Chr.6: 44139741-44139741 on Build GRCh38
rs995587748 Chr.6: 44139843-44139843 on Build GRCh38
rs767380854 Chr.6: 44139656-44139656 on Build GRCh38
rs1013332148 Chr.6: 44139859-44139859 on Build GRCh38
rs773051560 Chr.6: 44139765-44139765 on Build GRCh38
rs764557964 Chr.6: 44139682-44139682 on Build GRCh38
rs780776879 Chr.6: 44139769-44139769 on Build GRCh38
rs1046580857 Chr.6: 44140003-44140003 on Build GRCh38
rs780416151 Chr.6: 44139709-44139709 on Build GRCh38
rs1052230253 Chr.6: 44139818-44139818 on Build GRCh38
rs374498822 Chr.6: 44139730-44139730 on Build GRCh38
rs757664730 Chr.6: 44139691-44139691 on Build GRCh38
rs139340075 Chr.6: 44139768-44139768 on Build GRCh38
rs991266255 Chr.6: 44139987-44139987 on Build GRCh38
rs1028034275 Chr.6: 44139844-44139844 on Build GRCh38
rs1052232690 Chr.6: 44139668-44139668 on Build GRCh38
rs573253973 Chr.6: 44139956-44139956 on Build GRCh38
rs954998762 Chr.6: 44140008-44140008 on Build GRCh38
rs202216036 Chr.6: 44139677-44139677 on Build GRCh38
rs35031546 Chr.6: 44139880-44139881 on Build GRCh38
rs556807169 Chr.6: 44139868-44139868 on Build GRCh38
rs753414791 Chr.6: 44139675-44139675 on Build GRCh38
rs766005257 Chr.6: 44139672-44139672 on Build GRCh38
rs555084904 Chr.6: 44139950-44139950 on Build GRCh38
rs745312021 Chr.6: 44139779-44139779 on Build GRCh38
rs746175796 Chr.6: 44139704-44139704 on Build GRCh38
rs910573319 Chr.6: 44140033-44140033 on Build GRCh38
rs201285466 Chr.6: 44139778-44139778 on Build GRCh38
rs953970330 Chr.6: 44139845-44139845 on Build GRCh38
rs10948125 Chr.6: 44140061-44140061 on Build GRCh38
rs943397437 Chr.6: 44140040-44140040 on Build GRCh38
rs768754994 Chr.6: 44139722-44139722 on Build GRCh38

View Full Product Details