Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 45898776-45899316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC5
Entrez Gene ID
53405
Gene Name
chloride intracellular channel 5
Gene Aliases
DFNB102, DFNB103, MST130, MSTP130
UniGene
Hs.485489
Gene Chromosome Location
Chr.6: 45898451-46129809 on Build GRCh38
Transcripts
NM_001114086.1 NM_016929.4 NR_045672.1 NR_045673.1 XM_017010953.1

SNP Detail

SNP ID Chromosome Location
rs776977378 Chr.6: 45898890-45898890 on Build GRCh38
rs1035737536 Chr.6: 45898803-45898803 on Build GRCh38
rs1019961591 Chr.6: 45898827-45898827 on Build GRCh38
rs764644548 Chr.6: 45899131-45899131 on Build GRCh38
rs943017780 Chr.6: 45898950-45898950 on Build GRCh38
rs922969587 Chr.6: 45899210-45899210 on Build GRCh38
rs978524015 Chr.6: 45898875-45898875 on Build GRCh38
rs1128514 Chr.6: 45898945-45898945 on Build GRCh38
rs964686702 Chr.6: 45899182-45899182 on Build GRCh38
rs115050913 Chr.6: 45899249-45899249 on Build GRCh38
rs759003478 Chr.6: 45899118-45899118 on Build GRCh38
rs908438773 Chr.6: 45899283-45899283 on Build GRCh38
rs760075482 Chr.6: 45899025-45899025 on Build GRCh38
rs933105092 Chr.6: 45898971-45898971 on Build GRCh38
rs368212823 Chr.6: 45899095-45899095 on Build GRCh38
rs1037801549 Chr.6: 45899011-45899011 on Build GRCh38
rs1038680365 Chr.6: 45898954-45898954 on Build GRCh38
rs886423931 Chr.6: 45898989-45898989 on Build GRCh38
rs550414631 Chr.6: 45898931-45898931 on Build GRCh38
rs912981173 Chr.6: 45899281-45899281 on Build GRCh38
rs149527163 Chr.6: 45899080-45899080 on Build GRCh38
rs958667422 Chr.6: 45899197-45899197 on Build GRCh38
rs576805522 Chr.6: 45898952-45898952 on Build GRCh38
rs1032766212 Chr.6: 45899186-45899186 on Build GRCh38
rs148651951 Chr.6: 45899162-45899162 on Build GRCh38
rs1035726248 Chr.6: 45899057-45899057 on Build GRCh38
rs1001532182 Chr.6: 45899052-45899052 on Build GRCh38
rs111674768 Chr.6: 45899001-45899001 on Build GRCh38
rs766400573 Chr.6: 45898890-45898890 on Build GRCh38
rs899219397 Chr.6: 45899035-45899035 on Build GRCh38
rs763037111 Chr.6: 45898875-45898885 on Build GRCh38
rs949993117 Chr.6: 45899277-45899277 on Build GRCh38
rs994274316 Chr.6: 45899115-45899115 on Build GRCh38
rs72871404 Chr.6: 45899240-45899240 on Build GRCh38
rs112079441 Chr.6: 45899147-45899147 on Build GRCh38
rs1011837686 Chr.6: 45899029-45899029 on Build GRCh38
rs1052871406 Chr.6: 45899006-45899006 on Build GRCh38
rs767889021 Chr.6: 45899148-45899148 on Build GRCh38
rs546506435 Chr.6: 45899234-45899234 on Build GRCh38
rs940717972 Chr.6: 45899285-45899285 on Build GRCh38
rs530692009 Chr.6: 45898850-45898850 on Build GRCh38
rs1042944302 Chr.6: 45899039-45899039 on Build GRCh38
rs574589698 Chr.6: 45899053-45899053 on Build GRCh38
rs61002683 Chr.6: 45899040-45899040 on Build GRCh38
rs192949899 Chr.6: 45898819-45898819 on Build GRCh38
rs984367895 Chr.6: 45898942-45898942 on Build GRCh38
rs923022006 Chr.6: 45898955-45898955 on Build GRCh38
rs60094266 Chr.6: 45899007-45899007 on Build GRCh38
rs952884513 Chr.6: 45899120-45899120 on Build GRCh38
rs539100341 Chr.6: 45898975-45898975 on Build GRCh38
rs549765114 Chr.6: 45899251-45899251 on Build GRCh38
rs996360681 Chr.6: 45899037-45899037 on Build GRCh38
rs933052718 Chr.6: 45899236-45899236 on Build GRCh38
rs561704433 Chr.6: 45899279-45899279 on Build GRCh38
rs182866866 Chr.6: 45898891-45898891 on Build GRCh38
rs925668321 Chr.6: 45898934-45898934 on Build GRCh38
rs974373406 Chr.6: 45899208-45899208 on Build GRCh38

View Full Product Details