Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 46160979-46161519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPP5
Entrez Gene ID
59084
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative)
Gene Aliases
NPP-5, NPP5
UniGene
Hs.35198
Gene Chromosome Location
Chr.6: 46159182-46171045 on Build GRCh38
Transcripts
NM_001290072.1 NM_001290073.1 NM_021572.5 XM_005249260.3 XM_011514786.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3430710 c.1375C>A p.Q459K Chr.6: 46161385-46161385 on Build GRCh38
451575 c.1365A>C p.L455F Chr.6: 46161395-46161395 on Build GRCh38
6846647 c.1362T>G p.H454Q Chr.6: 46161398-46161398 on Build GRCh38
7261528 c.1289C>T p.S430L Chr.6: 46161471-46161471 on Build GRCh38
6993363 c.1248C>T p.L416L Chr.6: 46161512-46161512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749084146 Chr.6: 46161275-46161275 on Build GRCh38
rs565098925 Chr.6: 46161191-46161191 on Build GRCh38
rs554825669 Chr.6: 46161147-46161147 on Build GRCh38
rs147979315 Chr.6: 46161436-46161436 on Build GRCh38
rs145274280 Chr.6: 46161264-46161264 on Build GRCh38
rs768371818 Chr.6: 46161471-46161471 on Build GRCh38
rs760365040 Chr.6: 46161346-46161346 on Build GRCh38
rs189746500 Chr.6: 46161183-46161183 on Build GRCh38
rs769546954 Chr.6: 46161276-46161276 on Build GRCh38
rs939372935 Chr.6: 46161120-46161120 on Build GRCh38
rs1049105693 Chr.6: 46161163-46161163 on Build GRCh38
rs968298256 Chr.6: 46161359-46161359 on Build GRCh38
rs115235333 Chr.6: 46161301-46161301 on Build GRCh38
rs74445007 Chr.6: 46161173-46161173 on Build GRCh38
rs1053821556 Chr.6: 46161295-46161295 on Build GRCh38
rs577066582 Chr.6: 46161196-46161196 on Build GRCh38
rs771615362 Chr.6: 46161334-46161334 on Build GRCh38
rs73735712 Chr.6: 46161145-46161145 on Build GRCh38
rs140279334 Chr.6: 46161058-46161058 on Build GRCh38
rs761741298 Chr.6: 46161390-46161390 on Build GRCh38
rs915055982 Chr.6: 46161050-46161050 on Build GRCh38
rs745808391 Chr.6: 46161122-46161122 on Build GRCh38
rs546925640 Chr.6: 46161451-46161451 on Build GRCh38
rs67079261 Chr.6: 46161174-46161175 on Build GRCh38
rs1056377666 Chr.6: 46161132-46161132 on Build GRCh38
rs929379528 Chr.6: 46161162-46161162 on Build GRCh38
rs868115354 Chr.6: 46161236-46161236 on Build GRCh38
rs6941587 Chr.6: 46161230-46161230 on Build GRCh38
rs756673571 Chr.6: 46161449-46161449 on Build GRCh38
rs777263460 Chr.6: 46161407-46161408 on Build GRCh38
rs1012520580 Chr.6: 46161304-46161304 on Build GRCh38
rs781760020 Chr.6: 46161284-46161284 on Build GRCh38
rs939264788 Chr.6: 46161424-46161424 on Build GRCh38
rs1047153 Chr.6: 46161008-46161008 on Build GRCh38
rs557058138 Chr.6: 46161079-46161079 on Build GRCh38
rs771656751 Chr.6: 46161456-46161456 on Build GRCh38
rs751861066 Chr.6: 46161433-46161433 on Build GRCh38
rs766479290 Chr.6: 46161415-46161415 on Build GRCh38
rs760030741 Chr.6: 46161415-46161416 on Build GRCh38
rs181765859 Chr.6: 46161208-46161208 on Build GRCh38
rs369152079 Chr.6: 46161478-46161478 on Build GRCh38
rs751109599 Chr.6: 46161038-46161038 on Build GRCh38
rs926512950 Chr.6: 46161091-46161091 on Build GRCh38
rs750490393 Chr.6: 46161404-46161404 on Build GRCh38
rs776510857 Chr.6: 46161364-46161364 on Build GRCh38
rs745439501 Chr.6: 46161312-46161312 on Build GRCh38
rs746704534 Chr.6: 46161467-46161467 on Build GRCh38
rs1041531636 Chr.6: 46161218-46161218 on Build GRCh38
rs755252981 Chr.6: 46161435-46161435 on Build GRCh38
rs973130337 Chr.6: 46161067-46161067 on Build GRCh38
rs778572614 Chr.6: 46161310-46161310 on Build GRCh38
rs375879187 Chr.6: 46161458-46161458 on Build GRCh38
rs371294052 Chr.6: 46161437-46161437 on Build GRCh38
rs575125750 Chr.6: 46161104-46161104 on Build GRCh38
rs200453493 Chr.6: 46161360-46161360 on Build GRCh38
rs573173945 Chr.6: 46161179-46161179 on Build GRCh38
rs1029732502 Chr.6: 46161247-46161247 on Build GRCh38
rs1022010461 Chr.6: 46161352-46161352 on Build GRCh38
rs988024869 Chr.6: 46161042-46161042 on Build GRCh38
rs35054026 Chr.6: 46161173-46161174 on Build GRCh38
rs775244700 Chr.6: 46161342-46161342 on Build GRCh38
rs997878102 Chr.6: 46161244-46161244 on Build GRCh38
rs760943125 Chr.6: 46161396-46161396 on Build GRCh38
rs560495507 Chr.6: 46161309-46161309 on Build GRCh38
rs992159341 Chr.6: 46161425-46161425 on Build GRCh38
rs2235882 Chr.6: 46161057-46161057 on Build GRCh38
rs771201120 Chr.6: 46161316-46161323 on Build GRCh38

View Full Product Details