Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 46168069-46168573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPP5
Entrez Gene ID
59084
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative)
Gene Aliases
NPP-5, NPP5
UniGene
Hs.35198
Gene Chromosome Location
Chr.6: 46159182-46171045 on Build GRCh38
Transcripts
NM_001290072.1 NM_001290073.1 NM_021572.5 XM_005249260.3 XM_011514786.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1697309 c.178C>T p.H60Y Chr.6: 46168085-46168085 on Build GRCh38
5928795 c.98T>A p.V33D Chr.6: 46168165-46168165 on Build GRCh38
1697310 c.154C>T p.H52Y Chr.6: 46168109-46168109 on Build GRCh38
5959167 c.149A>G p.H50R Chr.6: 46168114-46168114 on Build GRCh38
5603475 c.101C>T p.S34F Chr.6: 46168162-46168162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957837370 Chr.6: 46168427-46168427 on Build GRCh38
rs915252799 Chr.6: 46168423-46168423 on Build GRCh38
rs146025312 Chr.6: 46168121-46168121 on Build GRCh38
rs771162160 Chr.6: 46168119-46168119 on Build GRCh38
rs768931482 Chr.6: 46168303-46168303 on Build GRCh38
rs372747904 Chr.6: 46168219-46168219 on Build GRCh38
rs778734822 Chr.6: 46168310-46168310 on Build GRCh38
rs939544559 Chr.6: 46168472-46168472 on Build GRCh38
rs144047978 Chr.6: 46168191-46168191 on Build GRCh38
rs772476966 Chr.6: 46168130-46168130 on Build GRCh38
rs34109856 Chr.6: 46168147-46168147 on Build GRCh38
rs778095361 Chr.6: 46168182-46168182 on Build GRCh38
rs190774314 Chr.6: 46168532-46168532 on Build GRCh38
rs777128942 Chr.6: 46168314-46168314 on Build GRCh38
rs777614014 Chr.6: 46168105-46168105 on Build GRCh38
rs751154937 Chr.6: 46168173-46168173 on Build GRCh38
rs374777514 Chr.6: 46168120-46168120 on Build GRCh38
rs200392605 Chr.6: 46168230-46168230 on Build GRCh38
rs554889165 Chr.6: 46168269-46168269 on Build GRCh38
rs926710200 Chr.6: 46168471-46168471 on Build GRCh38
rs749367453 Chr.6: 46168115-46168115 on Build GRCh38
rs779118415 Chr.6: 46168254-46168254 on Build GRCh38
rs78793102 Chr.6: 46168388-46168388 on Build GRCh38
rs987980465 Chr.6: 46168378-46168378 on Build GRCh38
rs1017999968 Chr.6: 46168232-46168232 on Build GRCh38
rs762485026 Chr.6: 46168155-46168155 on Build GRCh38
rs374042338 Chr.6: 46168433-46168433 on Build GRCh38
rs754899541 Chr.6: 46168130-46168133 on Build GRCh38
rs993756484 Chr.6: 46168506-46168506 on Build GRCh38
rs755915456 Chr.6: 46168228-46168228 on Build GRCh38
rs771070665 Chr.6: 46168249-46168249 on Build GRCh38
rs759847474 Chr.6: 46168129-46168129 on Build GRCh38
rs765876878 Chr.6: 46168167-46168167 on Build GRCh38
rs935245858 Chr.6: 46168352-46168352 on Build GRCh38
rs751656431 Chr.6: 46168315-46168316 on Build GRCh38
rs560326190 Chr.6: 46168434-46168434 on Build GRCh38
rs970767060 Chr.6: 46168453-46168453 on Build GRCh38
rs1032500013 Chr.6: 46168331-46168331 on Build GRCh38
rs934204620 Chr.6: 46168311-46168311 on Build GRCh38
rs769767805 Chr.6: 46168100-46168100 on Build GRCh38
rs577220991 Chr.6: 46168485-46168485 on Build GRCh38
rs775915886 Chr.6: 46168133-46168133 on Build GRCh38
rs746007899 Chr.6: 46168255-46168255 on Build GRCh38
rs1023314505 Chr.6: 46168436-46168436 on Build GRCh38
rs999729076 Chr.6: 46168275-46168275 on Build GRCh38
rs1056627917 Chr.6: 46168476-46168476 on Build GRCh38
rs747348756 Chr.6: 46168287-46168287 on Build GRCh38
rs752508122 Chr.6: 46168221-46168221 on Build GRCh38
rs3806995 Chr.6: 46168247-46168247 on Build GRCh38
rs142176948 Chr.6: 46168148-46168148 on Build GRCh38

View Full Product Details