Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 46170708-46171217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPP5
Entrez Gene ID
59084
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative)
Gene Aliases
NPP-5, NPP5
UniGene
Hs.35198
Gene Chromosome Location
Chr.6: 46159182-46171045 on Build GRCh38
Transcripts
NM_001290072.1 NM_001290073.1 NM_021572.5 XM_005249260.3 XM_011514786.2

SNP Detail

SNP ID Chromosome Location
rs983276301 Chr.6: 46170928-46170928 on Build GRCh38
rs900516968 Chr.6: 46170957-46170957 on Build GRCh38
rs73735714 Chr.6: 46171149-46171149 on Build GRCh38
rs1020525474 Chr.6: 46170893-46170893 on Build GRCh38
rs116032216 Chr.6: 46171095-46171095 on Build GRCh38
rs901487277 Chr.6: 46170929-46170929 on Build GRCh38
rs1010397787 Chr.6: 46170988-46170988 on Build GRCh38
rs1026562315 Chr.6: 46171045-46171046 on Build GRCh38
rs892947781 Chr.6: 46170781-46170781 on Build GRCh38
rs922941417 Chr.6: 46171111-46171111 on Build GRCh38
rs907831212 Chr.6: 46170932-46170932 on Build GRCh38
rs547924119 Chr.6: 46170954-46170954 on Build GRCh38
rs74545502 Chr.6: 46170918-46170918 on Build GRCh38
rs3822892 Chr.6: 46170878-46170878 on Build GRCh38
rs917655523 Chr.6: 46170921-46170921 on Build GRCh38
rs878999564 Chr.6: 46170906-46170906 on Build GRCh38
rs1041755650 Chr.6: 46171191-46171191 on Build GRCh38
rs568677268 Chr.6: 46171146-46171146 on Build GRCh38
rs865780145 Chr.6: 46170901-46170901 on Build GRCh38
rs1032430684 Chr.6: 46170907-46170907 on Build GRCh38
rs1037591789 Chr.6: 46170947-46170947 on Build GRCh38
rs960963046 Chr.6: 46170877-46170877 on Build GRCh38
rs1031528822 Chr.6: 46170764-46170764 on Build GRCh38
rs1051710812 Chr.6: 46171184-46171184 on Build GRCh38
rs941947872 Chr.6: 46170934-46170934 on Build GRCh38
rs959321273 Chr.6: 46171080-46171080 on Build GRCh38
rs1016908390 Chr.6: 46171130-46171130 on Build GRCh38
rs1044260637 Chr.6: 46171134-46171134 on Build GRCh38
rs914530430 Chr.6: 46171119-46171119 on Build GRCh38
rs1025593085 Chr.6: 46170902-46170902 on Build GRCh38
rs758102599 Chr.6: 46171174-46171174 on Build GRCh38
rs980829671 Chr.6: 46170848-46170849 on Build GRCh38
rs532018192 Chr.6: 46171120-46171120 on Build GRCh38
rs1056434578 Chr.6: 46171153-46171153 on Build GRCh38
rs1052769078 Chr.6: 46171115-46171115 on Build GRCh38
rs369205185 Chr.6: 46171025-46171025 on Build GRCh38
rs1011417653 Chr.6: 46170814-46170814 on Build GRCh38
rs747503668 Chr.6: 46170750-46170750 on Build GRCh38
rs371717891 Chr.6: 46170883-46170883 on Build GRCh38
rs907719627 Chr.6: 46171132-46171132 on Build GRCh38
rs946142943 Chr.6: 46171188-46171188 on Build GRCh38
rs527319019 Chr.6: 46171011-46171011 on Build GRCh38
rs562347089 Chr.6: 46170899-46170899 on Build GRCh38
rs781636396 Chr.6: 46170963-46170963 on Build GRCh38
rs540496214 Chr.6: 46170837-46170837 on Build GRCh38
rs554352838 Chr.6: 46171180-46171180 on Build GRCh38
rs931979734 Chr.6: 46170964-46170964 on Build GRCh38
rs964069786 Chr.6: 46171139-46171139 on Build GRCh38
rs952386811 Chr.6: 46171047-46171047 on Build GRCh38
rs1034996627 Chr.6: 46171002-46171002 on Build GRCh38
rs1022340958 Chr.6: 46170829-46170829 on Build GRCh38
rs949229675 Chr.6: 46170926-46170926 on Build GRCh38
rs374882505 Chr.6: 46170912-46170912 on Build GRCh38
rs947279649 Chr.6: 46171130-46171131 on Build GRCh38
rs572388465 Chr.6: 46170791-46170791 on Build GRCh38
rs941847130 Chr.6: 46171135-46171135 on Build GRCh38
rs1038107353 Chr.6: 46170845-46170845 on Build GRCh38
rs1005531951 Chr.6: 46170924-46170924 on Build GRCh38
rs998756078 Chr.6: 46170753-46170753 on Build GRCh38
rs576952993 Chr.6: 46170884-46170884 on Build GRCh38
rs542285496 Chr.6: 46171088-46171088 on Build GRCh38
rs745561044 Chr.6: 46171038-46171038 on Build GRCh38
rs993712872 Chr.6: 46171030-46171030 on Build GRCh38
rs1044614257 Chr.6: 46170958-46170962 on Build GRCh38
rs890531206 Chr.6: 46170968-46170968 on Build GRCh38
rs996673918 Chr.6: 46170867-46170867 on Build GRCh38
rs959040165 Chr.6: 46170916-46170916 on Build GRCh38
rs373168647 Chr.6: 46170896-46170896 on Build GRCh38
rs1020410693 Chr.6: 46170992-46170992 on Build GRCh38
rs886343318 Chr.6: 46170830-46170830 on Build GRCh38
rs369579314 Chr.6: 46171186-46171186 on Build GRCh38
rs904665760 Chr.6: 46170998-46170998 on Build GRCh38
rs921857369 Chr.6: 46171152-46171152 on Build GRCh38
rs900463724 Chr.6: 46170848-46170848 on Build GRCh38
rs111890908 Chr.6: 46171096-46171096 on Build GRCh38
rs1047701681 Chr.6: 46170794-46170794 on Build GRCh38
rs934276466 Chr.6: 46171112-46171112 on Build GRCh38
rs559716372 Chr.6: 46170969-46170969 on Build GRCh38
rs1052308888 Chr.6: 46170965-46170965 on Build GRCh38

View Full Product Details