Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 46716261-46716769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G7
Entrez Gene ID
7941
Gene Name
phospholipase A2 group VII
Gene Aliases
LDL-PLA2, LP-PLA2, PAFAD, PAFAH
UniGene
Hs.584823
Gene Chromosome Location
Chr.6: 46700954-46735836 on Build GRCh38
Transcripts
NM_001168357.1 NM_005084.3 XM_005249408.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7188150 c.329T>C p.F110S Chr.6: 46716431-46716431 on Build GRCh38
1312276 c.286C>A p.L96I Chr.6: 46716474-46716474 on Build GRCh38
4714925 c.245G>A p.R82H Chr.6: 46716515-46716515 on Build GRCh38
6872249 c.286C>T p.L96F Chr.6: 46716474-46716474 on Build GRCh38
4981408 c.363G>A p.L121L Chr.6: 46716397-46716397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899354628 Chr.6: 46716707-46716707 on Build GRCh38
rs765174107 Chr.6: 46716611-46716611 on Build GRCh38
rs758684014 Chr.6: 46716410-46716410 on Build GRCh38
rs763882543 Chr.6: 46716390-46716390 on Build GRCh38
rs748660580 Chr.6: 46716436-46716436 on Build GRCh38
rs752486194 Chr.6: 46716385-46716385 on Build GRCh38
rs753600819 Chr.6: 46716537-46716537 on Build GRCh38
rs1022231309 Chr.6: 46716625-46716626 on Build GRCh38
rs1026037266 Chr.6: 46716727-46716727 on Build GRCh38
rs936641315 Chr.6: 46716403-46716403 on Build GRCh38
rs753624242 Chr.6: 46716457-46716457 on Build GRCh38
rs200126173 Chr.6: 46716351-46716351 on Build GRCh38
rs779027670 Chr.6: 46716399-46716399 on Build GRCh38
rs767091605 Chr.6: 46716444-46716444 on Build GRCh38
rs151292733 Chr.6: 46716313-46716313 on Build GRCh38
rs960468078 Chr.6: 46716712-46716712 on Build GRCh38
rs748522672 Chr.6: 46716494-46716494 on Build GRCh38
rs199793731 Chr.6: 46716510-46716510 on Build GRCh38
rs143012249 Chr.6: 46716431-46716431 on Build GRCh38
rs1011987129 Chr.6: 46716416-46716416 on Build GRCh38
rs184940944 Chr.6: 46716469-46716469 on Build GRCh38
rs778258079 Chr.6: 46716507-46716507 on Build GRCh38
rs767216223 Chr.6: 46716379-46716379 on Build GRCh38
rs759114384 Chr.6: 46716443-46716443 on Build GRCh38
rs1036922546 Chr.6: 46716698-46716698 on Build GRCh38
rs374356451 Chr.6: 46716395-46716395 on Build GRCh38
rs200782042 Chr.6: 46716417-46716417 on Build GRCh38
rs527379312 Chr.6: 46716694-46716694 on Build GRCh38
rs776478288 Chr.6: 46716518-46716518 on Build GRCh38
rs368234407 Chr.6: 46716509-46716509 on Build GRCh38
rs372505803 Chr.6: 46716536-46716536 on Build GRCh38
rs759169916 Chr.6: 46716376-46716376 on Build GRCh38
rs768974261 Chr.6: 46716357-46716357 on Build GRCh38
rs758789308 Chr.6: 46716401-46716401 on Build GRCh38
rs191270134 Chr.6: 46716414-46716414 on Build GRCh38
rs144962369 Chr.6: 46716551-46716551 on Build GRCh38
rs200649164 Chr.6: 46716560-46716560 on Build GRCh38
rs138830410 Chr.6: 46716600-46716600 on Build GRCh38
rs746001962 Chr.6: 46716409-46716409 on Build GRCh38
rs200298077 Chr.6: 46716482-46716482 on Build GRCh38
rs936561152 Chr.6: 46716535-46716535 on Build GRCh38
rs866437639 Chr.6: 46716442-46716442 on Build GRCh38
rs562765483 Chr.6: 46716526-46716526 on Build GRCh38
rs200052639 Chr.6: 46716438-46716438 on Build GRCh38
rs560026957 Chr.6: 46716647-46716647 on Build GRCh38
rs755943605 Chr.6: 46716388-46716388 on Build GRCh38
rs747316945 Chr.6: 46716429-46716429 on Build GRCh38
rs759429602 Chr.6: 46716569-46716569 on Build GRCh38
rs140551939 Chr.6: 46716373-46716373 on Build GRCh38
rs765208515 Chr.6: 46716525-46716525 on Build GRCh38
rs865913859 Chr.6: 46716407-46716407 on Build GRCh38
rs775138182 Chr.6: 46716447-46716447 on Build GRCh38
rs150471892 Chr.6: 46716442-46716442 on Build GRCh38
rs775984119 Chr.6: 46716341-46716341 on Build GRCh38
rs372380410 Chr.6: 46716334-46716334 on Build GRCh38
rs201253286 Chr.6: 46716502-46716502 on Build GRCh38
rs923815733 Chr.6: 46716383-46716383 on Build GRCh38
rs750458482 Chr.6: 46716471-46716471 on Build GRCh38
rs971793902 Chr.6: 46716722-46716722 on Build GRCh38
rs556394219 Chr.6: 46716306-46716306 on Build GRCh38
rs755215220 Chr.6: 46716564-46716564 on Build GRCh38
rs930240375 Chr.6: 46716506-46716506 on Build GRCh38
rs202100699 Chr.6: 46716478-46716478 on Build GRCh38
rs1805017 Chr.6: 46716485-46716485 on Build GRCh38
rs143106370 Chr.6: 46716516-46716516 on Build GRCh38
rs765947390 Chr.6: 46716368-46716368 on Build GRCh38
rs541982618 Chr.6: 46716645-46716645 on Build GRCh38
rs201777305 Chr.6: 46716484-46716484 on Build GRCh38
rs144983904 Chr.6: 46716515-46716515 on Build GRCh38
rs766537476 Chr.6: 46716545-46716545 on Build GRCh38
rs1001910447 Chr.6: 46716671-46716671 on Build GRCh38
rs1053442004 Chr.6: 46716413-46716413 on Build GRCh38
rs865957744 Chr.6: 46716527-46716527 on Build GRCh38
rs760340474 Chr.6: 46716448-46716448 on Build GRCh38
rs763916484 Chr.6: 46716455-46716455 on Build GRCh38
rs200593058 Chr.6: 46716335-46716335 on Build GRCh38
rs780371256 Chr.6: 46716476-46716476 on Build GRCh38
rs758562180 Chr.6: 46716474-46716474 on Build GRCh38
rs201604552 Chr.6: 46716566-46716566 on Build GRCh38
rs74470054 Chr.6: 46716364-46716364 on Build GRCh38
rs747441548 Chr.6: 46716350-46716350 on Build GRCh38
rs761517434 Chr.6: 46716523-46716523 on Build GRCh38
rs940482656 Chr.6: 46716596-46716596 on Build GRCh38
rs150329930 Chr.6: 46716432-46716432 on Build GRCh38
rs189781341 Chr.6: 46716486-46716486 on Build GRCh38
rs930209067 Chr.6: 46716302-46716302 on Build GRCh38
rs906270883 Chr.6: 46716570-46716570 on Build GRCh38
rs201835224 Chr.6: 46716346-46716346 on Build GRCh38
rs777206289 Chr.6: 46716360-46716360 on Build GRCh38

View Full Product Details