Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 47573901-47574400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD2AP
Entrez Gene ID
23607
Gene Name
CD2 associated protein
Gene Aliases
CMS
UniGene
Hs.485518
Gene Chromosome Location
Chr.6: 47477746-47627263 on Build GRCh38
Transcripts
NM_012120.2 XM_005248976.1 XM_011514449.2 XM_017010641.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3628099 c.634C>T p.R212* Chr.6: 47574156-47574156 on Build GRCh38
4670442 c.630A>C p.K210N Chr.6: 47574152-47574152 on Build GRCh38
400040 c.558G>T p.G186G Chr.6: 47574080-47574080 on Build GRCh38
5528361 c.611C>T p.S204L Chr.6: 47574133-47574133 on Build GRCh38
7000257 c.661A>G p.K221E Chr.6: 47574183-47574183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756161725 Chr.6: 47574243-47574243 on Build GRCh38
rs754966889 Chr.6: 47574157-47574157 on Build GRCh38
rs1049658994 Chr.6: 47574314-47574314 on Build GRCh38
rs146384979 Chr.6: 47574344-47574345 on Build GRCh38
rs1055892576 Chr.6: 47573974-47573974 on Build GRCh38
rs764430418 Chr.6: 47574140-47574140 on Build GRCh38
rs775987563 Chr.6: 47574278-47574278 on Build GRCh38
rs1038175950 Chr.6: 47574209-47574209 on Build GRCh38
rs141242954 Chr.6: 47574188-47574188 on Build GRCh38
rs746327167 Chr.6: 47574269-47574269 on Build GRCh38
rs766301489 Chr.6: 47574215-47574215 on Build GRCh38
rs142643033 Chr.6: 47574075-47574075 on Build GRCh38
rs762800934 Chr.6: 47574207-47574207 on Build GRCh38
rs752880361 Chr.6: 47574229-47574229 on Build GRCh38
rs143630674 Chr.6: 47573927-47573927 on Build GRCh38
rs1009384125 Chr.6: 47574001-47574001 on Build GRCh38
rs377722480 Chr.6: 47574015-47574015 on Build GRCh38
rs753380875 Chr.6: 47574104-47574104 on Build GRCh38
rs758392587 Chr.6: 47574128-47574128 on Build GRCh38
rs977576473 Chr.6: 47574080-47574080 on Build GRCh38
rs776679863 Chr.6: 47574025-47574025 on Build GRCh38
rs150959557 Chr.6: 47574242-47574242 on Build GRCh38
rs996699348 Chr.6: 47574228-47574228 on Build GRCh38
rs762237115 Chr.6: 47574369-47574369 on Build GRCh38
rs150851309 Chr.6: 47574204-47574204 on Build GRCh38
rs535153702 Chr.6: 47574376-47574376 on Build GRCh38
rs999358581 Chr.6: 47574362-47574362 on Build GRCh38
rs1045885305 Chr.6: 47574193-47574193 on Build GRCh38
rs771234513 Chr.6: 47574260-47574260 on Build GRCh38
rs761363856 Chr.6: 47574200-47574200 on Build GRCh38
rs769463446 Chr.6: 47574289-47574289 on Build GRCh38
rs781129940 Chr.6: 47574227-47574227 on Build GRCh38
rs907053121 Chr.6: 47573958-47573958 on Build GRCh38
rs760110147 Chr.6: 47574100-47574100 on Build GRCh38
rs146010044 Chr.6: 47574158-47574158 on Build GRCh38
rs756790635 Chr.6: 47574205-47574205 on Build GRCh38
rs138163915 Chr.6: 47574073-47574073 on Build GRCh38
rs373030099 Chr.6: 47574084-47574084 on Build GRCh38
rs764906767 Chr.6: 47574116-47574116 on Build GRCh38
rs999078911 Chr.6: 47574111-47574111 on Build GRCh38
rs889107828 Chr.6: 47574320-47574320 on Build GRCh38
rs201232118 Chr.6: 47574164-47574164 on Build GRCh38
rs144554172 Chr.6: 47574302-47574302 on Build GRCh38
rs774798567 Chr.6: 47574179-47574179 on Build GRCh38
rs771289707 Chr.6: 47574168-47574168 on Build GRCh38
rs973720364 Chr.6: 47574303-47574303 on Build GRCh38
rs536276372 Chr.6: 47574354-47574354 on Build GRCh38
rs749595817 Chr.6: 47574165-47574165 on Build GRCh38
rs546274903 Chr.6: 47573983-47573983 on Build GRCh38
rs140627775 Chr.6: 47574218-47574218 on Build GRCh38
rs147211221 Chr.6: 47573964-47573964 on Build GRCh38
rs749542658 Chr.6: 47574254-47574254 on Build GRCh38
rs542265189 Chr.6: 47573932-47573932 on Build GRCh38
rs368107857 Chr.6: 47574262-47574262 on Build GRCh38
rs774852008 Chr.6: 47574091-47574091 on Build GRCh38
rs867687839 Chr.6: 47574093-47574093 on Build GRCh38
rs770034676 Chr.6: 47574031-47574031 on Build GRCh38
rs776242797 Chr.6: 47574195-47574195 on Build GRCh38
rs777127972 Chr.6: 47574016-47574016 on Build GRCh38
rs149993734 Chr.6: 47574224-47574224 on Build GRCh38
rs751286070 Chr.6: 47574105-47574105 on Build GRCh38
rs749616791 Chr.6: 47574279-47574279 on Build GRCh38
rs864622037 Chr.6: 47574082-47574082 on Build GRCh38
rs763690363 Chr.6: 47574102-47574102 on Build GRCh38
rs1008938735 Chr.6: 47574335-47574335 on Build GRCh38
rs1019365367 Chr.6: 47574336-47574336 on Build GRCh38
rs772700438 Chr.6: 47574272-47574272 on Build GRCh38
rs750307379 Chr.6: 47574118-47574118 on Build GRCh38
rs564923108 Chr.6: 47573997-47573997 on Build GRCh38
rs908328150 Chr.6: 47574197-47574197 on Build GRCh38
rs528212824 Chr.6: 47573977-47573977 on Build GRCh38
rs139926926 Chr.6: 47574182-47574182 on Build GRCh38
rs770123305 Chr.6: 47574160-47574160 on Build GRCh38
rs371396990 Chr.6: 47574029-47574029 on Build GRCh38
rs938724619 Chr.6: 47573966-47573966 on Build GRCh38
rs777904060 Chr.6: 47574251-47574251 on Build GRCh38

View Full Product Details