Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 47579094-47579624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD2AP
Entrez Gene ID
23607
Gene Name
CD2 associated protein
Gene Aliases
CMS
UniGene
Hs.485518
Gene Chromosome Location
Chr.6: 47477746-47627263 on Build GRCh38
Transcripts
NM_012120.2 XM_005248976.1 XM_011514449.2 XM_017010641.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
139648 c.973G>T p.A325S Chr.6: 47579454-47579454 on Build GRCh38
5202443 c.997_998delAA p.K333fs*13 Chr.6: 47579478-47579479 on Build GRCh38
6398345 c.905A>G p.E302G Chr.6: 47579386-47579386 on Build GRCh38
4403863 c.909T>C p.T303T Chr.6: 47579390-47579390 on Build GRCh38
266205 c.968A>T p.N323I Chr.6: 47579449-47579449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199995766 Chr.6: 47579371-47579371 on Build GRCh38
rs759885985 Chr.6: 47579504-47579504 on Build GRCh38
rs779152482 Chr.6: 47579522-47579531 on Build GRCh38
rs918990573 Chr.6: 47579335-47579336 on Build GRCh38
rs769681987 Chr.6: 47579478-47579478 on Build GRCh38
rs778481750 Chr.6: 47579406-47579406 on Build GRCh38
rs754370307 Chr.6: 47579541-47579541 on Build GRCh38
rs202236302 Chr.6: 47579459-47579459 on Build GRCh38
rs147188917 Chr.6: 47579415-47579415 on Build GRCh38
rs57280259 Chr.6: 47579348-47579349 on Build GRCh38
rs151172462 Chr.6: 47579447-47579447 on Build GRCh38
rs778326186 Chr.6: 47579339-47579340 on Build GRCh38
rs67064904 Chr.6: 47579347-47579348 on Build GRCh38
rs970779081 Chr.6: 47579259-47579259 on Build GRCh38
rs773234554 Chr.6: 47579357-47579357 on Build GRCh38
rs548872105 Chr.6: 47579220-47579220 on Build GRCh38
rs921601729 Chr.6: 47579423-47579423 on Build GRCh38
rs922615651 Chr.6: 47579199-47579199 on Build GRCh38
rs200545750 Chr.6: 47579508-47579508 on Build GRCh38
rs185936527 Chr.6: 47579496-47579496 on Build GRCh38
rs761792087 Chr.6: 47579354-47579354 on Build GRCh38
rs755425767 Chr.6: 47579500-47579501 on Build GRCh38
rs771076145 Chr.6: 47579491-47579491 on Build GRCh38
rs756568913 Chr.6: 47579345-47579346 on Build GRCh38
rs749955608 Chr.6: 47579414-47579414 on Build GRCh38
rs533078001 Chr.6: 47579128-47579128 on Build GRCh38
rs931626305 Chr.6: 47579448-47579448 on Build GRCh38
rs760223406 Chr.6: 47579537-47579537 on Build GRCh38
rs541244683 Chr.6: 47579540-47579540 on Build GRCh38
rs1021437195 Chr.6: 47579163-47579163 on Build GRCh38
rs778875298 Chr.6: 47579428-47579430 on Build GRCh38
rs572425310 Chr.6: 47579402-47579402 on Build GRCh38
rs762662122 Chr.6: 47579566-47579566 on Build GRCh38
rs9395281 Chr.6: 47579290-47579290 on Build GRCh38
rs762874586 Chr.6: 47579489-47579489 on Build GRCh38
rs959368959 Chr.6: 47579172-47579172 on Build GRCh38
rs140188898 Chr.6: 47579473-47579473 on Build GRCh38
rs918048154 Chr.6: 47579239-47579239 on Build GRCh38
rs539798344 Chr.6: 47579335-47579335 on Build GRCh38
rs768016875 Chr.6: 47579382-47579382 on Build GRCh38
rs536922417 Chr.6: 47579192-47579192 on Build GRCh38
rs370145728 Chr.6: 47579531-47579531 on Build GRCh38
rs532854337 Chr.6: 47579590-47579590 on Build GRCh38
rs543532433 Chr.6: 47579417-47579417 on Build GRCh38
rs114590237 Chr.6: 47579182-47579182 on Build GRCh38
rs1010777901 Chr.6: 47579158-47579158 on Build GRCh38
rs773216411 Chr.6: 47579479-47579479 on Build GRCh38
rs779504309 Chr.6: 47579424-47579424 on Build GRCh38
rs377756180 Chr.6: 47579449-47579449 on Build GRCh38
rs1000096046 Chr.6: 47579138-47579138 on Build GRCh38
rs763227419 Chr.6: 47579366-47579366 on Build GRCh38
rs748038105 Chr.6: 47579228-47579228 on Build GRCh38
rs906213854 Chr.6: 47579375-47579375 on Build GRCh38
rs868579791 Chr.6: 47579568-47579568 on Build GRCh38
rs937709961 Chr.6: 47579378-47579378 on Build GRCh38
rs776508783 Chr.6: 47579352-47579352 on Build GRCh38
rs559932679 Chr.6: 47579542-47579542 on Build GRCh38
rs75176322 Chr.6: 47579349-47579349 on Build GRCh38
rs551073783 Chr.6: 47579133-47579133 on Build GRCh38
rs772699707 Chr.6: 47579501-47579501 on Build GRCh38
rs1041594438 Chr.6: 47579487-47579487 on Build GRCh38
rs772078303 Chr.6: 47579229-47579229 on Build GRCh38
rs536243596 Chr.6: 47579262-47579267 on Build GRCh38
rs34735056 Chr.6: 47579334-47579335 on Build GRCh38
rs1029635038 Chr.6: 47579555-47579555 on Build GRCh38
rs574314971 Chr.6: 47579521-47579521 on Build GRCh38
rs759941391 Chr.6: 47579377-47579377 on Build GRCh38
rs766456040 Chr.6: 47579369-47579369 on Build GRCh38
rs1032772397 Chr.6: 47579140-47579140 on Build GRCh38
rs954019314 Chr.6: 47579594-47579594 on Build GRCh38
rs776063896 Chr.6: 47579530-47579530 on Build GRCh38
rs768441101 Chr.6: 47579351-47579351 on Build GRCh38
rs374532164 Chr.6: 47579446-47579446 on Build GRCh38
rs968649349 Chr.6: 47579135-47579135 on Build GRCh38
rs567085196 Chr.6: 47579274-47579274 on Build GRCh38
rs78311912 Chr.6: 47579350-47579350 on Build GRCh38

View Full Product Details