Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49785951-49786447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGK2
Entrez Gene ID
5232
Gene Name
phosphoglycerate kinase 2
Gene Aliases
HEL-S-272, PGKB, PGKPS, dJ417L20.2
UniGene
Hs.367727
Gene Chromosome Location
Chr.6: 49785651-49787340 on Build GRCh38
Transcripts
NM_138733.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3830722 c.1072G>T p.E358* Chr.6: 49786116-49786116 on Build GRCh38
3628312 c.778G>A p.E260K Chr.6: 49786410-49786410 on Build GRCh38
5042060 c.1237G>A p.A413T Chr.6: 49785951-49785951 on Build GRCh38
3628308 c.925C>T p.P309S Chr.6: 49786263-49786263 on Build GRCh38
4944375 c.1120G>C p.G374R Chr.6: 49786068-49786068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199822863 Chr.6: 49786043-49786043 on Build GRCh38
rs542531069 Chr.6: 49785994-49785994 on Build GRCh38
rs768181677 Chr.6: 49786170-49786170 on Build GRCh38
rs552379127 Chr.6: 49786303-49786303 on Build GRCh38
rs753580221 Chr.6: 49786073-49786073 on Build GRCh38
rs780200740 Chr.6: 49786098-49786098 on Build GRCh38
rs201535868 Chr.6: 49786169-49786169 on Build GRCh38
rs187805174 Chr.6: 49785993-49785993 on Build GRCh38
rs1017880499 Chr.6: 49786083-49786083 on Build GRCh38
rs779203149 Chr.6: 49786235-49786235 on Build GRCh38
rs773628964 Chr.6: 49786252-49786252 on Build GRCh38
rs775325509 Chr.6: 49786402-49786402 on Build GRCh38
rs776872781 Chr.6: 49786021-49786021 on Build GRCh38
rs747670363 Chr.6: 49786257-49786257 on Build GRCh38
rs757119246 Chr.6: 49785992-49785992 on Build GRCh38
rs770639068 Chr.6: 49786392-49786392 on Build GRCh38
rs759304137 Chr.6: 49786395-49786395 on Build GRCh38
rs750122364 Chr.6: 49786097-49786097 on Build GRCh38
rs761327233 Chr.6: 49786074-49786074 on Build GRCh38
rs779256225 Chr.6: 49786345-49786345 on Build GRCh38
rs192939731 Chr.6: 49786226-49786226 on Build GRCh38
rs199596449 Chr.6: 49786000-49786000 on Build GRCh38
rs778202087 Chr.6: 49786137-49786137 on Build GRCh38
rs564293391 Chr.6: 49786174-49786174 on Build GRCh38
rs774299538 Chr.6: 49786393-49786393 on Build GRCh38
rs754914944 Chr.6: 49786190-49786190 on Build GRCh38
rs770242126 Chr.6: 49786033-49786033 on Build GRCh38
rs377284913 Chr.6: 49786023-49786023 on Build GRCh38
rs200172498 Chr.6: 49786227-49786227 on Build GRCh38
rs766299030 Chr.6: 49786302-49786302 on Build GRCh38
rs774900961 Chr.6: 49786155-49786155 on Build GRCh38
rs775979768 Chr.6: 49786182-49786186 on Build GRCh38
rs754104374 Chr.6: 49786329-49786329 on Build GRCh38
rs948120131 Chr.6: 49786319-49786319 on Build GRCh38
rs371825744 Chr.6: 49786039-49786039 on Build GRCh38
rs952143615 Chr.6: 49786370-49786370 on Build GRCh38
rs771269272 Chr.6: 49786153-49786153 on Build GRCh38
rs868587165 Chr.6: 49786093-49786093 on Build GRCh38
rs972082609 Chr.6: 49786338-49786338 on Build GRCh38
rs1039355616 Chr.6: 49786151-49786151 on Build GRCh38
rs763586767 Chr.6: 49786061-49786061 on Build GRCh38
rs530905364 Chr.6: 49786198-49786198 on Build GRCh38
rs909689488 Chr.6: 49786401-49786401 on Build GRCh38
rs1031204504 Chr.6: 49786192-49786192 on Build GRCh38
rs767297908 Chr.6: 49786304-49786304 on Build GRCh38
rs202057550 Chr.6: 49786028-49786028 on Build GRCh38
rs775967199 Chr.6: 49786171-49786171 on Build GRCh38
rs867908055 Chr.6: 49786325-49786325 on Build GRCh38
rs772798273 Chr.6: 49786376-49786376 on Build GRCh38
rs780494411 Chr.6: 49786363-49786363 on Build GRCh38
rs114365007 Chr.6: 49786128-49786128 on Build GRCh38
rs765154147 Chr.6: 49786290-49786301 on Build GRCh38
rs202168239 Chr.6: 49786407-49786407 on Build GRCh38
rs373042304 Chr.6: 49786172-49786172 on Build GRCh38
rs919364563 Chr.6: 49786352-49786352 on Build GRCh38
rs143783188 Chr.6: 49786367-49786367 on Build GRCh38
rs144977238 Chr.6: 49786308-49786308 on Build GRCh38
rs774956783 Chr.6: 49786103-49786103 on Build GRCh38
rs769343937 Chr.6: 49786271-49786271 on Build GRCh38
rs767665164 Chr.6: 49786203-49786203 on Build GRCh38
rs757434404 Chr.6: 49786333-49786333 on Build GRCh38
rs148144531 Chr.6: 49786371-49786371 on Build GRCh38
rs773398615 Chr.6: 49786036-49786036 on Build GRCh38
rs765114201 Chr.6: 49786091-49786091 on Build GRCh38
rs760232499 Chr.6: 49786047-49786047 on Build GRCh38
rs760635624 Chr.6: 49786405-49786405 on Build GRCh38
rs866103316 Chr.6: 49786344-49786344 on Build GRCh38
rs759104371 Chr.6: 49786242-49786244 on Build GRCh38
rs865897397 Chr.6: 49786116-49786116 on Build GRCh38
rs751596092 Chr.6: 49786177-49786177 on Build GRCh38
rs781109448 Chr.6: 49786126-49786126 on Build GRCh38
rs760203253 Chr.6: 49785998-49785998 on Build GRCh38
rs766774056 Chr.6: 49786412-49786412 on Build GRCh38
rs199952447 Chr.6: 49786343-49786343 on Build GRCh38
rs749593590 Chr.6: 49786147-49786147 on Build GRCh38
rs756369568 Chr.6: 49786214-49786214 on Build GRCh38
rs751538516 Chr.6: 49786102-49786102 on Build GRCh38
rs746408780 Chr.6: 49786238-49786238 on Build GRCh38
rs757655601 Chr.6: 49786228-49786228 on Build GRCh38
rs752574287 Chr.6: 49786316-49786317 on Build GRCh38
rs752651158 Chr.6: 49786207-49786207 on Build GRCh38
rs867145392 Chr.6: 49786068-49786069 on Build GRCh38
rs140705328 Chr.6: 49786359-49786359 on Build GRCh38
rs141226697 Chr.6: 49786173-49786173 on Build GRCh38
rs41273702 Chr.6: 49786013-49786013 on Build GRCh38
rs776222096 Chr.6: 49786254-49786254 on Build GRCh38
rs139041045 Chr.6: 49786324-49786324 on Build GRCh38
rs747519956 Chr.6: 49786366-49786366 on Build GRCh38
rs762673991 Chr.6: 49786299-49786299 on Build GRCh38
rs1052199909 Chr.6: 49786351-49786351 on Build GRCh38
rs1048211283 Chr.6: 49786050-49786050 on Build GRCh38
rs755149321 Chr.6: 49786112-49786112 on Build GRCh38
rs748423631 Chr.6: 49786127-49786127 on Build GRCh38
rs151215524 Chr.6: 49786309-49786309 on Build GRCh38
rs868482064 Chr.6: 49786340-49786340 on Build GRCh38
rs773039741 Chr.6: 49786297-49786297 on Build GRCh38
rs748979046 Chr.6: 49786378-49786378 on Build GRCh38
rs751994592 Chr.6: 49786070-49786070 on Build GRCh38
rs201510519 Chr.6: 49786350-49786350 on Build GRCh38
rs373420238 Chr.6: 49786396-49786396 on Build GRCh38
rs766246242 Chr.6: 49786175-49786175 on Build GRCh38
rs780287346 Chr.6: 49785999-49785999 on Build GRCh38
rs35254270 Chr.6: 49786339-49786340 on Build GRCh38
rs758373597 Chr.6: 49785998-49785998 on Build GRCh38

View Full Product Details