Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 50843751-50844251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFAP2B
Entrez Gene ID
7021
Gene Name
transcription factor AP-2 beta
Gene Aliases
AP-2B, AP2-B
UniGene
Hs.33102
Gene Chromosome Location
Chr.6: 50817692-50847619 on Build GRCh38
Transcripts
NM_003221.3 XM_011514837.2 XM_017011233.1 XM_017011234.1 XM_017011235.1

SNP Detail

SNP ID Chromosome Location
rs1023717405 Chr.6: 50843819-50843819 on Build GRCh38
rs1044421187 Chr.6: 50844109-50844109 on Build GRCh38
rs932151794 Chr.6: 50843951-50843951 on Build GRCh38
rs567202499 Chr.6: 50844214-50844214 on Build GRCh38
rs962367849 Chr.6: 50843878-50843878 on Build GRCh38
rs893674250 Chr.6: 50843920-50843920 on Build GRCh38
rs762515397 Chr.6: 50843869-50843869 on Build GRCh38
rs1027852605 Chr.6: 50844142-50844142 on Build GRCh38
rs921058731 Chr.6: 50843776-50843776 on Build GRCh38
rs986346611 Chr.6: 50844157-50844157 on Build GRCh38
rs113608318 Chr.6: 50844023-50844023 on Build GRCh38
rs1054355015 Chr.6: 50844064-50844064 on Build GRCh38
rs962054452 Chr.6: 50844130-50844130 on Build GRCh38
rs761881613 Chr.6: 50844097-50844097 on Build GRCh38
rs751151722 Chr.6: 50843942-50843942 on Build GRCh38
rs895800973 Chr.6: 50843857-50843857 on Build GRCh38
rs977183444 Chr.6: 50844173-50844173 on Build GRCh38
rs893643254 Chr.6: 50844143-50844143 on Build GRCh38
rs767211941 Chr.6: 50844024-50844024 on Build GRCh38
rs902818600 Chr.6: 50844040-50844040 on Build GRCh38
rs912036353 Chr.6: 50844162-50844162 on Build GRCh38
rs1037309324 Chr.6: 50843855-50843855 on Build GRCh38
rs927333654 Chr.6: 50844118-50844118 on Build GRCh38
rs536690291 Chr.6: 50844050-50844050 on Build GRCh38
rs986293650 Chr.6: 50843991-50843991 on Build GRCh38
rs984270229 Chr.6: 50843818-50843818 on Build GRCh38
rs944430837 Chr.6: 50844011-50844011 on Build GRCh38
rs773102982 Chr.6: 50844120-50844120 on Build GRCh38
rs935693220 Chr.6: 50844058-50844058 on Build GRCh38
rs982607607 Chr.6: 50843847-50843847 on Build GRCh38
rs1012780046 Chr.6: 50843810-50843810 on Build GRCh38
rs917515048 Chr.6: 50844071-50844071 on Build GRCh38
rs983861559 Chr.6: 50844031-50844031 on Build GRCh38
rs1041874169 Chr.6: 50844039-50844039 on Build GRCh38
rs552527973 Chr.6: 50844119-50844119 on Build GRCh38
rs768673938 Chr.6: 50843988-50843988 on Build GRCh38
rs894436024 Chr.6: 50844077-50844077 on Build GRCh38
rs1020911150 Chr.6: 50843969-50843969 on Build GRCh38
rs920870962 Chr.6: 50843919-50843919 on Build GRCh38
rs777645982 Chr.6: 50844140-50844140 on Build GRCh38
rs547301591 Chr.6: 50844134-50844134 on Build GRCh38
rs912066957 Chr.6: 50843999-50843999 on Build GRCh38
rs188736094 Chr.6: 50843833-50843833 on Build GRCh38
rs953594276 Chr.6: 50844156-50844156 on Build GRCh38
rs759417902 Chr.6: 50843953-50843953 on Build GRCh38
rs766155044 Chr.6: 50843901-50843901 on Build GRCh38
rs776626894 Chr.6: 50844082-50844082 on Build GRCh38
rs756009770 Chr.6: 50844122-50844122 on Build GRCh38
rs966622180 Chr.6: 50844164-50844164 on Build GRCh38
rs973669182 Chr.6: 50843879-50843879 on Build GRCh38
rs1009438593 Chr.6: 50844151-50844151 on Build GRCh38
rs965410934 Chr.6: 50843979-50843979 on Build GRCh38
rs905949664 Chr.6: 50843893-50843893 on Build GRCh38

View Full Product Details