Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 52405379-52405894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAQR8
Entrez Gene ID
85315
Gene Name
progestin and adipoQ receptor family member 8
Gene Aliases
C6orf33, LMPB1, MPRB
UniGene
Hs.239388
Gene Chromosome Location
Chr.6: 52362128-52407777 on Build GRCh38
Transcripts
NM_133367.4

SNP Detail

SNP ID Chromosome Location
rs144775008 Chr.6: 52405628-52405628 on Build GRCh38
rs1018682620 Chr.6: 52405678-52405678 on Build GRCh38
rs1027258361 Chr.6: 52405731-52405731 on Build GRCh38
rs755487323 Chr.6: 52405864-52405864 on Build GRCh38
rs682123 Chr.6: 52405826-52405826 on Build GRCh38
rs1018297893 Chr.6: 52405404-52405404 on Build GRCh38
rs142295765 Chr.6: 52405409-52405409 on Build GRCh38
rs1056517705 Chr.6: 52405569-52405569 on Build GRCh38
rs3062685 Chr.6: 52405749-52405749 on Build GRCh38
rs531712807 Chr.6: 52405478-52405478 on Build GRCh38
rs1034352287 Chr.6: 52405742-52405742 on Build GRCh38
rs939144658 Chr.6: 52405537-52405537 on Build GRCh38
rs983342394 Chr.6: 52405755-52405755 on Build GRCh38
rs1040166458 Chr.6: 52405514-52405514 on Build GRCh38
rs920056704 Chr.6: 52405500-52405500 on Build GRCh38
rs557903524 Chr.6: 52405618-52405618 on Build GRCh38
rs1001942349 Chr.6: 52405723-52405727 on Build GRCh38
rs951497616 Chr.6: 52405754-52405754 on Build GRCh38
rs186794124 Chr.6: 52405610-52405610 on Build GRCh38
rs1008349734 Chr.6: 52405675-52405675 on Build GRCh38
rs1036635556 Chr.6: 52405644-52405644 on Build GRCh38
rs964285384 Chr.6: 52405441-52405441 on Build GRCh38
rs150039094 Chr.6: 52405749-52405751 on Build GRCh38
rs927541765 Chr.6: 52405462-52405462 on Build GRCh38
rs922988597 Chr.6: 52405572-52405572 on Build GRCh38
rs4489154 Chr.6: 52405559-52405559 on Build GRCh38
rs913394316 Chr.6: 52405503-52405503 on Build GRCh38
rs529948349 Chr.6: 52405719-52405719 on Build GRCh38
rs373801807 Chr.6: 52405589-52405589 on Build GRCh38
rs148432475 Chr.6: 52405747-52405749 on Build GRCh38
rs552233495 Chr.6: 52405866-52405866 on Build GRCh38
rs957604807 Chr.6: 52405472-52405472 on Build GRCh38
rs145558413 Chr.6: 52405700-52405701 on Build GRCh38
rs928843273 Chr.6: 52405783-52405783 on Build GRCh38
rs990402954 Chr.6: 52405769-52405769 on Build GRCh38
rs931643982 Chr.6: 52405582-52405582 on Build GRCh38
rs887733527 Chr.6: 52405627-52405627 on Build GRCh38
rs66914305 Chr.6: 52405749-52405749 on Build GRCh38
rs999325399 Chr.6: 52405634-52405634 on Build GRCh38
rs546343364 Chr.6: 52405808-52405808 on Build GRCh38
rs897647332 Chr.6: 52405668-52405668 on Build GRCh38
rs766510286 Chr.6: 52405510-52405510 on Build GRCh38
rs566658037 Chr.6: 52405815-52405815 on Build GRCh38
rs947967774 Chr.6: 52405601-52405601 on Build GRCh38
rs974355723 Chr.6: 52405486-52405486 on Build GRCh38
rs9395789 Chr.6: 52405706-52405706 on Build GRCh38
rs1050055287 Chr.6: 52405597-52405597 on Build GRCh38
rs891184582 Chr.6: 52405661-52405661 on Build GRCh38
rs894855245 Chr.6: 52405591-52405591 on Build GRCh38
rs980095572 Chr.6: 52405434-52405434 on Build GRCh38
rs907630728 Chr.6: 52405523-52405523 on Build GRCh38
rs983198089 Chr.6: 52405515-52405515 on Build GRCh38
rs868437300 Chr.6: 52405642-52405642 on Build GRCh38
rs780274085 Chr.6: 52405746-52405747 on Build GRCh38
rs960735723 Chr.6: 52405812-52405812 on Build GRCh38

View Full Product Details