Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 52902254-52902748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSTA3
Entrez Gene ID
2940
Gene Name
glutathione S-transferase alpha 3
Gene Aliases
GSTA3-3, GTA3
UniGene
Hs.102484
Gene Chromosome Location
Chr.6: 52896639-52909799 on Build GRCh38
Transcripts
NM_000847.4 XM_006715072.3 XM_011514533.2 XM_017010794.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4689118 c.140-1G>T p.? Chr.6: 52902479-52902479 on Build GRCh38
5931835 c.248G>A p.G83E Chr.6: 52902370-52902370 on Build GRCh38
5694841 c.269C>A p.A90D Chr.6: 52902349-52902349 on Build GRCh38
3355199 c.234A>T p.K78N Chr.6: 52902384-52902384 on Build GRCh38
6676567 c.160C>T p.Q54* Chr.6: 52902458-52902458 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776616362 Chr.6: 52902342-52902342 on Build GRCh38
rs747372433 Chr.6: 52902484-52902484 on Build GRCh38
rs1008656091 Chr.6: 52902608-52902608 on Build GRCh38
rs761156747 Chr.6: 52902493-52902493 on Build GRCh38
rs766938651 Chr.6: 52902500-52902501 on Build GRCh38
rs2281594 Chr.6: 52902527-52902527 on Build GRCh38
rs571435491 Chr.6: 52902609-52902609 on Build GRCh38
rs45558636 Chr.6: 52902276-52902276 on Build GRCh38
rs377560395 Chr.6: 52902325-52902325 on Build GRCh38
rs746638972 Chr.6: 52902341-52902341 on Build GRCh38
rs777287549 Chr.6: 52902487-52902487 on Build GRCh38
rs780798684 Chr.6: 52902322-52902322 on Build GRCh38
rs890721099 Chr.6: 52902379-52902379 on Build GRCh38
rs772516567 Chr.6: 52902366-52902366 on Build GRCh38
rs776341198 Chr.6: 52902622-52902622 on Build GRCh38
rs368823085 Chr.6: 52902474-52902474 on Build GRCh38
rs920554890 Chr.6: 52902653-52902653 on Build GRCh38
rs371173696 Chr.6: 52902439-52902439 on Build GRCh38
rs769072733 Chr.6: 52902486-52902486 on Build GRCh38
rs765746856 Chr.6: 52902405-52902405 on Build GRCh38
rs916732192 Chr.6: 52902717-52902717 on Build GRCh38
rs964535651 Chr.6: 52902620-52902620 on Build GRCh38
rs374383889 Chr.6: 52902332-52902332 on Build GRCh38
rs370957665 Chr.6: 52902331-52902331 on Build GRCh38
rs754558972 Chr.6: 52902410-52902410 on Build GRCh38
rs375580219 Chr.6: 52902443-52902443 on Build GRCh38
rs778820549 Chr.6: 52902459-52902459 on Build GRCh38
rs367578958 Chr.6: 52902502-52902502 on Build GRCh38
rs143379014 Chr.6: 52902464-52902464 on Build GRCh38
rs964819176 Chr.6: 52902274-52902274 on Build GRCh38
rs147033582 Chr.6: 52902372-52902372 on Build GRCh38
rs987266527 Chr.6: 52902279-52902279 on Build GRCh38
rs994918754 Chr.6: 52902438-52902438 on Build GRCh38
rs749342161 Chr.6: 52902475-52902475 on Build GRCh38
rs1034981792 Chr.6: 52902672-52902672 on Build GRCh38
rs375524853 Chr.6: 52902447-52902447 on Build GRCh38
rs960603701 Chr.6: 52902465-52902465 on Build GRCh38
rs45487194 Chr.6: 52902673-52902673 on Build GRCh38
rs551359543 Chr.6: 52902601-52902601 on Build GRCh38
rs754813450 Chr.6: 52902320-52902320 on Build GRCh38
rs1023984802 Chr.6: 52902713-52902713 on Build GRCh38
rs934952257 Chr.6: 52902561-52902561 on Build GRCh38
rs17851798 Chr.6: 52902429-52902429 on Build GRCh38
rs907596268 Chr.6: 52902675-52902675 on Build GRCh38
rs767157163 Chr.6: 52902523-52902523 on Build GRCh38
rs759293518 Chr.6: 52902504-52902504 on Build GRCh38
rs751102513 Chr.6: 52902316-52902316 on Build GRCh38
rs757382643 Chr.6: 52902453-52902453 on Build GRCh38
rs1049093325 Chr.6: 52902666-52902666 on Build GRCh38
rs749166755 Chr.6: 52902446-52902446 on Build GRCh38
rs983303688 Chr.6: 52902286-52902286 on Build GRCh38
rs995433937 Chr.6: 52902650-52902650 on Build GRCh38
rs545328003 Chr.6: 52902282-52902282 on Build GRCh38
rs1004957064 Chr.6: 52902667-52902667 on Build GRCh38
rs761608364 Chr.6: 52902666-52902666 on Build GRCh38
rs746140421 Chr.6: 52902463-52902463 on Build GRCh38
rs371341656 Chr.6: 52902526-52902526 on Build GRCh38
rs1052661 Chr.6: 52902407-52902407 on Build GRCh38
rs943302175 Chr.6: 52902284-52902284 on Build GRCh38
rs200269918 Chr.6: 52902387-52902387 on Build GRCh38
rs201344628 Chr.6: 52902492-52902493 on Build GRCh38
rs1050743307 Chr.6: 52902588-52902588 on Build GRCh38
rs774344905 Chr.6: 52902565-52902565 on Build GRCh38
rs565494491 Chr.6: 52902371-52902371 on Build GRCh38
rs767698466 Chr.6: 52902617-52902617 on Build GRCh38
rs550514135 Chr.6: 52902715-52902715 on Build GRCh38
rs946482051 Chr.6: 52902361-52902361 on Build GRCh38
rs557115297 Chr.6: 52902707-52902707 on Build GRCh38
rs762175496 Chr.6: 52902490-52902490 on Build GRCh38
rs912273373 Chr.6: 52902602-52902602 on Build GRCh38
rs750017171 Chr.6: 52902394-52902394 on Build GRCh38
rs765942843 Chr.6: 52902493-52902493 on Build GRCh38
rs1053326756 Chr.6: 52902378-52902378 on Build GRCh38
rs770850602 Chr.6: 52902340-52902340 on Build GRCh38
rs528399054 Chr.6: 52902543-52902543 on Build GRCh38
rs930364126 Chr.6: 52902352-52902352 on Build GRCh38
rs756114913 Chr.6: 52902441-52902441 on Build GRCh38
rs41273858 Chr.6: 52902401-52902401 on Build GRCh38
rs948243066 Chr.6: 52902718-52902718 on Build GRCh38
rs779530905 Chr.6: 52902319-52902319 on Build GRCh38

View Full Product Details