Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 54160374-54160799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MLIP
Entrez Gene ID
90523
Gene Name
muscular LMNA-interacting protein
Gene Aliases
C6orf142
UniGene
Hs.449276
Gene Chromosome Location
Chr.6: 54018916-54266325 on Build GRCh38
Transcripts
NM_001281746.1 NM_001281747.1 NM_138569.2 XM_005249476.4 XM_005249478.4 XM_005249480.4 XM_006715245.3 XM_017011465.1 XM_017011466.1 XM_017011467.1 XM_017011468.1 XM_017011469.1 XM_017011470.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7377240 c.750G>T p.E250D Chr.6: 54160432-54160432 on Build GRCh38
3430762 c.848C>A p.S283Y Chr.6: 54160753-54160753 on Build GRCh38
421262 c.891C>G p.V297V Chr.6: 54160796-54160796 on Build GRCh38
7216008 c.836A>G p.H279R Chr.6: 54160741-54160741 on Build GRCh38
742499 c.775C>A p.Q259K Chr.6: 54160540-54160540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776788683 Chr.6: 54160554-54160554 on Build GRCh38
rs749201502 Chr.6: 54160417-54160417 on Build GRCh38
rs763748491 Chr.6: 54160587-54160587 on Build GRCh38
rs568665457 Chr.6: 54160766-54160766 on Build GRCh38
rs757250001 Chr.6: 54160418-54160418 on Build GRCh38
rs745690556 Chr.6: 54160430-54160430 on Build GRCh38
rs745857669 Chr.6: 54160512-54160512 on Build GRCh38
rs752316753 Chr.6: 54160492-54160492 on Build GRCh38
rs755813932 Chr.6: 54160494-54160494 on Build GRCh38
rs1054132851 Chr.6: 54160663-54160663 on Build GRCh38
rs566392958 Chr.6: 54160733-54160733 on Build GRCh38
rs371695494 Chr.6: 54160602-54160602 on Build GRCh38
rs775853360 Chr.6: 54160439-54160439 on Build GRCh38
rs750078895 Chr.6: 54160465-54160465 on Build GRCh38
rs776314154 Chr.6: 54160709-54160709 on Build GRCh38
rs949030539 Chr.6: 54160537-54160537 on Build GRCh38
rs752943566 Chr.6: 54160767-54160767 on Build GRCh38
rs937761556 Chr.6: 54160667-54160667 on Build GRCh38
rs754819019 Chr.6: 54160755-54160755 on Build GRCh38
rs762365997 Chr.6: 54160467-54160467 on Build GRCh38
rs267601078 Chr.6: 54160533-54160533 on Build GRCh38
rs766718370 Chr.6: 54160488-54160488 on Build GRCh38
rs985345177 Chr.6: 54160419-54160419 on Build GRCh38
rs757083037 Chr.6: 54160506-54160506 on Build GRCh38
rs745498657 Chr.6: 54160727-54160728 on Build GRCh38
rs778894905 Chr.6: 54160509-54160509 on Build GRCh38
rs550666996 Chr.6: 54160497-54160497 on Build GRCh38
rs777869742 Chr.6: 54160774-54160774 on Build GRCh38
rs780039732 Chr.6: 54160515-54160515 on Build GRCh38
rs78446650 Chr.6: 54160741-54160741 on Build GRCh38
rs751409465 Chr.6: 54160625-54160625 on Build GRCh38
rs140994036 Chr.6: 54160555-54160555 on Build GRCh38
rs774720954 Chr.6: 54160582-54160582 on Build GRCh38
rs779951361 Chr.6: 54160617-54160617 on Build GRCh38
rs779508970 Chr.6: 54160593-54160593 on Build GRCh38
rs141526731 Chr.6: 54160747-54160747 on Build GRCh38
rs756312908 Chr.6: 54160770-54160770 on Build GRCh38
rs778506082 Chr.6: 54160421-54160421 on Build GRCh38
rs769888460 Chr.6: 54160634-54160634 on Build GRCh38
rs748387781 Chr.6: 54160633-54160633 on Build GRCh38
rs769035997 Chr.6: 54160539-54160539 on Build GRCh38
rs779423718 Chr.6: 54160434-54160434 on Build GRCh38
rs766745173 Chr.6: 54160567-54160567 on Build GRCh38
rs78760786 Chr.6: 54160628-54160628 on Build GRCh38
rs774639482 Chr.6: 54160696-54160696 on Build GRCh38
rs765775001 Chr.6: 54160470-54160470 on Build GRCh38
rs777304909 Chr.6: 54160411-54160411 on Build GRCh38
rs966130524 Chr.6: 54160758-54160758 on Build GRCh38
rs763411165 Chr.6: 54160558-54160558 on Build GRCh38
rs771459318 Chr.6: 54160690-54160690 on Build GRCh38
rs768837959 Chr.6: 54160450-54160450 on Build GRCh38
rs773388317 Chr.6: 54160475-54160475 on Build GRCh38
rs758286367 Chr.6: 54160614-54160614 on Build GRCh38
rs763361307 Chr.6: 54160484-54160484 on Build GRCh38
rs747278207 Chr.6: 54160446-54160446 on Build GRCh38
rs753349648 Chr.6: 54160503-54160503 on Build GRCh38
rs567369130 Chr.6: 54160598-54160598 on Build GRCh38
rs781554569 Chr.6: 54160632-54160632 on Build GRCh38
rs150973985 Chr.6: 54160407-54160407 on Build GRCh38
rs368364951 Chr.6: 54160732-54160732 on Build GRCh38
rs759880846 Chr.6: 54160700-54160700 on Build GRCh38
rs764752286 Chr.6: 54160727-54160727 on Build GRCh38
rs866075290 Chr.6: 54160753-54160753 on Build GRCh38
rs546553459 Chr.6: 54160680-54160680 on Build GRCh38
rs780753892 Chr.6: 54160704-54160708 on Build GRCh38
rs770049513 Chr.6: 54160690-54160691 on Build GRCh38
rs776324543 Chr.6: 54160531-54160531 on Build GRCh38
rs76731109 Chr.6: 54160605-54160605 on Build GRCh38
rs758097014 Chr.6: 54160590-54160590 on Build GRCh38
rs761418936 Chr.6: 54160710-54160710 on Build GRCh38
rs758774286 Chr.6: 54160504-54160505 on Build GRCh38
rs200245272 Chr.6: 54160513-54160513 on Build GRCh38
rs749754700 Chr.6: 54160689-54160689 on Build GRCh38
rs368115749 Chr.6: 54160453-54160453 on Build GRCh38
rs146495445 Chr.6: 54160597-54160597 on Build GRCh38
rs375095731 Chr.6: 54160642-54160642 on Build GRCh38
rs772490608 Chr.6: 54160701-54160701 on Build GRCh38
rs569843601 Chr.6: 54160600-54160600 on Build GRCh38
rs201931180 Chr.6: 54160688-54160688 on Build GRCh38
rs577497785 Chr.6: 54160728-54160728 on Build GRCh38
rs559583404 Chr.6: 54160663-54160664 on Build GRCh38
rs917676518 Chr.6: 54160535-54160535 on Build GRCh38
rs759152223 Chr.6: 54160584-54160584 on Build GRCh38
rs753068027 Chr.6: 54160473-54160474 on Build GRCh38

View Full Product Details