Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 70791528-70792043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMAP1
Entrez Gene ID
60682
Gene Name
small ArfGAP 1
Gene Aliases
SMAP-1
UniGene
Hs.485717
Gene Chromosome Location
Chr.6: 70667771-70862015 on Build GRCh38
Transcripts
NM_001044305.2 NM_001281439.1 NM_001281440.1 NM_021940.4 XM_005248760.4 XM_011536045.2 XM_011536046.2 XM_011536047.2 XM_011536048.2 XM_017011170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3928583 c.448G>T p.V150L Chr.6: 70791722-70791722 on Build GRCh38
5632038 c.439C>T p.Q147* Chr.6: 70791713-70791713 on Build GRCh38
484320 c.468A>G p.Q156Q Chr.6: 70791742-70791742 on Build GRCh38
4493705 c.419C>T p.S140F Chr.6: 70791693-70791693 on Build GRCh38
4778796 c.479A>G p.D160G Chr.6: 70791753-70791753 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777535059 Chr.6: 70791864-70791864 on Build GRCh38
rs779458735 Chr.6: 70791767-70791767 on Build GRCh38
rs933883156 Chr.6: 70791684-70791684 on Build GRCh38
rs993731544 Chr.6: 70791838-70791838 on Build GRCh38
rs569274930 Chr.6: 70791820-70791820 on Build GRCh38
rs771095583 Chr.6: 70791771-70791772 on Build GRCh38
rs752288565 Chr.6: 70791805-70791805 on Build GRCh38
rs951215787 Chr.6: 70791563-70791563 on Build GRCh38
rs550951914 Chr.6: 70791810-70791810 on Build GRCh38
rs939810068 Chr.6: 70791595-70791595 on Build GRCh38
rs1040800917 Chr.6: 70791844-70791844 on Build GRCh38
rs558744157 Chr.6: 70791949-70791949 on Build GRCh38
rs1024810250 Chr.6: 70791735-70791735 on Build GRCh38
rs975075848 Chr.6: 70791974-70791974 on Build GRCh38
rs759505091 Chr.6: 70791702-70791702 on Build GRCh38
rs1028586540 Chr.6: 70791666-70791666 on Build GRCh38
rs534106240 Chr.6: 70791923-70791923 on Build GRCh38
rs760304917 Chr.6: 70791742-70791742 on Build GRCh38
rs933934713 Chr.6: 70792017-70792017 on Build GRCh38
rs992962999 Chr.6: 70791729-70791729 on Build GRCh38
rs774157598 Chr.6: 70791800-70791800 on Build GRCh38
rs971625599 Chr.6: 70791831-70791831 on Build GRCh38
rs532461504 Chr.6: 70791801-70791801 on Build GRCh38
rs868453275 Chr.6: 70791999-70791999 on Build GRCh38
rs45559440 Chr.6: 70791633-70791633 on Build GRCh38
rs373507240 Chr.6: 70791704-70791704 on Build GRCh38
rs189857064 Chr.6: 70791587-70791587 on Build GRCh38
rs754287171 Chr.6: 70791759-70791759 on Build GRCh38
rs970036243 Chr.6: 70791750-70791750 on Build GRCh38
rs542355845 Chr.6: 70791603-70791603 on Build GRCh38
rs758993657 Chr.6: 70791776-70791776 on Build GRCh38
rs781282742 Chr.6: 70791774-70791775 on Build GRCh38
rs770676499 Chr.6: 70791795-70791795 on Build GRCh38
rs1041631629 Chr.6: 70792018-70792018 on Build GRCh38
rs750920486 Chr.6: 70791774-70791774 on Build GRCh38
rs183157351 Chr.6: 70791698-70791698 on Build GRCh38
rs922228600 Chr.6: 70791985-70791985 on Build GRCh38
rs932317894 Chr.6: 70791868-70791868 on Build GRCh38
rs747056974 Chr.6: 70791777-70791777 on Build GRCh38
rs377558692 Chr.6: 70791778-70791778 on Build GRCh38
rs117021599 Chr.6: 70791676-70791676 on Build GRCh38
rs563413668 Chr.6: 70791919-70791919 on Build GRCh38
rs984303677 Chr.6: 70791895-70791895 on Build GRCh38
rs45495098 Chr.6: 70791861-70791861 on Build GRCh38
rs577082965 Chr.6: 70791973-70791973 on Build GRCh38
rs942328890 Chr.6: 70791619-70791619 on Build GRCh38
rs759413182 Chr.6: 70791804-70791804 on Build GRCh38
rs45540932 Chr.6: 70791791-70791791 on Build GRCh38
rs574422981 Chr.6: 70791899-70791899 on Build GRCh38
rs781249095 Chr.6: 70791779-70791779 on Build GRCh38
rs544508275 Chr.6: 70791983-70791983 on Build GRCh38
rs753580446 Chr.6: 70791746-70791746 on Build GRCh38
rs752220620 Chr.6: 70791710-70791710 on Build GRCh38
rs956389493 Chr.6: 70791723-70791723 on Build GRCh38
rs1000727289 Chr.6: 70791743-70791743 on Build GRCh38
rs770810174 Chr.6: 70791643-70791643 on Build GRCh38
rs980039476 Chr.6: 70791764-70791764 on Build GRCh38
rs1041179741 Chr.6: 70791605-70791605 on Build GRCh38
rs909590698 Chr.6: 70791609-70791609 on Build GRCh38
rs1053770758 Chr.6: 70791950-70791950 on Build GRCh38
rs867827125 Chr.6: 70791817-70791817 on Build GRCh38
rs762643035 Chr.6: 70791638-70791638 on Build GRCh38

View Full Product Details