Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75601732-75602234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SENP6
Entrez Gene ID
26054
Gene Name
SUMO1/sentrin specific peptidase 6
Gene Aliases
SSP1, SUSP1
UniGene
Hs.485784
Gene Chromosome Location
Chr.6: 75601509-75718285 on Build GRCh38
Transcripts
NM_001100409.2 NM_001304792.1 NM_015571.3 XM_006715457.3 XM_011535746.2 XM_011535747.2 XM_017010719.1 XM_017010722.1 XM_017010723.1 XM_017010724.1 XM_017010725.1

SNP Detail

SNP ID Chromosome Location
rs899018702 Chr.6: 75601845-75601845 on Build GRCh38
rs975951717 Chr.6: 75601781-75601781 on Build GRCh38
rs1019915885 Chr.6: 75601770-75601770 on Build GRCh38
rs948622435 Chr.6: 75602018-75602018 on Build GRCh38
rs1039084229 Chr.6: 75602041-75602041 on Build GRCh38
rs35178767 Chr.6: 75602017-75602017 on Build GRCh38
rs536224337 Chr.6: 75602011-75602011 on Build GRCh38
rs1050439300 Chr.6: 75601989-75601989 on Build GRCh38
rs542892511 Chr.6: 75602009-75602009 on Build GRCh38
rs960076360 Chr.6: 75602147-75602147 on Build GRCh38
rs900177678 Chr.6: 75602046-75602046 on Build GRCh38
rs935567396 Chr.6: 75601820-75601820 on Build GRCh38
rs920412779 Chr.6: 75602021-75602021 on Build GRCh38
rs938707409 Chr.6: 75601806-75601806 on Build GRCh38
rs558792142 Chr.6: 75602205-75602205 on Build GRCh38
rs1031809143 Chr.6: 75602136-75602136 on Build GRCh38
rs1017276678 Chr.6: 75601888-75601888 on Build GRCh38
rs1022611050 Chr.6: 75602081-75602081 on Build GRCh38
rs964234287 Chr.6: 75601773-75601773 on Build GRCh38
rs997492289 Chr.6: 75602047-75602047 on Build GRCh38
rs992946394 Chr.6: 75602171-75602171 on Build GRCh38
rs992755770 Chr.6: 75602010-75602010 on Build GRCh38
rs948674170 Chr.6: 75601826-75601826 on Build GRCh38
rs972001488 Chr.6: 75601753-75601753 on Build GRCh38
rs915667517 Chr.6: 75601780-75601780 on Build GRCh38
rs573193547 Chr.6: 75602089-75602089 on Build GRCh38
rs915917776 Chr.6: 75602012-75602012 on Build GRCh38
rs924382110 Chr.6: 75602096-75602096 on Build GRCh38
rs915848113 Chr.6: 75601821-75601821 on Build GRCh38
rs1029451201 Chr.6: 75601859-75601859 on Build GRCh38
rs540494828 Chr.6: 75602170-75602170 on Build GRCh38
rs991180335 Chr.6: 75601916-75601916 on Build GRCh38
rs35708564 Chr.6: 75602050-75602050 on Build GRCh38
rs1008599829 Chr.6: 75601887-75601887 on Build GRCh38
rs959937228 Chr.6: 75601907-75601907 on Build GRCh38
rs929012784 Chr.6: 75601852-75601852 on Build GRCh38
rs925442122 Chr.6: 75601936-75601936 on Build GRCh38
rs1035574000 Chr.6: 75602062-75602062 on Build GRCh38
rs1047543189 Chr.6: 75601867-75601867 on Build GRCh38
rs1037901194 Chr.6: 75601829-75601829 on Build GRCh38
rs181831097 Chr.6: 75602132-75602132 on Build GRCh38
rs1043866227 Chr.6: 75601808-75601808 on Build GRCh38
rs997124546 Chr.6: 75601915-75601915 on Build GRCh38
rs970114130 Chr.6: 75602180-75602180 on Build GRCh38
rs889152329 Chr.6: 75601994-75601994 on Build GRCh38
rs948186174 Chr.6: 75601789-75601789 on Build GRCh38
rs915094555 Chr.6: 75601775-75601775 on Build GRCh38
rs71561434 Chr.6: 75602002-75602002 on Build GRCh38
rs374087556 Chr.6: 75601941-75601941 on Build GRCh38
rs879183803 Chr.6: 75602093-75602093 on Build GRCh38
rs538401494 Chr.6: 75601896-75601896 on Build GRCh38
rs933282351 Chr.6: 75601982-75601982 on Build GRCh38
rs1016099913 Chr.6: 75601880-75601881 on Build GRCh38
rs977457256 Chr.6: 75601965-75601965 on Build GRCh38
rs1023359957 Chr.6: 75602172-75602172 on Build GRCh38
rs1056974793 Chr.6: 75601818-75601818 on Build GRCh38
rs997543746 Chr.6: 75601857-75601857 on Build GRCh38
rs1021901273 Chr.6: 75602059-75602059 on Build GRCh38
rs924601099 Chr.6: 75601799-75601799 on Build GRCh38
rs944440207 Chr.6: 75602040-75602040 on Build GRCh38
rs557010369 Chr.6: 75601931-75601931 on Build GRCh38
rs973358464 Chr.6: 75602019-75602019 on Build GRCh38
rs1052612976 Chr.6: 75601966-75601966 on Build GRCh38
rs935513700 Chr.6: 75601943-75601943 on Build GRCh38
rs890173293 Chr.6: 75602036-75602036 on Build GRCh38
rs1031756865 Chr.6: 75601970-75601970 on Build GRCh38
rs35930438 Chr.6: 75602054-75602054 on Build GRCh38
rs904696887 Chr.6: 75602077-75602077 on Build GRCh38
rs969183849 Chr.6: 75601797-75601797 on Build GRCh38
rs947154888 Chr.6: 75601930-75601930 on Build GRCh38
rs914746684 Chr.6: 75601787-75601787 on Build GRCh38
rs991625267 Chr.6: 75601778-75601778 on Build GRCh38
rs1047502373 Chr.6: 75602035-75602035 on Build GRCh38
rs901822141 Chr.6: 75601848-75601848 on Build GRCh38
rs890109752 Chr.6: 75601879-75601879 on Build GRCh38
rs889212008 Chr.6: 75601877-75601878 on Build GRCh38
rs189124093 Chr.6: 75602033-75602033 on Build GRCh38
rs968818382 Chr.6: 75602086-75602086 on Build GRCh38
rs111525008 Chr.6: 75601762-75601762 on Build GRCh38
rs977488330 Chr.6: 75601798-75601798 on Build GRCh38
rs1036107143 Chr.6: 75601820-75601833 on Build GRCh38
rs959977748 Chr.6: 75602069-75602069 on Build GRCh38
rs1022181210 Chr.6: 75601918-75601918 on Build GRCh38
rs978452759 Chr.6: 75602087-75602087 on Build GRCh38
rs988274581 Chr.6: 75602168-75602168 on Build GRCh38

View Full Product Details