Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75840876-75841384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO6
Entrez Gene ID
4646
Gene Name
myosin VI
Gene Aliases
DFNA22, DFNB37
UniGene
Hs.149387
Gene Chromosome Location
Chr.6: 75749176-75919537 on Build GRCh38
Transcripts
NM_001300899.1 NM_004999.3 XM_005248719.3 XM_005248720.3 XM_005248721.3 XM_005248722.3 XM_005248724.3 XM_005248726.3 XM_017010899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5987546 c.712C>G p.Q238E Chr.6: 75841274-75841274 on Build GRCh38
6495642 c.816+1G>A p.? Chr.6: 75841379-75841379 on Build GRCh38
5097588 c.707G>A p.C236Y Chr.6: 75841269-75841269 on Build GRCh38
6107708 c.781G>A p.E261K Chr.6: 75841343-75841343 on Build GRCh38
162750 c.793T>G p.L265V Chr.6: 75841355-75841355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776814111 Chr.6: 75841249-75841249 on Build GRCh38
rs121912560 Chr.6: 75841299-75841299 on Build GRCh38
rs898440044 Chr.6: 75841134-75841134 on Build GRCh38
rs755566058 Chr.6: 75841303-75841303 on Build GRCh38
rs572338848 Chr.6: 75840921-75840921 on Build GRCh38
rs751465266 Chr.6: 75841171-75841171 on Build GRCh38
rs914864183 Chr.6: 75840964-75840964 on Build GRCh38
rs775574293 Chr.6: 75841237-75841237 on Build GRCh38
rs546262267 Chr.6: 75841011-75841011 on Build GRCh38
rs542523266 Chr.6: 75840945-75840945 on Build GRCh38
rs777614964 Chr.6: 75841317-75841317 on Build GRCh38
rs955410532 Chr.6: 75841258-75841258 on Build GRCh38
rs1013903021 Chr.6: 75840958-75840958 on Build GRCh38
rs12203461 Chr.6: 75841013-75841013 on Build GRCh38
rs766119690 Chr.6: 75841250-75841250 on Build GRCh38
rs760669746 Chr.6: 75841245-75841245 on Build GRCh38
rs755724636 Chr.6: 75841189-75841189 on Build GRCh38
rs753508452 Chr.6: 75841321-75841321 on Build GRCh38
rs746366118 Chr.6: 75841347-75841347 on Build GRCh38
rs575016261 Chr.6: 75840907-75840907 on Build GRCh38
rs930430931 Chr.6: 75841241-75841241 on Build GRCh38
rs34674679 Chr.6: 75841123-75841124 on Build GRCh38
rs752318101 Chr.6: 75841296-75841296 on Build GRCh38
rs147722766 Chr.6: 75841091-75841091 on Build GRCh38
rs529148986 Chr.6: 75841193-75841193 on Build GRCh38
rs375645908 Chr.6: 75841207-75841207 on Build GRCh38
rs770022805 Chr.6: 75841059-75841059 on Build GRCh38
rs752301635 Chr.6: 75841184-75841184 on Build GRCh38
rs766937820 Chr.6: 75841281-75841281 on Build GRCh38
rs561937228 Chr.6: 75841158-75841158 on Build GRCh38
rs1056192445 Chr.6: 75841023-75841023 on Build GRCh38
rs757719710 Chr.6: 75841322-75841322 on Build GRCh38
rs754475205 Chr.6: 75841180-75841180 on Build GRCh38
rs759456713 Chr.6: 75841278-75841278 on Build GRCh38
rs975226295 Chr.6: 75841152-75841152 on Build GRCh38
rs1038683374 Chr.6: 75841122-75841122 on Build GRCh38
rs772058658 Chr.6: 75841223-75841223 on Build GRCh38
rs112510570 Chr.6: 75840991-75840991 on Build GRCh38
rs531863014 Chr.6: 75841048-75841048 on Build GRCh38
rs966696313 Chr.6: 75841085-75841085 on Build GRCh38
rs751472354 Chr.6: 75841276-75841276 on Build GRCh38
rs756122330 Chr.6: 75841082-75841082 on Build GRCh38
rs780701022 Chr.6: 75841183-75841183 on Build GRCh38
rs111480295 Chr.6: 75840998-75840998 on Build GRCh38
rs768928382 Chr.6: 75841246-75841246 on Build GRCh38
rs746338729 Chr.6: 75841212-75841212 on Build GRCh38
rs370811316 Chr.6: 75841122-75841123 on Build GRCh38
rs956239103 Chr.6: 75840914-75840914 on Build GRCh38
rs989694064 Chr.6: 75841015-75841015 on Build GRCh38
rs771399392 Chr.6: 75841200-75841200 on Build GRCh38
rs749041026 Chr.6: 75841126-75841126 on Build GRCh38
rs3839374 Chr.6: 75841121-75841122 on Build GRCh38
rs780471577 Chr.6: 75841205-75841206 on Build GRCh38
rs779365282 Chr.6: 75841337-75841337 on Build GRCh38
rs990400397 Chr.6: 75840923-75840923 on Build GRCh38
rs868163831 Chr.6: 75841045-75841045 on Build GRCh38
rs750292374 Chr.6: 75841079-75841079 on Build GRCh38

View Full Product Details